Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 44861186-44861695 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTTG1IP
Entrez Gene ID
754
Gene Name
pituitary tumor-transforming 1 interacting protein
Gene Aliases
C21orf1, C21orf3, PBF
UniGene
Hs.474010
Gene Chromosome Location
Chr.21: 44849585-44873903 on Build GRCh38
Transcripts
NM_001286822.1 NM_004339.3 NR_104597.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5858764 c.211G>A p.V71I Chr.21: 44861229-44861229 on Build GRCh38
6161568 c.176_177GG>TC p.W59F Chr.21: 44861263-44861264 on Build GRCh38
2821416 c.216A>G p.T72T Chr.21: 44861224-44861224 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs777140471 Chr.21: 44861302-44861302 on Build GRCh38
rs373341271 Chr.21: 44861422-44861422 on Build GRCh38
rs778849261 Chr.21: 44861278-44861278 on Build GRCh38
rs547651507 Chr.21: 44861458-44861458 on Build GRCh38
rs751093743 Chr.21: 44861241-44861241 on Build GRCh38
rs181968469 Chr.21: 44861357-44861357 on Build GRCh38
rs1050457782 Chr.21: 44861578-44861578 on Build GRCh38
rs1031857801 Chr.21: 44861570-44861570 on Build GRCh38
rs144668992 Chr.21: 44861255-44861255 on Build GRCh38
rs778834109 Chr.21: 44861522-44861522 on Build GRCh38
rs1017735492 Chr.21: 44861223-44861223 on Build GRCh38
rs956328809 Chr.21: 44861317-44861317 on Build GRCh38
rs200698166 Chr.21: 44861220-44861220 on Build GRCh38
rs1045846874 Chr.21: 44861626-44861626 on Build GRCh38
rs188877011 Chr.21: 44861210-44861210 on Build GRCh38
rs200392558 Chr.21: 44861549-44861550 on Build GRCh38
rs938412549 Chr.21: 44861416-44861416 on Build GRCh38
rs970282197 Chr.21: 44861631-44861631 on Build GRCh38
rs11537961 Chr.21: 44861242-44861242 on Build GRCh38
rs776897199 Chr.21: 44861650-44861650 on Build GRCh38
rs1001849106 Chr.21: 44861424-44861424 on Build GRCh38
rs150849428 Chr.21: 44861499-44861499 on Build GRCh38
rs113180682 Chr.21: 44861669-44861669 on Build GRCh38
rs142345359 Chr.21: 44861227-44861227 on Build GRCh38
rs1010051813 Chr.21: 44861400-44861400 on Build GRCh38
rs572176295 Chr.21: 44861228-44861228 on Build GRCh38
rs776852385 Chr.21: 44861286-44861287 on Build GRCh38
rs745862954 Chr.21: 44861541-44861541 on Build GRCh38
rs755949360 Chr.21: 44861264-44861264 on Build GRCh38
rs755245744 Chr.21: 44861272-44861272 on Build GRCh38
rs770767563 Chr.21: 44861429-44861429 on Build GRCh38
rs186136293 Chr.21: 44861437-44861437 on Build GRCh38
rs925480435 Chr.21: 44861316-44861316 on Build GRCh38
rs951674917 Chr.21: 44861470-44861470 on Build GRCh38
rs755774187 Chr.21: 44861288-44861288 on Build GRCh38
rs1014531336 Chr.21: 44861670-44861670 on Build GRCh38
rs934224225 Chr.21: 44861330-44861330 on Build GRCh38
rs777506076 Chr.21: 44861353-44861353 on Build GRCh38
rs922754588 Chr.21: 44861503-44861503 on Build GRCh38
rs1045954455 Chr.21: 44861402-44861402 on Build GRCh38
rs772696653 Chr.21: 44861218-44861218 on Build GRCh38
rs969660218 Chr.21: 44861358-44861358 on Build GRCh38
rs235276 Chr.21: 44861399-44861399 on Build GRCh38
rs763445809 Chr.21: 44861240-44861240 on Build GRCh38
rs891919795 Chr.21: 44861590-44861590 on Build GRCh38
rs761164176 Chr.21: 44861221-44861221 on Build GRCh38
rs1001378266 Chr.21: 44861634-44861634 on Build GRCh38
rs369693004 Chr.21: 44861296-44861296 on Build GRCh38
rs575726615 Chr.21: 44861363-44861363 on Build GRCh38
rs914240952 Chr.21: 44861343-44861343 on Build GRCh38
rs1054512392 Chr.21: 44861391-44861391 on Build GRCh38
rs779842845 Chr.21: 44861268-44861268 on Build GRCh38
rs559719844 Chr.21: 44861490-44861490 on Build GRCh38
rs913261795 Chr.21: 44861591-44861591 on Build GRCh38
rs1034308259 Chr.21: 44861456-44861456 on Build GRCh38
rs771383132 Chr.21: 44861298-44861298 on Build GRCh38
rs935991930 Chr.21: 44861380-44861380 on Build GRCh38
rs749467135 Chr.21: 44861271-44861271 on Build GRCh38
rs533935724 Chr.21: 44861325-44861325 on Build GRCh38
rs565747192 Chr.21: 44861668-44861668 on Build GRCh38
rs778474487 Chr.21: 44861292-44861292 on Build GRCh38
rs997977758 Chr.21: 44861551-44861551 on Build GRCh38
rs537520081 Chr.21: 44861559-44861559 on Build GRCh38
rs979731648 Chr.21: 44861393-44861394 on Build GRCh38
rs772430025 Chr.21: 44861289-44861289 on Build GRCh38
rs1005849579 Chr.21: 44861498-44861498 on Build GRCh38
rs570178779 Chr.21: 44861540-44861540 on Build GRCh38
rs540961273 Chr.21: 44861239-44861239 on Build GRCh38
rs1036896079 Chr.21: 44861455-44861455 on Build GRCh38
rs567627131 Chr.21: 44861615-44861615 on Build GRCh38
rs549430542 Chr.21: 44861571-44861571 on Build GRCh38
rs1039033346 Chr.21: 44861515-44861515 on Build GRCh38
rs955607007 Chr.21: 44861508-44861508 on Build GRCh38
rs907026476 Chr.21: 44861630-44861630 on Build GRCh38
rs931266403 Chr.21: 44861500-44861501 on Build GRCh38
rs113683225 Chr.21: 44861647-44861647 on Build GRCh38
rs535767775 Chr.21: 44861646-44861646 on Build GRCh38
rs961787324 Chr.21: 44861500-44861500 on Build GRCh38
rs775235646 Chr.21: 44861231-44861231 on Build GRCh38
rs551672503 Chr.21: 44861501-44861501 on Build GRCh38
rs986077433 Chr.21: 44861514-44861514 on Build GRCh38
rs1054481390 Chr.21: 44861542-44861542 on Build GRCh38
rs757127661 Chr.21: 44861254-44861254 on Build GRCh38

View Full Product Details