Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 29730747-29731253 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CABP7
Entrez Gene ID
164633
Gene Name
calcium binding protein 7
Gene Aliases
CALN2
UniGene
Hs.643608
Gene Chromosome Location
Chr.22: 29720090-29731833 on Build GRCh38
Transcripts
NM_182527.2 NM_001003692.1 NM_001318129.1 NM_019103.2

Gene Symbol
ZMAT5
Entrez Gene ID
55954
Gene Name
zinc finger matrin-type 5
Gene Aliases
SNRNP20, U11/U12-20K, ZC3H19
UniGene
Hs.713647
Gene Chromosome Location
Chr.22: 29730956-29767011 on Build GRCh38
Transcripts
NM_182527.2 NM_001003692.1 NM_001318129.1 NM_019103.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5028793 c.493C>A p.P165T Chr.22: 29731245-29731245 on Build GRCh38
5028794 c.493C>A p.P165T Chr.22: 29731245-29731245 on Build GRCh38
6161999 c.489G>A p.L163L Chr.22: 29731249-29731249 on Build GRCh38
6162000 c.489G>A p.L163L Chr.22: 29731249-29731249 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs373784620 Chr.22: 29731199-29731199 on Build GRCh38
rs3180412 Chr.22: 29730957-29730957 on Build GRCh38
rs893070873 Chr.22: 29730806-29730806 on Build GRCh38
rs947326735 Chr.22: 29730959-29730959 on Build GRCh38
rs377024133 Chr.22: 29730859-29730859 on Build GRCh38
rs755316473 Chr.22: 29731217-29731217 on Build GRCh38
rs928371009 Chr.22: 29730928-29730928 on Build GRCh38
rs578214349 Chr.22: 29730838-29730838 on Build GRCh38
rs746709714 Chr.22: 29731158-29731158 on Build GRCh38
rs564157656 Chr.22: 29731017-29731017 on Build GRCh38
rs561882076 Chr.22: 29731070-29731070 on Build GRCh38
rs535319312 Chr.22: 29730852-29730852 on Build GRCh38
rs909693080 Chr.22: 29730792-29730792 on Build GRCh38
rs1011878612 Chr.22: 29730818-29730818 on Build GRCh38
rs981186752 Chr.22: 29730918-29730918 on Build GRCh38
rs6006230 Chr.22: 29731146-29731146 on Build GRCh38
rs914187889 Chr.22: 29730958-29730958 on Build GRCh38
rs776032024 Chr.22: 29731200-29731201 on Build GRCh38
rs767717505 Chr.22: 29731205-29731205 on Build GRCh38
rs773751326 Chr.22: 29730771-29730771 on Build GRCh38
rs750516038 Chr.22: 29731207-29731207 on Build GRCh38
rs962209340 Chr.22: 29731093-29731093 on Build GRCh38
rs756709454 Chr.22: 29731211-29731211 on Build GRCh38
rs563982480 Chr.22: 29731157-29731157 on Build GRCh38
rs762088255 Chr.22: 29731203-29731203 on Build GRCh38
rs540115769 Chr.22: 29731055-29731055 on Build GRCh38
rs1028999379 Chr.22: 29731058-29731058 on Build GRCh38
rs764426459 Chr.22: 29731196-29731196 on Build GRCh38
rs754274299 Chr.22: 29731215-29731215 on Build GRCh38
rs776848530 Chr.22: 29731230-29731230 on Build GRCh38
rs199783629 Chr.22: 29731212-29731212 on Build GRCh38
rs962178361 Chr.22: 29730915-29730915 on Build GRCh38
rs545766435 Chr.22: 29731007-29731007 on Build GRCh38
rs780684995 Chr.22: 29730942-29730942 on Build GRCh38
rs191059008 Chr.22: 29730817-29730817 on Build GRCh38
rs985410566 Chr.22: 29730890-29730890 on Build GRCh38
rs1041559328 Chr.22: 29730821-29730821 on Build GRCh38
rs958489025 Chr.22: 29731130-29731130 on Build GRCh38
rs770623089 Chr.22: 29731147-29731147 on Build GRCh38
rs981593246 Chr.22: 29731119-29731119 on Build GRCh38
rs558356305 Chr.22: 29730776-29730776 on Build GRCh38
rs139936953 Chr.22: 29730879-29730879 on Build GRCh38
rs529076576 Chr.22: 29731138-29731138 on Build GRCh38
rs901360329 Chr.22: 29731222-29731222 on Build GRCh38
rs542095847 Chr.22: 29730892-29730892 on Build GRCh38
rs1015117116 Chr.22: 29731091-29731091 on Build GRCh38
rs116658301 Chr.22: 29730937-29730937 on Build GRCh38
rs983972538 Chr.22: 29731074-29731074 on Build GRCh38
rs1016811442 Chr.22: 29731083-29731083 on Build GRCh38
rs182886551 Chr.22: 29730847-29730847 on Build GRCh38
rs1018897208 Chr.22: 29731001-29731001 on Build GRCh38
rs1019029049 Chr.22: 29730839-29730839 on Build GRCh38
rs1006508869 Chr.22: 29731087-29731087 on Build GRCh38
rs528502671 Chr.22: 29731054-29731054 on Build GRCh38
rs751175453 Chr.22: 29730870-29730870 on Build GRCh38
rs536737405 Chr.22: 29730805-29730805 on Build GRCh38
rs776559783 Chr.22: 29731184-29731184 on Build GRCh38
rs749053253 Chr.22: 29731129-29731129 on Build GRCh38
rs768458582 Chr.22: 29731145-29731145 on Build GRCh38
rs991332586 Chr.22: 29731121-29731121 on Build GRCh38
rs935546990 Chr.22: 29730904-29730904 on Build GRCh38
rs61737046 Chr.22: 29731228-29731228 on Build GRCh38
rs903051599 Chr.22: 29730997-29730997 on Build GRCh38
rs769215451 Chr.22: 29731199-29731200 on Build GRCh38
rs779873674 Chr.22: 29731068-29731068 on Build GRCh38
rs562574386 Chr.22: 29731193-29731193 on Build GRCh38
rs745328470 Chr.22: 29731187-29731188 on Build GRCh38
rs111846036 Chr.22: 29730822-29730822 on Build GRCh38
rs142160138 Chr.22: 29730986-29730986 on Build GRCh38
rs747806140 Chr.22: 29731223-29731223 on Build GRCh38
rs9614043 Chr.22: 29730910-29730910 on Build GRCh38

View Full Product Details