Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 35741304-35741829 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RBFOX2
Entrez Gene ID
23543
Gene Name
RNA binding protein, fox-1 homolog 2
Gene Aliases
FOX2, Fox-2, HNRBP2, HRNBP2, RBM9, RTA, dJ106I20.3, fxh
UniGene
Hs.282998
Gene Chromosome Location
Chr.22: 35738736-36028892 on Build GRCh38
Transcripts
NM_001031695.2 NM_001082576.1 NM_001082577.1 NM_001082578.1 NM_001082579.1 NM_014309.2 XM_005261428.2 XM_005261429.2 XM_005261430.2 XM_005261431.2 XM_005261432.2 XM_005261433.2 XM_005261435.1 XM_005261437.1 XM_006724185.2 XM_006724186.2 XM_006724187.2 XM_006724188.2 XM_006724189.3 XM_006724190.1 XM_006724191.1 XM_006724192.2 XM_006724194.2 XM_017028686.1 XM_017028687.1 XM_017028688.1 XM_017028689.1 XM_017028690.1 XM_017028691.1 XM_017028692.1 XM_017028693.1 XM_017028694.1 XM_017028695.1 XM_017028696.1 XM_017028697.1 XM_017028698.1

SNP Detail

SNP ID Chromosome Location
rs188061884 Chr.22: 35741606-35741606 on Build GRCh38
rs529527074 Chr.22: 35741513-35741513 on Build GRCh38
rs190892855 Chr.22: 35741411-35741411 on Build GRCh38
rs1045974092 Chr.22: 35741376-35741376 on Build GRCh38
rs113159456 Chr.22: 35741771-35741771 on Build GRCh38
rs932814625 Chr.22: 35741781-35741781 on Build GRCh38
rs928878249 Chr.22: 35741378-35741378 on Build GRCh38
rs1023056612 Chr.22: 35741616-35741616 on Build GRCh38
rs766994301 Chr.22: 35741761-35741761 on Build GRCh38
rs544372078 Chr.22: 35741544-35741544 on Build GRCh38
rs564316384 Chr.22: 35741554-35741554 on Build GRCh38
rs182183370 Chr.22: 35741481-35741481 on Build GRCh38
rs944041410 Chr.22: 35741699-35741699 on Build GRCh38
rs981433410 Chr.22: 35741638-35741638 on Build GRCh38
rs1023305850 Chr.22: 35741515-35741515 on Build GRCh38
rs1021358797 Chr.22: 35741480-35741480 on Build GRCh38
rs899960457 Chr.22: 35741475-35741475 on Build GRCh38
rs546092124 Chr.22: 35741347-35741347 on Build GRCh38
rs572918295 Chr.22: 35741400-35741400 on Build GRCh38
rs969883641 Chr.22: 35741527-35741527 on Build GRCh38
rs989090617 Chr.22: 35741734-35741734 on Build GRCh38
rs889526863 Chr.22: 35741795-35741795 on Build GRCh38
rs746839830 Chr.22: 35741452-35741452 on Build GRCh38
rs915884296 Chr.22: 35741342-35741342 on Build GRCh38
rs762062748 Chr.22: 35741485-35741485 on Build GRCh38
rs970168688 Chr.22: 35741633-35741633 on Build GRCh38
rs1041726106 Chr.22: 35741459-35741459 on Build GRCh38
rs982856857 Chr.22: 35741685-35741686 on Build GRCh38
rs769831675 Chr.22: 35741493-35741493 on Build GRCh38
rs779901901 Chr.22: 35741406-35741406 on Build GRCh38
rs947559972 Chr.22: 35741750-35741750 on Build GRCh38
rs1053683238 Chr.22: 35741437-35741437 on Build GRCh38
rs191100074 Chr.22: 35741643-35741643 on Build GRCh38
rs914692914 Chr.22: 35741745-35741745 on Build GRCh38
rs947110207 Chr.22: 35741453-35741453 on Build GRCh38
rs994123719 Chr.22: 35741496-35741496 on Build GRCh38
rs1049477068 Chr.22: 35741474-35741474 on Build GRCh38
rs151061359 Chr.22: 35741371-35741371 on Build GRCh38
rs529359407 Chr.22: 35741796-35741796 on Build GRCh38
rs772682645 Chr.22: 35741723-35741723 on Build GRCh38
rs766741087 Chr.22: 35741396-35741397 on Build GRCh38
rs1027208478 Chr.22: 35741542-35741542 on Build GRCh38
rs763066840 Chr.22: 35741752-35741752 on Build GRCh38
rs919985075 Chr.22: 35741599-35741599 on Build GRCh38
rs775363208 Chr.22: 35741624-35741624 on Build GRCh38
rs768448693 Chr.22: 35741479-35741479 on Build GRCh38
rs908541426 Chr.22: 35741690-35741691 on Build GRCh38
rs5755937 Chr.22: 35741484-35741484 on Build GRCh38
rs533116513 Chr.22: 35741570-35741570 on Build GRCh38
rs934889021 Chr.22: 35741645-35741645 on Build GRCh38

View Full Product Details