Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 42551310-42551857 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SEC22C
Entrez Gene ID
9117
Gene Name
SEC22 homolog C, vesicle trafficking protein
Gene Aliases
SEC22L3
UniGene
Hs.445892
Gene Chromosome Location
Chr.3: 42547967-42601080 on Build GRCh38
Transcripts
NM_001201572.1 NM_001201584.1 NM_004206.3 NM_032970.3 XM_005265533.1 XM_005265534.1 XM_005265535.1 XM_011534212.1 XM_017007438.1 XM_017007439.1

SNP Detail

SNP ID Chromosome Location
rs561888963 Chr.3: 42551354-42551354 on Build GRCh38
rs138539315 Chr.3: 42551758-42551758 on Build GRCh38
rs754818046 Chr.3: 42551538-42551538 on Build GRCh38
rs1036832031 Chr.3: 42551608-42551608 on Build GRCh38
rs944613717 Chr.3: 42551628-42551628 on Build GRCh38
rs998482252 Chr.3: 42551812-42551812 on Build GRCh38
rs973548644 Chr.3: 42551811-42551811 on Build GRCh38
rs527562784 Chr.3: 42551720-42551720 on Build GRCh38
rs561987180 Chr.3: 42551673-42551673 on Build GRCh38
rs773403243 Chr.3: 42551540-42551540 on Build GRCh38
rs903032733 Chr.3: 42551646-42551646 on Build GRCh38
rs777049125 Chr.3: 42551684-42551685 on Build GRCh38
rs748303482 Chr.3: 42551524-42551524 on Build GRCh38
rs1012911756 Chr.3: 42551702-42551702 on Build GRCh38
rs932638204 Chr.3: 42551567-42551567 on Build GRCh38
rs342538 Chr.3: 42551410-42551410 on Build GRCh38
rs971537455 Chr.3: 42551467-42551467 on Build GRCh38
rs1024506504 Chr.3: 42551757-42551757 on Build GRCh38
rs1024537807 Chr.3: 42551461-42551461 on Build GRCh38
rs545880030 Chr.3: 42551512-42551512 on Build GRCh38
rs1036934809 Chr.3: 42551351-42551351 on Build GRCh38
rs342537 Chr.3: 42551382-42551382 on Build GRCh38
rs1043387986 Chr.3: 42551761-42551761 on Build GRCh38
rs570393965 Chr.3: 42551776-42551780 on Build GRCh38
rs181907056 Chr.3: 42551807-42551807 on Build GRCh38
rs753582954 Chr.3: 42551429-42551429 on Build GRCh38
rs1000287438 Chr.3: 42551378-42551378 on Build GRCh38
rs1004426660 Chr.3: 42551797-42551797 on Build GRCh38
rs921314039 Chr.3: 42551539-42551539 on Build GRCh38
rs1041533625 Chr.3: 42551640-42551640 on Build GRCh38
rs879034885 Chr.3: 42551647-42551647 on Build GRCh38
rs1016154315 Chr.3: 42551802-42551802 on Build GRCh38
rs911845510 Chr.3: 42551597-42551597 on Build GRCh38
rs894447099 Chr.3: 42551684-42551684 on Build GRCh38
rs576571476 Chr.3: 42551502-42551502 on Build GRCh38
rs992137476 Chr.3: 42551457-42551457 on Build GRCh38
rs931696759 Chr.3: 42551565-42551565 on Build GRCh38
rs375744320 Chr.3: 42551572-42551572 on Build GRCh38
rs1031816476 Chr.3: 42551509-42551509 on Build GRCh38
rs531297022 Chr.3: 42551659-42551659 on Build GRCh38
rs939846762 Chr.3: 42551336-42551336 on Build GRCh38
rs920382855 Chr.3: 42551825-42551825 on Build GRCh38
rs878913801 Chr.3: 42551648-42551648 on Build GRCh38
rs372393093 Chr.3: 42551482-42551482 on Build GRCh38
rs902275500 Chr.3: 42551770-42551770 on Build GRCh38
rs76371810 Chr.3: 42551519-42551519 on Build GRCh38
rs1022040568 Chr.3: 42551460-42551460 on Build GRCh38
rs940086918 Chr.3: 42551606-42551606 on Build GRCh38
rs908725754 Chr.3: 42551492-42551492 on Build GRCh38
rs548201997 Chr.3: 42551669-42551669 on Build GRCh38
rs903231623 Chr.3: 42551376-42551376 on Build GRCh38

View Full Product Details