Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 45713804-45714316 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SACM1L
Entrez Gene ID
22908
Gene Name
SAC1 suppressor of actin mutations 1-like (yeast)
Gene Aliases
SAC1
UniGene
Hs.156509
Gene Chromosome Location
Chr.3: 45689241-45745425 on Build GRCh38
Transcripts
NM_001319072.1 NM_001319073.1 NM_014016.4 XM_011533500.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6433791 c.545T>C p.V182A Chr.3: 45714047-45714047 on Build GRCh38
1240120 c.572A>G p.H191R Chr.3: 45714074-45714074 on Build GRCh38
6188963 c.575G>T p.G192V Chr.3: 45714077-45714077 on Build GRCh38
6771306 c.551G>A p.R184Q Chr.3: 45714053-45714053 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs969147363 Chr.3: 45713968-45713968 on Build GRCh38
rs773470861 Chr.3: 45714022-45714022 on Build GRCh38
rs6764981 Chr.3: 45714144-45714144 on Build GRCh38
rs778580468 Chr.3: 45714101-45714101 on Build GRCh38
rs768476198 Chr.3: 45714124-45714124 on Build GRCh38
rs767390774 Chr.3: 45714139-45714139 on Build GRCh38
rs1046859014 Chr.3: 45713898-45713898 on Build GRCh38
rs919082849 Chr.3: 45714243-45714243 on Build GRCh38
rs71619608 Chr.3: 45714245-45714245 on Build GRCh38
rs76872275 Chr.3: 45714239-45714239 on Build GRCh38
rs952779736 Chr.3: 45713911-45713911 on Build GRCh38
rs963073228 Chr.3: 45714188-45714188 on Build GRCh38
rs746756381 Chr.3: 45714123-45714123 on Build GRCh38
rs981803066 Chr.3: 45714010-45714010 on Build GRCh38
rs112551935 Chr.3: 45713882-45713882 on Build GRCh38
rs574843246 Chr.3: 45714027-45714027 on Build GRCh38
rs1055093057 Chr.3: 45714280-45714280 on Build GRCh38
rs985828151 Chr.3: 45713837-45713837 on Build GRCh38
rs893914470 Chr.3: 45714281-45714281 on Build GRCh38
rs760291257 Chr.3: 45714255-45714256 on Build GRCh38
rs769001960 Chr.3: 45714131-45714132 on Build GRCh38
rs549146108 Chr.3: 45714237-45714237 on Build GRCh38
rs112928052 Chr.3: 45714238-45714240 on Build GRCh38
rs1007408561 Chr.3: 45713877-45713877 on Build GRCh38
rs772886658 Chr.3: 45714239-45714241 on Build GRCh38
rs1011082909 Chr.3: 45714066-45714066 on Build GRCh38
rs756814737 Chr.3: 45714098-45714098 on Build GRCh38
rs185377042 Chr.3: 45713887-45713887 on Build GRCh38
rs913475536 Chr.3: 45714104-45714104 on Build GRCh38
rs565598315 Chr.3: 45714053-45714053 on Build GRCh38
rs973222152 Chr.3: 45714223-45714223 on Build GRCh38
rs375105714 Chr.3: 45714054-45714054 on Build GRCh38
rs761087165 Chr.3: 45714129-45714130 on Build GRCh38
rs373301031 Chr.3: 45714237-45714237 on Build GRCh38
rs1002327484 Chr.3: 45714000-45714000 on Build GRCh38
rs906515890 Chr.3: 45713965-45713965 on Build GRCh38
rs6765078 Chr.3: 45714241-45714241 on Build GRCh38
rs983798322 Chr.3: 45713932-45713932 on Build GRCh38
rs201390331 Chr.3: 45714234-45714237 on Build GRCh38
rs375638527 Chr.3: 45714241-45714242 on Build GRCh38
rs796973405 Chr.3: 45714241-45714242 on Build GRCh38
rs751900129 Chr.3: 45714063-45714063 on Build GRCh38
rs35864358 Chr.3: 45714241-45714241 on Build GRCh38
rs938002924 Chr.3: 45714279-45714279 on Build GRCh38
rs527332670 Chr.3: 45714168-45714168 on Build GRCh38
rs531637374 Chr.3: 45713905-45713905 on Build GRCh38
rs755741002 Chr.3: 45714067-45714067 on Build GRCh38
rs893862425 Chr.3: 45714029-45714029 on Build GRCh38
rs200685145 Chr.3: 45714240-45714241 on Build GRCh38
rs994012286 Chr.3: 45713897-45713897 on Build GRCh38
rs537876297 Chr.3: 45714288-45714288 on Build GRCh38
rs967379861 Chr.3: 45714069-45714069 on Build GRCh38
rs540016479 Chr.3: 45714282-45714283 on Build GRCh38
rs753612806 Chr.3: 45714096-45714096 on Build GRCh38
rs1055207530 Chr.3: 45714012-45714012 on Build GRCh38
rs746104091 Chr.3: 45714239-45714240 on Build GRCh38
rs749654295 Chr.3: 45713952-45713952 on Build GRCh38
rs200961807 Chr.3: 45714117-45714117 on Build GRCh38
rs566285051 Chr.3: 45714163-45714163 on Build GRCh38
rs762058385 Chr.3: 45714005-45714005 on Build GRCh38
rs910417459 Chr.3: 45713864-45713864 on Build GRCh38
rs929237310 Chr.3: 45714245-45714245 on Build GRCh38
rs927989894 Chr.3: 45714255-45714255 on Build GRCh38
rs543740378 Chr.3: 45714014-45714014 on Build GRCh38
rs752089310 Chr.3: 45714031-45714031 on Build GRCh38
rs5019353 Chr.3: 45714238-45714238 on Build GRCh38
rs762986078 Chr.3: 45714025-45714025 on Build GRCh38
rs775668094 Chr.3: 45714119-45714120 on Build GRCh38
rs1037456535 Chr.3: 45713884-45713884 on Build GRCh38
rs1023752282 Chr.3: 45713957-45713957 on Build GRCh38
rs982047659 Chr.3: 45714246-45714246 on Build GRCh38
rs777539028 Chr.3: 45714074-45714074 on Build GRCh38
rs889863284 Chr.3: 45713872-45713872 on Build GRCh38
rs929210263 Chr.3: 45713888-45713888 on Build GRCh38
rs201461620 Chr.3: 45714105-45714105 on Build GRCh38

View Full Product Details