Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 107571010-107571528 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VAV3
Entrez Gene ID
10451
Gene Name
vav guanine nucleotide exchange factor 3
Gene Aliases
-
UniGene
Hs.267659
Gene Chromosome Location
Chr.1: 107571160-107964923 on Build GRCh38
Transcripts
NM_001079874.1 NM_006113.4 XM_005270360.2 XM_017000053.1 XM_017000055.1 XM_017000056.1

SNP Detail

SNP ID Chromosome Location
rs916148231 Chr.1: 107571132-107571132 on Build GRCh38
rs1038804274 Chr.1: 107571493-107571493 on Build GRCh38
rs532870032 Chr.1: 107571208-107571208 on Build GRCh38
rs896014040 Chr.1: 107571369-107571369 on Build GRCh38
rs3790681 Chr.1: 107571183-107571183 on Build GRCh38
rs8458 Chr.1: 107571234-107571234 on Build GRCh38
rs879458292 Chr.1: 107571280-107571280 on Build GRCh38
rs571262744 Chr.1: 107571237-107571237 on Build GRCh38
rs964515486 Chr.1: 107571489-107571500 on Build GRCh38
rs970717612 Chr.1: 107571397-107571397 on Build GRCh38
rs924934096 Chr.1: 107571216-107571216 on Build GRCh38
rs1006039851 Chr.1: 107571185-107571185 on Build GRCh38
rs780738472 Chr.1: 107571404-107571404 on Build GRCh38
rs916082227 Chr.1: 107571342-107571342 on Build GRCh38
rs779421069 Chr.1: 107571272-107571272 on Build GRCh38
rs540185217 Chr.1: 107571121-107571121 on Build GRCh38
rs12410842 Chr.1: 107571080-107571080 on Build GRCh38
rs567097555 Chr.1: 107571350-107571350 on Build GRCh38
rs889029854 Chr.1: 107571317-107571317 on Build GRCh38
rs747605006 Chr.1: 107571427-107571427 on Build GRCh38
rs1029585429 Chr.1: 107571242-107571242 on Build GRCh38
rs1051660464 Chr.1: 107571054-107571054 on Build GRCh38
rs796904827 Chr.1: 107571046-107571046 on Build GRCh38
rs993062088 Chr.1: 107571123-107571123 on Build GRCh38
rs1045384484 Chr.1: 107571063-107571063 on Build GRCh38
rs1032438723 Chr.1: 107571429-107571429 on Build GRCh38
rs994006723 Chr.1: 107571353-107571353 on Build GRCh38
rs8676 Chr.1: 107571243-107571243 on Build GRCh38
rs754104002 Chr.1: 107571124-107571124 on Build GRCh38
rs141569623 Chr.1: 107571376-107571376 on Build GRCh38
rs761019947 Chr.1: 107571086-107571086 on Build GRCh38
rs957705633 Chr.1: 107571449-107571449 on Build GRCh38
rs1016385024 Chr.1: 107571079-107571079 on Build GRCh38
rs745880260 Chr.1: 107571407-107571407 on Build GRCh38
rs567777531 Chr.1: 107571125-107571125 on Build GRCh38
rs139302493 Chr.1: 107571356-107571356 on Build GRCh38
rs887693081 Chr.1: 107571447-107571447 on Build GRCh38
rs996890816 Chr.1: 107571235-107571235 on Build GRCh38
rs894220051 Chr.1: 107571032-107571032 on Build GRCh38
rs993561339 Chr.1: 107571085-107571085 on Build GRCh38
rs906873812 Chr.1: 107571067-107571067 on Build GRCh38
rs988322999 Chr.1: 107571473-107571473 on Build GRCh38
rs544350115 Chr.1: 107571273-107571273 on Build GRCh38
rs558637805 Chr.1: 107571495-107571495 on Build GRCh38
rs147326269 Chr.1: 107571277-107571277 on Build GRCh38
rs957888776 Chr.1: 107571318-107571318 on Build GRCh38
rs1023211549 Chr.1: 107571392-107571392 on Build GRCh38
rs552773395 Chr.1: 107571359-107571359 on Build GRCh38
rs572795705 Chr.1: 107571372-107571372 on Build GRCh38
rs190950060 Chr.1: 107571451-107571451 on Build GRCh38
rs899136499 Chr.1: 107571349-107571349 on Build GRCh38
rs750890540 Chr.1: 107571285-107571285 on Build GRCh38
rs990329599 Chr.1: 107571339-107571339 on Build GRCh38
rs1006591258 Chr.1: 107571454-107571454 on Build GRCh38
rs3790682 Chr.1: 107571041-107571041 on Build GRCh38
rs1017889783 Chr.1: 107571218-107571218 on Build GRCh38
rs1026454061 Chr.1: 107571105-107571105 on Build GRCh38

View Full Product Details