Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 108560152-108560665 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM102B
Entrez Gene ID
284611
Gene Name
family with sequence similarity 102 member B
Gene Aliases
SYM-3B
UniGene
Hs.200230
Gene Chromosome Location
Chr.1: 108560349-108639327 on Build GRCh38
Transcripts
NM_001010883.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6658028 c.8A>T p.K3M Chr.1: 108560436-108560436 on Build GRCh38
7366932 c.74A>G p.N25S Chr.1: 108560502-108560502 on Build GRCh38
414204 c.59C>T p.S20L Chr.1: 108560487-108560487 on Build GRCh38
6658029 c.77G>T p.G26V Chr.1: 108560505-108560505 on Build GRCh38
2116830 c.60A>G p.S20S Chr.1: 108560488-108560488 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs561380422 Chr.1: 108560290-108560290 on Build GRCh38
rs780223694 Chr.1: 108560554-108560554 on Build GRCh38
rs376138081 Chr.1: 108560565-108560565 on Build GRCh38
rs373919018 Chr.1: 108560612-108560612 on Build GRCh38
rs544681986 Chr.1: 108560506-108560506 on Build GRCh38
rs916091733 Chr.1: 108560468-108560468 on Build GRCh38
rs985752305 Chr.1: 108560188-108560188 on Build GRCh38
rs759539534 Chr.1: 108560608-108560608 on Build GRCh38
rs530385405 Chr.1: 108560544-108560544 on Build GRCh38
rs200430461 Chr.1: 108560531-108560531 on Build GRCh38
rs974206481 Chr.1: 108560373-108560373 on Build GRCh38
rs890760752 Chr.1: 108560627-108560627 on Build GRCh38
rs777913977 Chr.1: 108560530-108560536 on Build GRCh38
rs542025471 Chr.1: 108560378-108560379 on Build GRCh38
rs539121326 Chr.1: 108560172-108560172 on Build GRCh38
rs918680870 Chr.1: 108560210-108560210 on Build GRCh38
rs72978732 Chr.1: 108560585-108560585 on Build GRCh38
rs770993896 Chr.1: 108560452-108560452 on Build GRCh38
rs565032126 Chr.1: 108560509-108560509 on Build GRCh38
rs540398636 Chr.1: 108560382-108560382 on Build GRCh38
rs1007066096 Chr.1: 108560339-108560339 on Build GRCh38
rs544631979 Chr.1: 108560277-108560277 on Build GRCh38
rs764509618 Chr.1: 108560394-108560394 on Build GRCh38
rs974025346 Chr.1: 108560273-108560295 on Build GRCh38
rs369092652 Chr.1: 108560405-108560405 on Build GRCh38
rs375624183 Chr.1: 108560426-108560426 on Build GRCh38
rs970146955 Chr.1: 108560479-108560479 on Build GRCh38
rs756138301 Chr.1: 108560415-108560415 on Build GRCh38
rs917950900 Chr.1: 108560247-108560247 on Build GRCh38
rs775971090 Chr.1: 108560392-108560392 on Build GRCh38
rs749270477 Chr.1: 108560566-108560567 on Build GRCh38
rs762063164 Chr.1: 108560568-108560568 on Build GRCh38
rs911375350 Chr.1: 108560192-108560192 on Build GRCh38
rs770369037 Chr.1: 108560461-108560461 on Build GRCh38
rs765133858 Chr.1: 108560610-108560610 on Build GRCh38
rs763378751 Chr.1: 108560393-108560393 on Build GRCh38
rs868356259 Chr.1: 108560308-108560308 on Build GRCh38
rs760153524 Chr.1: 108560380-108560380 on Build GRCh38
rs1018496919 Chr.1: 108560347-108560347 on Build GRCh38
rs747246331 Chr.1: 108560433-108560433 on Build GRCh38
rs113771630 Chr.1: 108560409-108560409 on Build GRCh38
rs753798782 Chr.1: 108560522-108560522 on Build GRCh38
rs751275001 Chr.1: 108560403-108560403 on Build GRCh38
rs781325876 Chr.1: 108560427-108560428 on Build GRCh38
rs754788417 Chr.1: 108560392-108560418 on Build GRCh38
rs752474055 Chr.1: 108560533-108560533 on Build GRCh38
rs751190875 Chr.1: 108560632-108560632 on Build GRCh38
rs745910731 Chr.1: 108560459-108560459 on Build GRCh38
rs955810226 Chr.1: 108560176-108560176 on Build GRCh38
rs941542933 Chr.1: 108560200-108560200 on Build GRCh38
rs201561046 Chr.1: 108560591-108560591 on Build GRCh38
rs918796950 Chr.1: 108560399-108560399 on Build GRCh38
rs767056033 Chr.1: 108560406-108560406 on Build GRCh38
rs763896151 Chr.1: 108560616-108560616 on Build GRCh38
rs957501194 Chr.1: 108560462-108560462 on Build GRCh38
rs781389459 Chr.1: 108560453-108560453 on Build GRCh38
rs905671523 Chr.1: 108560274-108560274 on Build GRCh38
rs201974354 Chr.1: 108560502-108560502 on Build GRCh38
rs935938008 Chr.1: 108560519-108560519 on Build GRCh38
rs767064898 Chr.1: 108560495-108560495 on Build GRCh38
rs1010026244 Chr.1: 108560324-108560324 on Build GRCh38
rs769279537 Chr.1: 108560563-108560563 on Build GRCh38
rs559124584 Chr.1: 108560215-108560215 on Build GRCh38
rs781236837 Chr.1: 108560549-108560549 on Build GRCh38
rs754799492 Chr.1: 108560427-108560427 on Build GRCh38
rs1056866730 Chr.1: 108560227-108560227 on Build GRCh38
rs765890989 Chr.1: 108560598-108560598 on Build GRCh38
rs949995910 Chr.1: 108560253-108560253 on Build GRCh38
rs749816739 Chr.1: 108560562-108560562 on Build GRCh38
rs756330277 Chr.1: 108560444-108560449 on Build GRCh38
rs929316347 Chr.1: 108560542-108560542 on Build GRCh38
rs763504206 Chr.1: 108560481-108560481 on Build GRCh38
rs575543393 Chr.1: 108560256-108560256 on Build GRCh38
rs374266127 Chr.1: 108560482-108560482 on Build GRCh38
rs575078510 Chr.1: 108560327-108560327 on Build GRCh38
rs771797681 Chr.1: 108560579-108560579 on Build GRCh38
rs1027098602 Chr.1: 108560367-108560390 on Build GRCh38
rs761590574 Chr.1: 108560485-108560485 on Build GRCh38
rs776168072 Chr.1: 108560465-108560465 on Build GRCh38
rs752666702 Chr.1: 108560434-108560436 on Build GRCh38
rs762677111 Chr.1: 108560385-108560386 on Build GRCh38
rs534065919 Chr.1: 108560277-108560303 on Build GRCh38
rs779837976 Chr.1: 108560423-108560423 on Build GRCh38
rs974102418 Chr.1: 108560207-108560207 on Build GRCh38
rs769786624 Chr.1: 108560551-108560551 on Build GRCh38
rs1029722702 Chr.1: 108560328-108560328 on Build GRCh38
rs955093696 Chr.1: 108560338-108560338 on Build GRCh38

View Full Product Details