Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 127004948-127005445 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLXNA1
Entrez Gene ID
5361
Gene Name
plexin A1
Gene Aliases
NOV, NOVP, PLEXIN-A1, PLXN1
UniGene
Hs.432329
Gene Chromosome Location
Chr.3: 126982119-127037392 on Build GRCh38
Transcripts
NM_032242.3 XM_011512908.2 XM_011512909.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3059023 c.1785C>T p.G595G Chr.3: 127005131-127005131 on Build GRCh38
6870479 c.1761G>A p.W587* Chr.3: 127005107-127005107 on Build GRCh38
108806 c.1749G>A p.V583V Chr.3: 127005095-127005095 on Build GRCh38
3059021 c.1707G>A p.Q569Q Chr.3: 127004972-127004972 on Build GRCh38
4715879 c.1887G>A p.T629T Chr.3: 127005233-127005233 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs73196539 Chr.3: 127005308-127005308 on Build GRCh38
rs753095268 Chr.3: 127005271-127005271 on Build GRCh38
rs762958945 Chr.3: 127005198-127005198 on Build GRCh38
rs752717809 Chr.3: 127005187-127005187 on Build GRCh38
rs184131664 Chr.3: 127005224-127005224 on Build GRCh38
rs952174835 Chr.3: 127005409-127005409 on Build GRCh38
rs767016402 Chr.3: 127005208-127005208 on Build GRCh38
rs750588302 Chr.3: 127005005-127005005 on Build GRCh38
rs370519091 Chr.3: 127005210-127005210 on Build GRCh38
rs541827996 Chr.3: 127005289-127005289 on Build GRCh38
rs755963552 Chr.3: 127005016-127005016 on Build GRCh38
rs766246674 Chr.3: 127005110-127005110 on Build GRCh38
rs534632249 Chr.3: 127005209-127005209 on Build GRCh38
rs370070914 Chr.3: 127005039-127005039 on Build GRCh38
rs374361480 Chr.3: 127005062-127005062 on Build GRCh38
rs759267125 Chr.3: 127005184-127005184 on Build GRCh38
rs771092009 Chr.3: 127005172-127005172 on Build GRCh38
rs149350048 Chr.3: 127005095-127005095 on Build GRCh38
rs778864044 Chr.3: 127005038-127005038 on Build GRCh38
rs758697652 Chr.3: 127005274-127005274 on Build GRCh38
rs919459505 Chr.3: 127005370-127005370 on Build GRCh38
rs779633986 Chr.3: 127005270-127005270 on Build GRCh38
rs575231185 Chr.3: 127005055-127005055 on Build GRCh38
rs993951099 Chr.3: 127004969-127004969 on Build GRCh38
rs370707852 Chr.3: 127005235-127005235 on Build GRCh38
rs762794822 Chr.3: 127005241-127005241 on Build GRCh38
rs569605295 Chr.3: 127005178-127005178 on Build GRCh38
rs550659032 Chr.3: 127005283-127005283 on Build GRCh38
rs371942161 Chr.3: 127005146-127005146 on Build GRCh38
rs1020760593 Chr.3: 127005075-127005075 on Build GRCh38
rs761286405 Chr.3: 127005248-127005248 on Build GRCh38
rs764283137 Chr.3: 127004980-127004980 on Build GRCh38
rs759769082 Chr.3: 127005234-127005234 on Build GRCh38
rs368237189 Chr.3: 127005074-127005074 on Build GRCh38
rs754851533 Chr.3: 127005037-127005037 on Build GRCh38
rs547309194 Chr.3: 127005111-127005111 on Build GRCh38
rs985068323 Chr.3: 127005137-127005137 on Build GRCh38
rs771388048 Chr.3: 127005068-127005068 on Build GRCh38
rs757917976 Chr.3: 127005045-127005045 on Build GRCh38
rs929447994 Chr.3: 127005400-127005400 on Build GRCh38
rs117043507 Chr.3: 127004976-127004976 on Build GRCh38
rs961427561 Chr.3: 127005342-127005342 on Build GRCh38
rs767572061 Chr.3: 127005077-127005077 on Build GRCh38
rs753856121 Chr.3: 127005023-127005023 on Build GRCh38
rs189195790 Chr.3: 127005019-127005019 on Build GRCh38
rs976733753 Chr.3: 127005348-127005348 on Build GRCh38
rs767049981 Chr.3: 127005251-127005251 on Build GRCh38
rs201695917 Chr.3: 127005120-127005120 on Build GRCh38
rs539560790 Chr.3: 127005319-127005319 on Build GRCh38
rs761788745 Chr.3: 127005382-127005382 on Build GRCh38
rs758044637 Chr.3: 127005124-127005124 on Build GRCh38
rs150750071 Chr.3: 127005186-127005186 on Build GRCh38
rs553298543 Chr.3: 127005346-127005346 on Build GRCh38
rs775444611 Chr.3: 127005238-127005238 on Build GRCh38
rs574788327 Chr.3: 127005232-127005232 on Build GRCh38
rs774942164 Chr.3: 127005151-127005151 on Build GRCh38
rs774316438 Chr.3: 127004985-127004985 on Build GRCh38
rs769006559 Chr.3: 127005154-127005154 on Build GRCh38
rs766083933 Chr.3: 127005022-127005022 on Build GRCh38
rs748705785 Chr.3: 127005214-127005214 on Build GRCh38
rs769072483 Chr.3: 127005070-127005070 on Build GRCh38
rs753616251 Chr.3: 127005345-127005345 on Build GRCh38
rs149221143 Chr.3: 127005158-127005158 on Build GRCh38
rs143376196 Chr.3: 127005171-127005171 on Build GRCh38
rs1024605465 Chr.3: 127005339-127005339 on Build GRCh38
rs376837581 Chr.3: 127005233-127005233 on Build GRCh38
rs985071890 Chr.3: 127005420-127005420 on Build GRCh38
rs781305806 Chr.3: 127005293-127005293 on Build GRCh38
rs967938675 Chr.3: 127005096-127005096 on Build GRCh38
rs910849453 Chr.3: 127005333-127005333 on Build GRCh38
rs1013431227 Chr.3: 127005080-127005080 on Build GRCh38
rs745884301 Chr.3: 127005294-127005294 on Build GRCh38
rs145552902 Chr.3: 127005131-127005131 on Build GRCh38
rs774854361 Chr.3: 127005072-127005072 on Build GRCh38
rs140490374 Chr.3: 127005213-127005213 on Build GRCh38
rs772590873 Chr.3: 127005223-127005223 on Build GRCh38
rs887810163 Chr.3: 127005219-127005219 on Build GRCh38
rs372511113 Chr.3: 127005105-127005105 on Build GRCh38
rs374749572 Chr.3: 127005228-127005228 on Build GRCh38
rs376751815 Chr.3: 127005189-127005189 on Build GRCh38
rs544545201 Chr.3: 127005182-127005182 on Build GRCh38
rs200374384 Chr.3: 127005132-127005132 on Build GRCh38
rs368596147 Chr.3: 127004999-127004999 on Build GRCh38
rs753291183 Chr.3: 127005340-127005340 on Build GRCh38
rs576626735 Chr.3: 127005207-127005207 on Build GRCh38
rs773489726 Chr.3: 127005093-127005093 on Build GRCh38
rs751485086 Chr.3: 127005246-127005246 on Build GRCh38
rs761839227 Chr.3: 127004994-127004994 on Build GRCh38
rs991892572 Chr.3: 127005156-127005156 on Build GRCh38
rs754766599 Chr.3: 127005118-127005118 on Build GRCh38
rs764129635 Chr.3: 127005242-127005242 on Build GRCh38
rs777622915 Chr.3: 127005050-127005050 on Build GRCh38
rs116047844 Chr.3: 127005199-127005199 on Build GRCh38
rs1033537516 Chr.3: 127005307-127005307 on Build GRCh38
rs374592740 Chr.3: 127005267-127005267 on Build GRCh38
rs145298497 Chr.3: 127005136-127005136 on Build GRCh38
rs200869892 Chr.3: 127005064-127005064 on Build GRCh38
rs749619543 Chr.3: 127005152-127005152 on Build GRCh38
rs370503227 Chr.3: 127005043-127005044 on Build GRCh38
rs750083584 Chr.3: 127005265-127005265 on Build GRCh38
rs923494024 Chr.3: 127005357-127005357 on Build GRCh38
rs372767653 Chr.3: 127005067-127005067 on Build GRCh38

View Full Product Details