Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 167288781-167289288 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZBBX
Entrez Gene ID
79740
Gene Name
zinc finger B-box domain containing
Gene Aliases
-
UniGene
Hs.478143
Gene Chromosome Location
Chr.3: 167178399-167380405 on Build GRCh38
Transcripts
NM_001199201.1 NM_001199202.1 NM_024687.3 XM_011513144.1 XM_011513145.2 XM_011513148.2 XM_011513149.2 XM_011513151.2 XM_011513152.2 XM_011513153.2 XM_017007186.1 XM_017007187.1 XM_017007188.1 XM_017007189.1 XM_017007190.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5524518 c.1948C>T p.L650L Chr.3: 167288915-167288915 on Build GRCh38
6499995 c.1979G>T p.R660I Chr.3: 167288884-167288884 on Build GRCh38
6204965 c.1909T>G p.L637V Chr.3: 167288954-167288954 on Build GRCh38
5433637 c.1989A>C p.Q663H Chr.3: 167288874-167288874 on Build GRCh38
6215036 c.1950G>A p.L650L Chr.3: 167288913-167288913 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs955422513 Chr.3: 167288849-167288849 on Build GRCh38
rs1054624436 Chr.3: 167289170-167289170 on Build GRCh38
rs547732133 Chr.3: 167289034-167289034 on Build GRCh38
rs1051065731 Chr.3: 167289023-167289023 on Build GRCh38
rs942975747 Chr.3: 167289114-167289114 on Build GRCh38
rs918679082 Chr.3: 167289000-167289000 on Build GRCh38
rs1055924248 Chr.3: 167289183-167289183 on Build GRCh38
rs180837087 Chr.3: 167288847-167288847 on Build GRCh38
rs780669297 Chr.3: 167288888-167288888 on Build GRCh38
rs201896466 Chr.3: 167288867-167288867 on Build GRCh38
rs894230366 Chr.3: 167289202-167289202 on Build GRCh38
rs755473675 Chr.3: 167288913-167288913 on Build GRCh38
rs541380394 Chr.3: 167289109-167289109 on Build GRCh38
rs1047339316 Chr.3: 167289113-167289113 on Build GRCh38
rs911321639 Chr.3: 167288876-167288876 on Build GRCh38
rs868489342 Chr.3: 167288948-167288948 on Build GRCh38
rs748193059 Chr.3: 167288947-167288947 on Build GRCh38
rs531908149 Chr.3: 167288952-167288952 on Build GRCh38
rs890026621 Chr.3: 167289039-167289039 on Build GRCh38
rs568698217 Chr.3: 167289006-167289006 on Build GRCh38
rs941450756 Chr.3: 167288920-167288920 on Build GRCh38
rs920133388 Chr.3: 167289040-167289040 on Build GRCh38
rs758817191 Chr.3: 167288845-167288845 on Build GRCh38
rs186109409 Chr.3: 167289020-167289020 on Build GRCh38
rs550112607 Chr.3: 167288977-167288977 on Build GRCh38
rs78204964 Chr.3: 167289223-167289223 on Build GRCh38
rs566110723 Chr.3: 167289134-167289134 on Build GRCh38
rs928271663 Chr.3: 167289149-167289149 on Build GRCh38
rs1031404125 Chr.3: 167288837-167288837 on Build GRCh38
rs7625651 Chr.3: 167288824-167288824 on Build GRCh38
rs781310128 Chr.3: 167288934-167288934 on Build GRCh38
rs930206064 Chr.3: 167289057-167289057 on Build GRCh38

View Full Product Details