Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 169773906-169774438 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYNN
Entrez Gene ID
55892
Gene Name
myoneurin
Gene Aliases
OSZF, SBBIZ1, ZBTB31, ZNF902
UniGene
Hs.507025
Gene Chromosome Location
Chr.3: 169772247-169789716 on Build GRCh38
Transcripts
NM_001185118.1 NM_001185119.1 NM_018657.4 NR_033702.1 NR_033703.1 XM_005247621.4 XM_005247622.3 XM_005247624.3 XM_011512987.1 XM_017006864.1 XM_017006865.1 XM_017006866.1 XM_017006867.1 XM_017006868.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4704748 c.51G>A p.Q17Q Chr.3: 169774346-169774346 on Build GRCh38
6255468 c.36G>T p.E12D Chr.3: 169774331-169774331 on Build GRCh38
7252780 c.127G>A p.A43T Chr.3: 169774422-169774422 on Build GRCh38
3731394 c.67_80del14 p.L23fs*9 Chr.3: 169774362-169774375 on Build GRCh38
3427305 c.11C>T p.S4L Chr.3: 169774306-169774306 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs952570872 Chr.3: 169774080-169774080 on Build GRCh38
rs770427794 Chr.3: 169774265-169774265 on Build GRCh38
rs777192345 Chr.3: 169774251-169774251 on Build GRCh38
rs778944352 Chr.3: 169774010-169774010 on Build GRCh38
rs572735755 Chr.3: 169774172-169774172 on Build GRCh38
rs1031278848 Chr.3: 169774057-169774057 on Build GRCh38
rs144366076 Chr.3: 169774209-169774209 on Build GRCh38
rs1015546859 Chr.3: 169774098-169774098 on Build GRCh38
rs150417121 Chr.3: 169774400-169774400 on Build GRCh38
rs926141456 Chr.3: 169774247-169774247 on Build GRCh38
rs766552335 Chr.3: 169774307-169774307 on Build GRCh38
rs974167476 Chr.3: 169774167-169774167 on Build GRCh38
rs113007230 Chr.3: 169774192-169774192 on Build GRCh38
rs753509538 Chr.3: 169773960-169773960 on Build GRCh38
rs397877370 Chr.3: 169774229-169774230 on Build GRCh38
rs1042850179 Chr.3: 169774250-169774250 on Build GRCh38
rs1021375143 Chr.3: 169773942-169773942 on Build GRCh38
rs762770402 Chr.3: 169774180-169774184 on Build GRCh38
rs899586451 Chr.3: 169773930-169773930 on Build GRCh38
rs752302853 Chr.3: 169774056-169774056 on Build GRCh38
rs777353210 Chr.3: 169774074-169774074 on Build GRCh38
rs916696868 Chr.3: 169774119-169774119 on Build GRCh38
rs374675135 Chr.3: 169774205-169774205 on Build GRCh38
rs577077796 Chr.3: 169774277-169774277 on Build GRCh38
rs1011410438 Chr.3: 169774237-169774237 on Build GRCh38
rs769021933 Chr.3: 169774247-169774250 on Build GRCh38
rs754996008 Chr.3: 169773989-169773989 on Build GRCh38
rs187779073 Chr.3: 169774063-169774063 on Build GRCh38
rs762163070 Chr.3: 169774267-169774267 on Build GRCh38
rs11553294 Chr.3: 169774367-169774367 on Build GRCh38
rs1047022624 Chr.3: 169774195-169774195 on Build GRCh38
rs368002667 Chr.3: 169774329-169774329 on Build GRCh38
rs199548793 Chr.3: 169774199-169774199 on Build GRCh38
rs34847803 Chr.3: 169774228-169774229 on Build GRCh38
rs542089926 Chr.3: 169774004-169774004 on Build GRCh38
rs571627140 Chr.3: 169774072-169774072 on Build GRCh38
rs902920337 Chr.3: 169774301-169774301 on Build GRCh38
rs899700193 Chr.3: 169774218-169774218 on Build GRCh38
rs912171297 Chr.3: 169774308-169774308 on Build GRCh38
rs10936599 Chr.3: 169774313-169774313 on Build GRCh38
rs912500283 Chr.3: 169774198-169774198 on Build GRCh38
rs763197153 Chr.3: 169774306-169774306 on Build GRCh38
rs769346608 Chr.3: 169774245-169774245 on Build GRCh38
rs930008351 Chr.3: 169774187-169774187 on Build GRCh38
rs558494994 Chr.3: 169773962-169773962 on Build GRCh38
rs969452071 Chr.3: 169774126-169774126 on Build GRCh38
rs1053287765 Chr.3: 169774287-169774287 on Build GRCh38
rs1035739899 Chr.3: 169774325-169774325 on Build GRCh38
rs967072113 Chr.3: 169773953-169773953 on Build GRCh38
rs879208832 Chr.3: 169774281-169774281 on Build GRCh38
rs568315645 Chr.3: 169773961-169773961 on Build GRCh38
rs1004153760 Chr.3: 169774081-169774081 on Build GRCh38
rs773120189 Chr.3: 169774280-169774280 on Build GRCh38
rs764003058 Chr.3: 169774319-169774319 on Build GRCh38
rs746657214 Chr.3: 169774197-169774197 on Build GRCh38
rs2251795 Chr.3: 169773941-169773941 on Build GRCh38
rs757948379 Chr.3: 169774066-169774066 on Build GRCh38
rs757082996 Chr.3: 169774381-169774381 on Build GRCh38
rs192213312 Chr.3: 169774169-169774169 on Build GRCh38

View Full Product Details