Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 180989253-180989759 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC19
Entrez Gene ID
131118
Gene Name
DnaJ heat shock protein family (Hsp40) member C19
Gene Aliases
PAM18, TIM14, TIMM14
UniGene
Hs.230601
Gene Chromosome Location
Chr.3: 180983709-180989774 on Build GRCh38
Transcripts
NM_001190233.1 NM_145261.3 NR_033721.1 NR_033722.1 NR_033723.1 NR_046073.1

SNP Detail

SNP ID Chromosome Location
rs751535411 Chr.3: 180989613-180989613 on Build GRCh38
rs763925180 Chr.3: 180989694-180989694 on Build GRCh38
rs986484341 Chr.3: 180989664-180989664 on Build GRCh38
rs1049235893 Chr.3: 180989373-180989373 on Build GRCh38
rs990675081 Chr.3: 180989344-180989344 on Build GRCh38
rs1036428783 Chr.3: 180989739-180989739 on Build GRCh38
rs768938396 Chr.3: 180989661-180989661 on Build GRCh38
rs755871059 Chr.3: 180989554-180989554 on Build GRCh38
rs769945722 Chr.3: 180989669-180989669 on Build GRCh38
rs773549822 Chr.3: 180989462-180989462 on Build GRCh38
rs749953478 Chr.3: 180989709-180989709 on Build GRCh38
rs746420330 Chr.3: 180989642-180989642 on Build GRCh38
rs977872020 Chr.3: 180989632-180989632 on Build GRCh38
rs764915383 Chr.3: 180989699-180989699 on Build GRCh38
rs767566082 Chr.3: 180989687-180989687 on Build GRCh38
rs370546709 Chr.3: 180989549-180989549 on Build GRCh38
rs748266916 Chr.3: 180989666-180989666 on Build GRCh38
rs944947579 Chr.3: 180989282-180989282 on Build GRCh38
rs534362204 Chr.3: 180989570-180989570 on Build GRCh38
rs756805866 Chr.3: 180989634-180989634 on Build GRCh38
rs1049182549 Chr.3: 180989729-180989729 on Build GRCh38
rs527471494 Chr.3: 180989688-180989688 on Build GRCh38
rs1014870228 Chr.3: 180989403-180989403 on Build GRCh38
rs946887546 Chr.3: 180989706-180989706 on Build GRCh38
rs779876511 Chr.3: 180989701-180989702 on Build GRCh38
rs886058205 Chr.3: 180989723-180989723 on Build GRCh38
rs761954093 Chr.3: 180989610-180989610 on Build GRCh38
rs1042635506 Chr.3: 180989723-180989723 on Build GRCh38
rs887821410 Chr.3: 180989380-180989380 on Build GRCh38
rs777635354 Chr.3: 180989630-180989630 on Build GRCh38
rs923612072 Chr.3: 180989281-180989281 on Build GRCh38
rs758947037 Chr.3: 180989651-180989651 on Build GRCh38
rs932139341 Chr.3: 180989367-180989367 on Build GRCh38
rs186253120 Chr.3: 180989737-180989737 on Build GRCh38
rs776284659 Chr.3: 180989474-180989474 on Build GRCh38
rs959437530 Chr.3: 180989341-180989341 on Build GRCh38
rs765170683 Chr.3: 180989611-180989611 on Build GRCh38
rs900650896 Chr.3: 180989326-180989326 on Build GRCh38
rs763537144 Chr.3: 180989468-180989469 on Build GRCh38
rs373661392 Chr.3: 180989566-180989566 on Build GRCh38
rs889550007 Chr.3: 180989467-180989467 on Build GRCh38
rs766521833 Chr.3: 180989564-180989564 on Build GRCh38
rs562479769 Chr.3: 180989469-180989469 on Build GRCh38
rs775794757 Chr.3: 180989583-180989583 on Build GRCh38
rs918275176 Chr.3: 180989416-180989416 on Build GRCh38
rs746819742 Chr.3: 180989707-180989707 on Build GRCh38
rs780519927 Chr.3: 180989655-180989655 on Build GRCh38
rs777545342 Chr.3: 180989561-180989561 on Build GRCh38
rs763066001 Chr.3: 180989678-180989678 on Build GRCh38
rs991058537 Chr.3: 180989623-180989623 on Build GRCh38
rs367841424 Chr.3: 180989584-180989584 on Build GRCh38
rs755959273 Chr.3: 180989628-180989628 on Build GRCh38
rs756823021 Chr.3: 180989696-180989696 on Build GRCh38
rs772451930 Chr.3: 180989581-180989581 on Build GRCh38
rs761598764 Chr.3: 180989534-180989534 on Build GRCh38
rs747436067 Chr.3: 180989658-180989658 on Build GRCh38
rs533013026 Chr.3: 180989614-180989614 on Build GRCh38
rs778238451 Chr.3: 180989637-180989637 on Build GRCh38
rs181271620 Chr.3: 180989631-180989631 on Build GRCh38
rs767619862 Chr.3: 180989679-180989679 on Build GRCh38
rs751727548 Chr.3: 180989567-180989567 on Build GRCh38
rs894004098 Chr.3: 180989704-180989704 on Build GRCh38
rs933675162 Chr.3: 180989453-180989453 on Build GRCh38
rs748887407 Chr.3: 180989562-180989562 on Build GRCh38
rs757894812 Chr.3: 180989714-180989714 on Build GRCh38
rs1003850979 Chr.3: 180989532-180989532 on Build GRCh38
rs1039601154 Chr.3: 180989698-180989698 on Build GRCh38
rs753552829 Chr.3: 180989695-180989695 on Build GRCh38
rs1034974123 Chr.3: 180989533-180989533 on Build GRCh38
rs568583800 Chr.3: 180989295-180989295 on Build GRCh38
rs375730162 Chr.3: 180989609-180989609 on Build GRCh38
rs918515573 Chr.3: 180989647-180989647 on Build GRCh38
rs909764795 Chr.3: 180989356-180989356 on Build GRCh38
rs752587219 Chr.3: 180989616-180989616 on Build GRCh38
rs1039241486 Chr.3: 180989502-180989502 on Build GRCh38
rs776922253 Chr.3: 180989662-180989662 on Build GRCh38
rs972600083 Chr.3: 180989641-180989641 on Build GRCh38
rs546277315 Chr.3: 180989736-180989736 on Build GRCh38
rs910957972 Chr.3: 180989673-180989673 on Build GRCh38
rs544220508 Chr.3: 180989394-180989394 on Build GRCh38
rs767529677 Chr.3: 180989615-180989615 on Build GRCh38
rs933615918 Chr.3: 180989654-180989654 on Build GRCh38
rs372756221 Chr.3: 180989602-180989602 on Build GRCh38
rs560226416 Chr.3: 180989674-180989674 on Build GRCh38

View Full Product Details