Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 185649296-185649792 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IGF2BP2
Entrez Gene ID
10644
Gene Name
insulin like growth factor 2 mRNA binding protein 2
Gene Aliases
IMP-2, IMP2, VICKZ2
UniGene
Hs.35354
Gene Chromosome Location
Chr.3: 185643739-185825051 on Build GRCh38
Transcripts
NM_001007225.1 NM_001291869.1 NM_001291872.1 NM_001291873.1 NM_001291874.1 NM_001291875.1 NM_006548.4 XM_006713477.3 XM_006713478.3 XM_017005557.1 XM_017005558.1 XM_017005559.1 XM_017005560.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4809129 c.1542C>G p.I514M Chr.3: 185649454-185649454 on Build GRCh38
6402419 c.1462G>T p.A488S Chr.3: 185649534-185649534 on Build GRCh38
281868 c.1500C>A p.N500K Chr.3: 185649496-185649496 on Build GRCh38
5398805 c.1348G>A p.E450K Chr.3: 185649477-185649477 on Build GRCh38
4769721 c.1544G>C p.R515T Chr.3: 185649452-185649452 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs546462111 Chr.3: 185649743-185649743 on Build GRCh38
rs909208096 Chr.3: 185649652-185649652 on Build GRCh38
rs550309596 Chr.3: 185649705-185649705 on Build GRCh38
rs558614643 Chr.3: 185649628-185649628 on Build GRCh38
rs938748208 Chr.3: 185649704-185649704 on Build GRCh38
rs753123780 Chr.3: 185649428-185649428 on Build GRCh38
rs769555482 Chr.3: 185649541-185649541 on Build GRCh38
rs986396151 Chr.3: 185649647-185649647 on Build GRCh38
rs760862269 Chr.3: 185649565-185649565 on Build GRCh38
rs552281158 Chr.3: 185649390-185649390 on Build GRCh38
rs757747673 Chr.3: 185649460-185649460 on Build GRCh38
rs964733655 Chr.3: 185649713-185649713 on Build GRCh38
rs1044027005 Chr.3: 185649656-185649657 on Build GRCh38
rs574792013 Chr.3: 185649685-185649685 on Build GRCh38
rs116832755 Chr.3: 185649626-185649626 on Build GRCh38
rs753375785 Chr.3: 185649361-185649361 on Build GRCh38
rs764499000 Chr.3: 185649566-185649566 on Build GRCh38
rs142886977 Chr.3: 185649686-185649686 on Build GRCh38
rs748167323 Chr.3: 185649532-185649532 on Build GRCh38
rs1021510048 Chr.3: 185649754-185649754 on Build GRCh38
rs111523243 Chr.3: 185649358-185649358 on Build GRCh38
rs368015923 Chr.3: 185649581-185649581 on Build GRCh38
rs576968169 Chr.3: 185649654-185649654 on Build GRCh38
rs745590647 Chr.3: 185649374-185649374 on Build GRCh38
rs568053766 Chr.3: 185649554-185649554 on Build GRCh38
rs767573568 Chr.3: 185649421-185649421 on Build GRCh38
rs568932583 Chr.3: 185649600-185649600 on Build GRCh38
rs765613789 Chr.3: 185649576-185649576 on Build GRCh38
rs942007426 Chr.3: 185649672-185649672 on Build GRCh38
rs758874089 Chr.3: 185649537-185649538 on Build GRCh38
rs772367775 Chr.3: 185649558-185649558 on Build GRCh38
rs747577247 Chr.3: 185649569-185649575 on Build GRCh38
rs754390433 Chr.3: 185649458-185649458 on Build GRCh38
rs570120081 Chr.3: 185649714-185649714 on Build GRCh38
rs542518803 Chr.3: 185649320-185649320 on Build GRCh38
rs778201083 Chr.3: 185649360-185649360 on Build GRCh38
rs757200480 Chr.3: 185649333-185649333 on Build GRCh38
rs775727112 Chr.3: 185649561-185649561 on Build GRCh38
rs764409132 Chr.3: 185649445-185649445 on Build GRCh38
rs779535002 Chr.3: 185649486-185649486 on Build GRCh38
rs541128313 Chr.3: 185649656-185649656 on Build GRCh38
rs190883724 Chr.3: 185649575-185649575 on Build GRCh38
rs932204916 Chr.3: 185649319-185649319 on Build GRCh38
rs758092673 Chr.3: 185649484-185649484 on Build GRCh38
rs77247478 Chr.3: 185649336-185649336 on Build GRCh38
rs771619154 Chr.3: 185649378-185649378 on Build GRCh38
rs760985009 Chr.3: 185649437-185649437 on Build GRCh38
rs377684115 Chr.3: 185649414-185649414 on Build GRCh38
rs371608235 Chr.3: 185649384-185649384 on Build GRCh38
rs553052033 Chr.3: 185649671-185649671 on Build GRCh38
rs528084524 Chr.3: 185649461-185649461 on Build GRCh38
rs139682125 Chr.3: 185649403-185649403 on Build GRCh38
rs537848740 Chr.3: 185649370-185649370 on Build GRCh38
rs374386875 Chr.3: 185649389-185649389 on Build GRCh38
rs1002826974 Chr.3: 185649657-185649657 on Build GRCh38
rs546664849 Chr.3: 185649476-185649476 on Build GRCh38
rs370946762 Chr.3: 185649517-185649517 on Build GRCh38
rs781125157 Chr.3: 185649523-185649523 on Build GRCh38
rs774498754 Chr.3: 185649551-185649551 on Build GRCh38
rs767994973 Chr.3: 185649355-185649355 on Build GRCh38
rs780689764 Chr.3: 185649543-185649544 on Build GRCh38
rs779913085 Chr.3: 185649383-185649383 on Build GRCh38
rs575892850 Chr.3: 185649718-185649718 on Build GRCh38
rs746630574 Chr.3: 185649511-185649511 on Build GRCh38
rs749846827 Chr.3: 185649472-185649472 on Build GRCh38
rs956878826 Chr.3: 185649612-185649612 on Build GRCh38
rs776279089 Chr.3: 185649573-185649573 on Build GRCh38
rs770571942 Chr.3: 185649362-185649362 on Build GRCh38
rs761822372 Chr.3: 185649393-185649393 on Build GRCh38
rs769274279 Chr.3: 185649512-185649512 on Build GRCh38
rs759812324 Chr.3: 185649413-185649413 on Build GRCh38
rs373131894 Chr.3: 185649357-185649357 on Build GRCh38
rs932255829 Chr.3: 185649635-185649635 on Build GRCh38

View Full Product Details