Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 17802854-17803364 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCAF16
Entrez Gene ID
54876
Gene Name
DDB1 and CUL4 associated factor 16
Gene Aliases
C4orf30
UniGene
Hs.614787
Gene Chromosome Location
Chr.4: 17798236-17810761 on Build GRCh38
Transcripts
NM_017741.3 XM_005248169.3 XM_005248171.3 XM_006713966.3 XM_017008327.1 XM_017008328.1

SNP Detail

SNP ID Chromosome Location
rs917386823 Chr.4: 17803136-17803136 on Build GRCh38
rs980692333 Chr.4: 17803186-17803186 on Build GRCh38
rs900370608 Chr.4: 17803074-17803074 on Build GRCh38
rs991961981 Chr.4: 17803119-17803119 on Build GRCh38
rs758557428 Chr.4: 17803101-17803101 on Build GRCh38
rs76846350 Chr.4: 17803043-17803043 on Build GRCh38
rs1009827142 Chr.4: 17803132-17803132 on Build GRCh38
rs926183110 Chr.4: 17803200-17803200 on Build GRCh38
rs932500256 Chr.4: 17802968-17802968 on Build GRCh38
rs386672191 Chr.4: 17803084-17803085 on Build GRCh38
rs968055965 Chr.4: 17803182-17803182 on Build GRCh38
rs773987029 Chr.4: 17803043-17803044 on Build GRCh38
rs530186129 Chr.4: 17802995-17802995 on Build GRCh38
rs146397295 Chr.4: 17803150-17803150 on Build GRCh38
rs552921026 Chr.4: 17803313-17803313 on Build GRCh38
rs757182872 Chr.4: 17803169-17803169 on Build GRCh38
rs533021670 Chr.4: 17803085-17803085 on Build GRCh38
rs983078471 Chr.4: 17803092-17803092 on Build GRCh38
rs954145335 Chr.4: 17803248-17803248 on Build GRCh38
rs111465602 Chr.4: 17803094-17803094 on Build GRCh38
rs1036869894 Chr.4: 17803078-17803078 on Build GRCh38
rs757529352 Chr.4: 17803283-17803283 on Build GRCh38
rs1028438985 Chr.4: 17803231-17803231 on Build GRCh38
rs753223328 Chr.4: 17803143-17803143 on Build GRCh38
rs961468960 Chr.4: 17803285-17803285 on Build GRCh38
rs559421731 Chr.4: 17803084-17803084 on Build GRCh38
rs1044143659 Chr.4: 17803086-17803086 on Build GRCh38
rs939950282 Chr.4: 17803061-17803061 on Build GRCh38
rs986636297 Chr.4: 17803055-17803055 on Build GRCh38
rs149154868 Chr.4: 17803102-17803102 on Build GRCh38
rs35039817 Chr.4: 17803042-17803043 on Build GRCh38
rs530613520 Chr.4: 17803111-17803111 on Build GRCh38
rs896772622 Chr.4: 17803110-17803110 on Build GRCh38
rs189074138 Chr.4: 17803137-17803137 on Build GRCh38
rs3178753 Chr.4: 17803224-17803224 on Build GRCh38
rs972636122 Chr.4: 17803310-17803310 on Build GRCh38
rs761619023 Chr.4: 17803135-17803136 on Build GRCh38
rs1000848165 Chr.4: 17803185-17803185 on Build GRCh38
rs912501741 Chr.4: 17803056-17803056 on Build GRCh38
rs763852166 Chr.4: 17803149-17803149 on Build GRCh38
rs1008944250 Chr.4: 17803027-17803027 on Build GRCh38
rs935973371 Chr.4: 17802929-17802929 on Build GRCh38
rs191994263 Chr.4: 17803261-17803261 on Build GRCh38
rs77332074 Chr.4: 17803042-17803042 on Build GRCh38
rs953375642 Chr.4: 17803268-17803268 on Build GRCh38

View Full Product Details