Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 17804634-17805128 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCAF16
Entrez Gene ID
54876
Gene Name
DDB1 and CUL4 associated factor 16
Gene Aliases
C4orf30
UniGene
Hs.614787
Gene Chromosome Location
Chr.4: 17798236-17810761 on Build GRCh38
Transcripts
NM_017741.3 XM_005248169.3 XM_005248171.3 XM_006713966.3 XM_017008327.1 XM_017008328.1

SNP Detail

SNP ID Chromosome Location
rs1020184235 Chr.4: 17804761-17804761 on Build GRCh38
rs1001406259 Chr.4: 17804689-17804689 on Build GRCh38
rs576497298 Chr.4: 17805085-17805085 on Build GRCh38
rs1006510916 Chr.4: 17804893-17804893 on Build GRCh38
rs1025451074 Chr.4: 17804927-17804927 on Build GRCh38
rs1041221329 Chr.4: 17804795-17804795 on Build GRCh38
rs1056030225 Chr.4: 17804725-17804725 on Build GRCh38
rs895609492 Chr.4: 17804732-17804732 on Build GRCh38
rs1029167768 Chr.4: 17804858-17804858 on Build GRCh38
rs956597296 Chr.4: 17805069-17805069 on Build GRCh38
rs993581028 Chr.4: 17804925-17804925 on Build GRCh38
rs573685120 Chr.4: 17804792-17804792 on Build GRCh38
rs772568622 Chr.4: 17805071-17805071 on Build GRCh38
rs565025640 Chr.4: 17804806-17804806 on Build GRCh38
rs548701567 Chr.4: 17805008-17805008 on Build GRCh38
rs141759496 Chr.4: 17804673-17804673 on Build GRCh38
rs1048371644 Chr.4: 17805005-17805005 on Build GRCh38
rs1040737615 Chr.4: 17805094-17805094 on Build GRCh38
rs770159725 Chr.4: 17805084-17805085 on Build GRCh38
rs998072368 Chr.4: 17804837-17804837 on Build GRCh38
rs879545408 Chr.4: 17804775-17804775 on Build GRCh38
rs550681423 Chr.4: 17804928-17804928 on Build GRCh38
rs1032791116 Chr.4: 17804801-17804801 on Build GRCh38
rs937423083 Chr.4: 17805043-17805043 on Build GRCh38
rs780990874 Chr.4: 17805086-17805087 on Build GRCh38
rs1055847143 Chr.4: 17805091-17805091 on Build GRCh38
rs567015802 Chr.4: 17805038-17805038 on Build GRCh38
rs542695114 Chr.4: 17804981-17804981 on Build GRCh38
rs150129650 Chr.4: 17804740-17804740 on Build GRCh38
rs911383098 Chr.4: 17804903-17804903 on Build GRCh38
rs1016268862 Chr.4: 17804901-17804902 on Build GRCh38
rs138477206 Chr.4: 17804864-17804864 on Build GRCh38
rs930005054 Chr.4: 17805004-17805004 on Build GRCh38
rs936593049 Chr.4: 17804705-17804705 on Build GRCh38
rs965541713 Chr.4: 17804907-17804907 on Build GRCh38
rs753397932 Chr.4: 17804917-17804917 on Build GRCh38
rs892171823 Chr.4: 17804769-17804769 on Build GRCh38
rs753463325 Chr.4: 17805085-17805087 on Build GRCh38
rs888514854 Chr.4: 17804677-17804677 on Build GRCh38
rs989380908 Chr.4: 17805067-17805067 on Build GRCh38
rs542330331 Chr.4: 17804658-17804658 on Build GRCh38
rs569257754 Chr.4: 17804957-17804957 on Build GRCh38
rs559945982 Chr.4: 17804747-17804747 on Build GRCh38
rs970854883 Chr.4: 17804943-17804943 on Build GRCh38
rs74667233 Chr.4: 17805101-17805104 on Build GRCh38
rs962059864 Chr.4: 17804910-17804910 on Build GRCh38
rs769415169 Chr.4: 17804666-17804666 on Build GRCh38
rs922308023 Chr.4: 17805104-17805105 on Build GRCh38
rs182377100 Chr.4: 17805093-17805093 on Build GRCh38

View Full Product Details