Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 88046553-88047052 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PKD2
Entrez Gene ID
5311
Gene Name
polycystin 2, transient receptor potential cation channel
Gene Aliases
APKD2, PC2, PKD4, Pc-2, TRPP2
UniGene
Hs.181272
Gene Chromosome Location
Chr.4: 88007647-88077779 on Build GRCh38
Transcripts
NM_000297.3 XM_011532028.2 XM_011532029.1 XM_011532030.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4625364 c.1339G>T p.A447S Chr.4: 88046661-88046661 on Build GRCh38
5401795 c.1390C>T p.R464* Chr.4: 88046712-88046712 on Build GRCh38
4397239 c.1348G>A p.G450S Chr.4: 88046670-88046670 on Build GRCh38
6168351 c.1419G>T p.L473L Chr.4: 88046741-88046741 on Build GRCh38
3696806 c.1544T>C p.V515A Chr.4: 88046866-88046866 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779504028 Chr.4: 88046612-88046612 on Build GRCh38
rs761548665 Chr.4: 88046596-88046596 on Build GRCh38
rs1131408 Chr.4: 88046668-88046668 on Build GRCh38
rs372922210 Chr.4: 88046592-88046592 on Build GRCh38
rs553111488 Chr.4: 88047001-88047001 on Build GRCh38
rs975218214 Chr.4: 88046624-88046624 on Build GRCh38
rs746049242 Chr.4: 88046809-88046809 on Build GRCh38
rs202245969 Chr.4: 88046721-88046721 on Build GRCh38
rs868456726 Chr.4: 88046684-88046684 on Build GRCh38
rs751653904 Chr.4: 88046782-88046782 on Build GRCh38
rs768723527 Chr.4: 88046838-88046838 on Build GRCh38
rs367862195 Chr.4: 88046648-88046648 on Build GRCh38
rs893730744 Chr.4: 88046999-88046999 on Build GRCh38
rs779703488 Chr.4: 88046820-88046820 on Build GRCh38
rs912607560 Chr.4: 88046824-88046824 on Build GRCh38
rs75762896 Chr.4: 88046767-88046767 on Build GRCh38
rs955223389 Chr.4: 88046964-88046964 on Build GRCh38
rs757279620 Chr.4: 88046787-88046787 on Build GRCh38
rs771670643 Chr.4: 88046916-88046916 on Build GRCh38
rs377670696 Chr.4: 88046914-88046914 on Build GRCh38
rs770946743 Chr.4: 88046599-88046601 on Build GRCh38
rs747299173 Chr.4: 88046636-88046636 on Build GRCh38
rs1003853373 Chr.4: 88046627-88046627 on Build GRCh38
rs121918042 Chr.4: 88046712-88046712 on Build GRCh38
rs939097843 Chr.4: 88046822-88046822 on Build GRCh38
rs747688660 Chr.4: 88046843-88046843 on Build GRCh38
rs376559147 Chr.4: 88046905-88046905 on Build GRCh38
rs754776342 Chr.4: 88046913-88046913 on Build GRCh38
rs865844739 Chr.4: 88046746-88046746 on Build GRCh38
rs767208570 Chr.4: 88046597-88046597 on Build GRCh38
rs201504575 Chr.4: 88046766-88046766 on Build GRCh38
rs772965121 Chr.4: 88046847-88046847 on Build GRCh38
rs144390370 Chr.4: 88046713-88046713 on Build GRCh38
rs755990722 Chr.4: 88046812-88046812 on Build GRCh38
rs761346302 Chr.4: 88046720-88046720 on Build GRCh38
rs17013754 Chr.4: 88046681-88046681 on Build GRCh38
rs745700907 Chr.4: 88046664-88046664 on Build GRCh38
rs760230159 Chr.4: 88046856-88046856 on Build GRCh38
rs768345552 Chr.4: 88046696-88046696 on Build GRCh38
rs749114897 Chr.4: 88046910-88046910 on Build GRCh38
rs781335033 Chr.4: 88046658-88046658 on Build GRCh38
rs750719254 Chr.4: 88046898-88046898 on Build GRCh38
rs1033463795 Chr.4: 88046759-88046759 on Build GRCh38
rs777817801 Chr.4: 88046621-88046621 on Build GRCh38
rs368920605 Chr.4: 88046915-88046915 on Build GRCh38
rs999100043 Chr.4: 88046989-88046989 on Build GRCh38
rs529196998 Chr.4: 88046690-88046690 on Build GRCh38
rs749042509 Chr.4: 88046823-88046823 on Build GRCh38
rs528130217 Chr.4: 88046577-88046577 on Build GRCh38
rs143581690 Chr.4: 88046868-88046868 on Build GRCh38
rs977431912 Chr.4: 88046808-88046808 on Build GRCh38
rs759983737 Chr.4: 88046737-88046737 on Build GRCh38
rs886041114 Chr.4: 88046642-88046642 on Build GRCh38
rs755246607 Chr.4: 88046607-88046607 on Build GRCh38
rs765496869 Chr.4: 88046755-88046755 on Build GRCh38
rs1029501022 Chr.4: 88046935-88046935 on Build GRCh38
rs899740654 Chr.4: 88046886-88046886 on Build GRCh38
rs750002416 Chr.4: 88046735-88046735 on Build GRCh38
rs753113036 Chr.4: 88046613-88046613 on Build GRCh38
rs1801612 Chr.4: 88046676-88046676 on Build GRCh38
rs369470196 Chr.4: 88046887-88046887 on Build GRCh38
rs746121408 Chr.4: 88046726-88046726 on Build GRCh38
rs1002113808 Chr.4: 88046739-88046739 on Build GRCh38
rs139753109 Chr.4: 88046799-88046799 on Build GRCh38
rs780261536 Chr.4: 88046907-88046907 on Build GRCh38
rs763247154 Chr.4: 88046865-88046865 on Build GRCh38
rs754146126 Chr.4: 88046757-88046757 on Build GRCh38
rs773998492 Chr.4: 88046716-88046716 on Build GRCh38
rs761111484 Chr.4: 88046659-88046659 on Build GRCh38
rs121918043 Chr.4: 88046854-88046854 on Build GRCh38
rs752024467 Chr.4: 88046871-88046871 on Build GRCh38
rs376901684 Chr.4: 88046879-88046879 on Build GRCh38
rs138683448 Chr.4: 88046607-88046612 on Build GRCh38
rs754239825 Chr.4: 88046601-88046601 on Build GRCh38
rs758520988 Chr.4: 88046620-88046620 on Build GRCh38
rs145716012 Chr.4: 88046864-88046864 on Build GRCh38
rs373937061 Chr.4: 88046845-88046845 on Build GRCh38
rs772592995 Chr.4: 88046919-88046919 on Build GRCh38
rs772302417 Chr.4: 88046892-88046896 on Build GRCh38
rs1021232807 Chr.4: 88047027-88047027 on Build GRCh38
rs931474348 Chr.4: 88046862-88046862 on Build GRCh38
rs149780942 Chr.4: 88046839-88046839 on Build GRCh38
rs139700679 Chr.4: 88046711-88046711 on Build GRCh38
rs201328200 Chr.4: 88046781-88046781 on Build GRCh38
rs775489701 Chr.4: 88046898-88046904 on Build GRCh38
rs1001830856 Chr.4: 88046933-88046933 on Build GRCh38
rs1033535717 Chr.4: 88046993-88046993 on Build GRCh38

View Full Product Details