Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 88051772-88052269 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PKD2
Entrez Gene ID
5311
Gene Name
polycystin 2, transient receptor potential cation channel
Gene Aliases
APKD2, PC2, PKD4, Pc-2, TRPP2
UniGene
Hs.181272
Gene Chromosome Location
Chr.4: 88007647-88077779 on Build GRCh38
Transcripts
NM_000297.3 XM_011532028.2 XM_011532029.1 XM_011532030.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
735372 c.1696G>T p.V566L Chr.4: 88052138-88052138 on Build GRCh38
4635168 c.1557G>T p.V519V Chr.4: 88051999-88051999 on Build GRCh38
5346646 c.1583G>C p.R528T Chr.4: 88052025-88052025 on Build GRCh38
3409607 c.1564A>G p.I522V Chr.4: 88052006-88052006 on Build GRCh38
3826416 c.1637C>T p.P546L Chr.4: 88052079-88052079 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1030058583 Chr.4: 88051886-88051886 on Build GRCh38
rs773726125 Chr.4: 88052082-88052082 on Build GRCh38
rs2728104 Chr.4: 88051854-88051854 on Build GRCh38
rs113552046 Chr.4: 88051957-88051957 on Build GRCh38
rs764415003 Chr.4: 88051959-88051959 on Build GRCh38
rs577408238 Chr.4: 88051870-88051870 on Build GRCh38
rs765487433 Chr.4: 88052098-88052098 on Build GRCh38
rs773865084 Chr.4: 88051834-88051834 on Build GRCh38
rs561247655 Chr.4: 88052193-88052193 on Build GRCh38
rs754868200 Chr.4: 88052022-88052022 on Build GRCh38
rs760705469 Chr.4: 88052083-88052083 on Build GRCh38
rs1054620336 Chr.4: 88052176-88052176 on Build GRCh38
rs930075115 Chr.4: 88052078-88052078 on Build GRCh38
rs776300102 Chr.4: 88051951-88051951 on Build GRCh38
rs770436317 Chr.4: 88052041-88052041 on Build GRCh38
rs930768993 Chr.4: 88051806-88051806 on Build GRCh38
rs906120327 Chr.4: 88051838-88051838 on Build GRCh38
rs537588879 Chr.4: 88051842-88051842 on Build GRCh38
rs575988557 Chr.4: 88051828-88051828 on Build GRCh38
rs761774250 Chr.4: 88052185-88052185 on Build GRCh38
rs763927072 Chr.4: 88051984-88051984 on Build GRCh38
rs754169081 Chr.4: 88052091-88052091 on Build GRCh38
rs370420329 Chr.4: 88051970-88051970 on Build GRCh38
rs779744978 Chr.4: 88052172-88052172 on Build GRCh38
rs377348948 Chr.4: 88052221-88052221 on Build GRCh38
rs893437481 Chr.4: 88052228-88052228 on Build GRCh38
rs369266202 Chr.4: 88051844-88051844 on Build GRCh38
rs183202128 Chr.4: 88051837-88051837 on Build GRCh38
rs377454143 Chr.4: 88052087-88052087 on Build GRCh38
rs777708333 Chr.4: 88052040-88052040 on Build GRCh38
rs760530043 Chr.4: 88051956-88051957 on Build GRCh38
rs374402854 Chr.4: 88052006-88052006 on Build GRCh38
rs756934555 Chr.4: 88051864-88051864 on Build GRCh38
rs79352425 Chr.4: 88051874-88051874 on Build GRCh38
rs762299492 Chr.4: 88051972-88051972 on Build GRCh38
rs918671443 Chr.4: 88051818-88051818 on Build GRCh38
rs748489649 Chr.4: 88052191-88052191 on Build GRCh38
rs758273926 Chr.4: 88052036-88052036 on Build GRCh38
rs990728570 Chr.4: 88051973-88051973 on Build GRCh38
rs907849580 Chr.4: 88051816-88051816 on Build GRCh38
rs777603721 Chr.4: 88052125-88052125 on Build GRCh38
rs369908107 Chr.4: 88052096-88052096 on Build GRCh38
rs145297759 Chr.4: 88052059-88052059 on Build GRCh38
rs780865432 Chr.4: 88052042-88052042 on Build GRCh38
rs901376210 Chr.4: 88051867-88051867 on Build GRCh38
rs758164143 Chr.4: 88052116-88052116 on Build GRCh38
rs778836129 Chr.4: 88052028-88052028 on Build GRCh38
rs148920907 Chr.4: 88051997-88051997 on Build GRCh38
rs374450718 Chr.4: 88051987-88051987 on Build GRCh38
rs75331451 Chr.4: 88051944-88051944 on Build GRCh38
rs751486110 Chr.4: 88052136-88052136 on Build GRCh38
rs1053375327 Chr.4: 88052222-88052222 on Build GRCh38
rs749927657 Chr.4: 88052205-88052205 on Build GRCh38
rs150924889 Chr.4: 88052180-88052180 on Build GRCh38
rs552483463 Chr.4: 88052047-88052047 on Build GRCh38
rs758358935 Chr.4: 88051809-88051809 on Build GRCh38
rs767706213 Chr.4: 88051975-88051975 on Build GRCh38
rs752554282 Chr.4: 88052100-88052100 on Build GRCh38
rs776718970 Chr.4: 88051993-88051993 on Build GRCh38
rs983462333 Chr.4: 88052048-88052048 on Build GRCh38
rs984104344 Chr.4: 88052102-88052102 on Build GRCh38
rs764677571 Chr.4: 88052204-88052205 on Build GRCh38
rs554558249 Chr.4: 88051821-88051821 on Build GRCh38
rs534844913 Chr.4: 88052075-88052075 on Build GRCh38
rs145498002 Chr.4: 88051920-88051920 on Build GRCh38
rs147630070 Chr.4: 88052076-88052076 on Build GRCh38
rs909924001 Chr.4: 88052150-88052150 on Build GRCh38
rs375543446 Chr.4: 88052029-88052029 on Build GRCh38
rs769071283 Chr.4: 88052046-88052046 on Build GRCh38
rs770503297 Chr.4: 88051945-88051945 on Build GRCh38
rs773637938 Chr.4: 88052201-88052201 on Build GRCh38
rs761228697 Chr.4: 88052205-88052205 on Build GRCh38
rs1010023800 Chr.4: 88051881-88051881 on Build GRCh38
rs1048440582 Chr.4: 88051835-88051835 on Build GRCh38
rs148433182 Chr.4: 88051933-88051933 on Build GRCh38
rs142580445 Chr.4: 88052175-88052175 on Build GRCh38

View Full Product Details