Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 107989587-107990128 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HADH
Entrez Gene ID
3033
Gene Name
hydroxyacyl-CoA dehydrogenase
Gene Aliases
HAD, HADH1, HADHSC, HCDH, HHF4, MSCHAD, SCHAD
UniGene
Hs.438289
Gene Chromosome Location
Chr.4: 107989714-108035175 on Build GRCh38
Transcripts
NM_001184705.2 NM_005327.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6420624 c.110T>C p.L37P Chr.4: 107990042-107990042 on Build GRCh38
5958926 c.92delC p.T31fs*7 Chr.4: 107990024-107990024 on Build GRCh38
7144280 c.62C>G p.S21W Chr.4: 107989994-107989994 on Build GRCh38
1209270 c.56C>T p.S19L Chr.4: 107989988-107989988 on Build GRCh38
6420625 c.110T>C p.L37P Chr.4: 107990042-107990042 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376518945 Chr.4: 107989898-107989898 on Build GRCh38
rs565804691 Chr.4: 107989858-107989858 on Build GRCh38
rs768559344 Chr.4: 107989875-107989875 on Build GRCh38
rs981960508 Chr.4: 107989732-107989732 on Build GRCh38
rs764905879 Chr.4: 107989889-107989889 on Build GRCh38
rs749548991 Chr.4: 107989911-107989911 on Build GRCh38
rs748294294 Chr.4: 107989859-107989860 on Build GRCh38
rs950643391 Chr.4: 107990083-107990083 on Build GRCh38
rs748286204 Chr.4: 107989899-107989899 on Build GRCh38
rs776852612 Chr.4: 107989876-107989876 on Build GRCh38
rs1054046630 Chr.4: 107989820-107989820 on Build GRCh38
rs1030732728 Chr.4: 107989866-107989866 on Build GRCh38
rs745742772 Chr.4: 107989813-107989813 on Build GRCh38
rs745696025 Chr.4: 107989867-107989867 on Build GRCh38
rs772843585 Chr.4: 107990070-107990070 on Build GRCh38
rs985119383 Chr.4: 107989659-107989659 on Build GRCh38
rs755018737 Chr.4: 107989830-107989830 on Build GRCh38
rs759364616 Chr.4: 107990089-107990089 on Build GRCh38
rs938663402 Chr.4: 107989888-107989888 on Build GRCh38
rs777924030 Chr.4: 107990024-107990024 on Build GRCh38
rs953885090 Chr.4: 107989652-107989652 on Build GRCh38
rs771772152 Chr.4: 107990066-107990067 on Build GRCh38
rs896957328 Chr.4: 107989635-107989635 on Build GRCh38
rs750884066 Chr.4: 107989838-107989838 on Build GRCh38
rs1037062665 Chr.4: 107989769-107989769 on Build GRCh38
rs926541644 Chr.4: 107989650-107989650 on Build GRCh38
rs770111560 Chr.4: 107989904-107989904 on Build GRCh38
rs775923392 Chr.4: 107990056-107990056 on Build GRCh38
rs1026883928 Chr.4: 107989646-107989646 on Build GRCh38
rs774673150 Chr.4: 107989879-107989879 on Build GRCh38
rs926097 Chr.4: 107990085-107990085 on Build GRCh38
rs981993093 Chr.4: 107989648-107989648 on Build GRCh38
rs781319494 Chr.4: 107990004-107990004 on Build GRCh38
rs372250758 Chr.4: 107989683-107989683 on Build GRCh38
rs752625929 Chr.4: 107990108-107990108 on Build GRCh38
rs1041175521 Chr.4: 107989807-107989807 on Build GRCh38
rs924787579 Chr.4: 107989689-107989689 on Build GRCh38
rs145521658 Chr.4: 107990035-107990035 on Build GRCh38
rs146017663 Chr.4: 107989950-107989950 on Build GRCh38
rs574132278 Chr.4: 107989882-107989882 on Build GRCh38
rs571161759 Chr.4: 107989871-107989871 on Build GRCh38
rs74428123 Chr.4: 107990031-107990031 on Build GRCh38
rs907304228 Chr.4: 107989622-107989622 on Build GRCh38
rs28703310 Chr.4: 107989868-107989868 on Build GRCh38
rs1032418920 Chr.4: 107989710-107989710 on Build GRCh38
rs749063053 Chr.4: 107989927-107989928 on Build GRCh38
rs761268836 Chr.4: 107989967-107989967 on Build GRCh38
rs372068563 Chr.4: 107990105-107990105 on Build GRCh38
rs536959577 Chr.4: 107989882-107989882 on Build GRCh38
rs754944563 Chr.4: 107989995-107989995 on Build GRCh38
rs567325509 Chr.4: 107989931-107989931 on Build GRCh38
rs763502531 Chr.4: 107989844-107989844 on Build GRCh38
rs376591232 Chr.4: 107989730-107989730 on Build GRCh38
rs1003322641 Chr.4: 107989843-107989843 on Build GRCh38
rs995855684 Chr.4: 107989639-107989639 on Build GRCh38
rs537335460 Chr.4: 107989818-107989818 on Build GRCh38
rs751280475 Chr.4: 107990087-107990087 on Build GRCh38
rs753433361 Chr.4: 107989860-107989860 on Build GRCh38
rs886058980 Chr.4: 107989953-107989953 on Build GRCh38
rs900111101 Chr.4: 107989696-107989696 on Build GRCh38
rs111537232 Chr.4: 107989691-107989691 on Build GRCh38
rs563383731 Chr.4: 107989927-107989927 on Build GRCh38
rs900864178 Chr.4: 107990052-107990052 on Build GRCh38
rs746985153 Chr.4: 107989874-107989874 on Build GRCh38
rs760705185 Chr.4: 107989828-107989828 on Build GRCh38
rs1002873270 Chr.4: 107989623-107989623 on Build GRCh38
rs948950853 Chr.4: 107989719-107989719 on Build GRCh38
rs879941987 Chr.4: 107990084-107990085 on Build GRCh38
rs907188251 Chr.4: 107989738-107989738 on Build GRCh38
rs749983874 Chr.4: 107989977-107989977 on Build GRCh38
rs765919410 Chr.4: 107989741-107989741 on Build GRCh38
rs774784972 Chr.4: 107989921-107989921 on Build GRCh38
rs1009249997 Chr.4: 107989829-107989829 on Build GRCh38
rs761180931 Chr.4: 107990059-107990059 on Build GRCh38
rs570893633 Chr.4: 107989688-107989688 on Build GRCh38
rs760216198 Chr.4: 107990087-107990088 on Build GRCh38
rs752222234 Chr.4: 107989856-107989856 on Build GRCh38
rs775613409 Chr.4: 107989822-107989822 on Build GRCh38
rs201600831 Chr.4: 107989982-107989982 on Build GRCh38
rs10017687 Chr.4: 107989740-107989740 on Build GRCh38
rs990341309 Chr.4: 107989870-107989870 on Build GRCh38
rs770646232 Chr.4: 107989969-107989971 on Build GRCh38
rs145598361 Chr.4: 107989615-107989615 on Build GRCh38
rs748128617 Chr.4: 107990008-107990008 on Build GRCh38
rs374248298 Chr.4: 107989979-107989979 on Build GRCh38
rs913723815 Chr.4: 107989634-107989635 on Build GRCh38
rs756893286 Chr.4: 107989886-107989886 on Build GRCh38
rs760104610 Chr.4: 107989880-107989880 on Build GRCh38
rs770197760 Chr.4: 107989877-107989877 on Build GRCh38
rs970448250 Chr.4: 107989836-107989836 on Build GRCh38
rs534966818 Chr.4: 107990071-107990071 on Build GRCh38
rs201094947 Chr.4: 107990092-107990092 on Build GRCh38
rs1019310746 Chr.4: 107989853-107989853 on Build GRCh38
rs758077730 Chr.4: 107989894-107989894 on Build GRCh38
rs773991596 Chr.4: 107989982-107989987 on Build GRCh38
rs528952923 Chr.4: 107989847-107989847 on Build GRCh38
rs778695590 Chr.4: 107989810-107989810 on Build GRCh38
rs770666363 Chr.4: 107989803-107989803 on Build GRCh38
rs764746023 Chr.4: 107989973-107989973 on Build GRCh38
rs1015398413 Chr.4: 107989627-107989627 on Build GRCh38
rs777890650 Chr.4: 107989892-107989906 on Build GRCh38
rs769224699 Chr.4: 107990060-107990060 on Build GRCh38
rs773267851 Chr.4: 107989909-107989909 on Build GRCh38
rs995346679 Chr.4: 107989805-107989805 on Build GRCh38
rs765688267 Chr.4: 107990106-107990115 on Build GRCh38
rs759111271 Chr.4: 107990065-107990066 on Build GRCh38
rs994692222 Chr.4: 107990038-107990038 on Build GRCh38
rs766800315 Chr.4: 107989878-107989878 on Build GRCh38
rs756215599 Chr.4: 107990009-107990009 on Build GRCh38
rs17038278 Chr.4: 107989685-107989685 on Build GRCh38
rs772452366 Chr.4: 107990048-107990048 on Build GRCh38
rs563886314 Chr.4: 107989837-107989837 on Build GRCh38
rs541455934 Chr.4: 107989799-107989799 on Build GRCh38
rs760202 Chr.4: 107989862-107989862 on Build GRCh38
rs137853101 Chr.4: 107990050-107990050 on Build GRCh38
rs886058979 Chr.4: 107989738-107989738 on Build GRCh38
rs143346680 Chr.4: 107989970-107989970 on Build GRCh38
rs548783898 Chr.4: 107989857-107989857 on Build GRCh38
rs931331922 Chr.4: 107989751-107989751 on Build GRCh38
rs749406566 Chr.4: 107990027-107990027 on Build GRCh38
rs139920805 Chr.4: 107989966-107989966 on Build GRCh38
rs533668226 Chr.4: 107989817-107989817 on Build GRCh38
rs17550794 Chr.4: 107989895-107989895 on Build GRCh38
rs564766802 Chr.4: 107989812-107989812 on Build GRCh38
rs779135938 Chr.4: 107990032-107990032 on Build GRCh38
rs555570196 Chr.4: 107989984-107989984 on Build GRCh38
rs372830966 Chr.4: 107989869-107989869 on Build GRCh38
rs957813463 Chr.4: 107990096-107990096 on Build GRCh38
rs1003291250 Chr.4: 107989981-107989981 on Build GRCh38
rs1037114786 Chr.4: 107989892-107989892 on Build GRCh38
rs1018755859 Chr.4: 107989801-107989802 on Build GRCh38
rs931381209 Chr.4: 107989666-107989666 on Build GRCh38
rs535101967 Chr.4: 107989707-107989707 on Build GRCh38
rs756911636 Chr.4: 107989861-107989861 on Build GRCh38
rs767443792 Chr.4: 107990100-107990100 on Build GRCh38
rs370266024 Chr.4: 107989913-107989913 on Build GRCh38
rs766844144 Chr.4: 107989850-107989850 on Build GRCh38
rs182097151 Chr.4: 107989831-107989831 on Build GRCh38
rs781415476 Chr.4: 107989848-107989848 on Build GRCh38
rs776940529 Chr.4: 107989832-107989832 on Build GRCh38
rs746854470 Chr.4: 107989897-107989897 on Build GRCh38
rs1019258745 Chr.4: 107989660-107989660 on Build GRCh38
rs541975295 Chr.4: 107989725-107989725 on Build GRCh38
rs61735986 Chr.4: 107989768-107989768 on Build GRCh38
rs965015323 Chr.4: 107989854-107989854 on Build GRCh38
rs914821653 Chr.4: 107989714-107989714 on Build GRCh38
rs771973054 Chr.4: 107989821-107989821 on Build GRCh38
rs973688128 Chr.4: 107989852-107989852 on Build GRCh38

View Full Product Details