Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 62390655-62391154 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DIMT1
Entrez Gene ID
27292
Gene Name
DIM1 dimethyladenosine transferase 1 homolog
Gene Aliases
DIM1, DIMT1L, HSA9761, HUSSY5
UniGene
Hs.731665
Gene Chromosome Location
Chr.5: 62388523-62403939 on Build GRCh38
Transcripts
NM_014473.2 XM_011543323.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2149863 c.897delC p.R300fs*>14 Chr.5: 62390878-62390878 on Build GRCh38
3855804 c.815T>C p.I272T Chr.5: 62390960-62390960 on Build GRCh38
4559735 c.817G>A p.A273T Chr.5: 62390958-62390958 on Build GRCh38
4407484 c.793-7T>C p.? Chr.5: 62390989-62390989 on Build GRCh38
5868677 c.875C>T p.S292F Chr.5: 62390900-62390900 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs981295040 Chr.5: 62391040-62391040 on Build GRCh38
rs760056933 Chr.5: 62390843-62390843 on Build GRCh38
rs558683005 Chr.5: 62390818-62390818 on Build GRCh38
rs758259989 Chr.5: 62391020-62391020 on Build GRCh38
rs1019898184 Chr.5: 62390898-62390898 on Build GRCh38
rs149430822 Chr.5: 62391064-62391064 on Build GRCh38
rs753406288 Chr.5: 62390986-62390986 on Build GRCh38
rs915827426 Chr.5: 62390683-62390688 on Build GRCh38
rs575757232 Chr.5: 62391052-62391052 on Build GRCh38
rs778930323 Chr.5: 62390904-62390904 on Build GRCh38
rs994190323 Chr.5: 62390944-62390944 on Build GRCh38
rs539107433 Chr.5: 62390815-62390815 on Build GRCh38
rs138970333 Chr.5: 62390719-62390719 on Build GRCh38
rs775964162 Chr.5: 62390965-62390965 on Build GRCh38
rs981060930 Chr.5: 62390749-62390749 on Build GRCh38
rs542444877 Chr.5: 62391012-62391012 on Build GRCh38
rs770298254 Chr.5: 62390957-62390957 on Build GRCh38
rs750041272 Chr.5: 62390881-62390881 on Build GRCh38
rs755691846 Chr.5: 62390889-62390889 on Build GRCh38
rs554080328 Chr.5: 62390910-62390910 on Build GRCh38
rs371008256 Chr.5: 62390832-62390832 on Build GRCh38
rs928562817 Chr.5: 62391041-62391043 on Build GRCh38
rs934355822 Chr.5: 62390774-62390774 on Build GRCh38
rs763443006 Chr.5: 62390974-62390974 on Build GRCh38
rs767258344 Chr.5: 62390867-62390867 on Build GRCh38
rs756264589 Chr.5: 62390709-62390709 on Build GRCh38
rs892887670 Chr.5: 62390808-62390808 on Build GRCh38
rs540502594 Chr.5: 62390776-62390776 on Build GRCh38
rs764336943 Chr.5: 62390979-62390979 on Build GRCh38
rs747173587 Chr.5: 62390938-62390938 on Build GRCh38
rs148664994 Chr.5: 62390951-62390951 on Build GRCh38
rs765691960 Chr.5: 62390855-62390855 on Build GRCh38
rs868017897 Chr.5: 62390912-62390912 on Build GRCh38
rs374427823 Chr.5: 62391016-62391016 on Build GRCh38
rs755000236 Chr.5: 62390903-62390903 on Build GRCh38
rs778093735 Chr.5: 62390917-62390917 on Build GRCh38
rs372819795 Chr.5: 62391043-62391043 on Build GRCh38
rs754483907 Chr.5: 62391009-62391009 on Build GRCh38
rs534698558 Chr.5: 62390859-62390859 on Build GRCh38
rs536894522 Chr.5: 62390726-62390726 on Build GRCh38
rs1008506359 Chr.5: 62390866-62390866 on Build GRCh38
rs556773433 Chr.5: 62390793-62390793 on Build GRCh38
rs759225708 Chr.5: 62390753-62390753 on Build GRCh38
rs749525056 Chr.5: 62390999-62390999 on Build GRCh38
rs771157668 Chr.5: 62391019-62391019 on Build GRCh38
rs562311773 Chr.5: 62391034-62391034 on Build GRCh38
rs767739835 Chr.5: 62390861-62390862 on Build GRCh38
rs750140575 Chr.5: 62390982-62390982 on Build GRCh38
rs777448341 Chr.5: 62391033-62391033 on Build GRCh38
rs753351247 Chr.5: 62390895-62390895 on Build GRCh38
rs115925120 Chr.5: 62390794-62390794 on Build GRCh38
rs763169283 Chr.5: 62390861-62390861 on Build GRCh38
rs760270737 Chr.5: 62390983-62390983 on Build GRCh38
rs2257389 Chr.5: 62390820-62390820 on Build GRCh38
rs759545716 Chr.5: 62390956-62390956 on Build GRCh38
rs779784803 Chr.5: 62391046-62391046 on Build GRCh38
rs777168295 Chr.5: 62390825-62390825 on Build GRCh38

View Full Product Details