Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 79640671-79641183 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PAPD4
Entrez Gene ID
167153
Gene Name
PAP associated domain containing 4
Gene Aliases
GLD2, TUT2
UniGene
Hs.418198
Gene Chromosome Location
Chr.5: 79612416-79686648 on Build GRCh38
Transcripts
NM_001114393.1 NM_001114394.1 NM_001297744.1 NM_001297745.1 NM_173797.3 XM_011543219.2 XM_011543220.2 XM_011543221.2 XM_011543222.2 XM_011543223.2 XM_011543224.2 XM_011543225.2 XM_011543226.2 XM_011543227.2 XM_011543228.2 XM_011543229.2 XM_011543230.2 XM_011543231.2 XM_011543232.2 XM_011543233.2 XM_017009151.1 XM_017009152.1 XM_017009153.1 XM_017009154.1 XM_017009155.1 XM_017009156.1 XM_017009157.1 XM_017009158.1 XM_017009159.1 XM_017009160.1 XM_017009161.1 XM_017009162.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5495808 c.532G>C p.E178Q Chr.5: 79640917-79640917 on Build GRCh38
483081 c.578C>T p.P193L Chr.5: 79640963-79640963 on Build GRCh38
4006062 c.617G>T p.G206V Chr.5: 79641141-79641141 on Build GRCh38
7181947 c.468G>A p.L156L Chr.5: 79640853-79640853 on Build GRCh38
1219222 c.620T>G p.F207C Chr.5: 79641144-79641144 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs547181173 Chr.5: 79641015-79641015 on Build GRCh38
rs112011692 Chr.5: 79640776-79640776 on Build GRCh38
rs745889213 Chr.5: 79641068-79641068 on Build GRCh38
rs778958719 Chr.5: 79641073-79641073 on Build GRCh38
rs751793561 Chr.5: 79640975-79640975 on Build GRCh38
rs748969844 Chr.5: 79640823-79640824 on Build GRCh38
rs753234338 Chr.5: 79640954-79640954 on Build GRCh38
rs201191682 Chr.5: 79640958-79640958 on Build GRCh38
rs1041657408 Chr.5: 79641030-79641030 on Build GRCh38
rs983069116 Chr.5: 79640889-79640889 on Build GRCh38
rs1032754111 Chr.5: 79640758-79640758 on Build GRCh38
rs990271347 Chr.5: 79640821-79640821 on Build GRCh38
rs879363444 Chr.5: 79641025-79641025 on Build GRCh38
rs375395619 Chr.5: 79640782-79640782 on Build GRCh38
rs141449959 Chr.5: 79640726-79640726 on Build GRCh38
rs565317621 Chr.5: 79640768-79640768 on Build GRCh38
rs1025873033 Chr.5: 79640772-79640772 on Build GRCh38
rs770519264 Chr.5: 79640829-79640829 on Build GRCh38
rs769333063 Chr.5: 79641150-79641150 on Build GRCh38
rs763427631 Chr.5: 79641153-79641153 on Build GRCh38
rs759059333 Chr.5: 79640923-79640923 on Build GRCh38
rs77993880 Chr.5: 79641079-79641079 on Build GRCh38
rs764416651 Chr.5: 79640971-79640971 on Build GRCh38
rs372529761 Chr.5: 79640969-79640969 on Build GRCh38
rs749234860 Chr.5: 79640820-79640820 on Build GRCh38
rs932725218 Chr.5: 79640965-79640965 on Build GRCh38
rs768650053 Chr.5: 79640828-79640828 on Build GRCh38
rs781570996 Chr.5: 79641017-79641017 on Build GRCh38
rs752481773 Chr.5: 79641147-79641147 on Build GRCh38
rs183803124 Chr.5: 79640791-79640791 on Build GRCh38
rs373046925 Chr.5: 79641154-79641154 on Build GRCh38
rs757521589 Chr.5: 79640778-79640778 on Build GRCh38
rs942374422 Chr.5: 79640978-79640978 on Build GRCh38
rs769270213 Chr.5: 79640916-79640917 on Build GRCh38
rs902837241 Chr.5: 79641061-79641061 on Build GRCh38
rs765682008 Chr.5: 79640948-79640948 on Build GRCh38
rs78953757 Chr.5: 79641089-79641089 on Build GRCh38
rs142919591 Chr.5: 79641115-79641115 on Build GRCh38
rs116627352 Chr.5: 79640838-79640838 on Build GRCh38
rs746821002 Chr.5: 79641090-79641090 on Build GRCh38
rs756287243 Chr.5: 79641072-79641072 on Build GRCh38
rs763792902 Chr.5: 79640827-79640827 on Build GRCh38
rs770511074 Chr.5: 79641095-79641095 on Build GRCh38
rs765905954 Chr.5: 79640916-79640916 on Build GRCh38
rs760299421 Chr.5: 79640900-79640900 on Build GRCh38
rs889437577 Chr.5: 79640711-79640711 on Build GRCh38
rs757490464 Chr.5: 79641001-79641001 on Build GRCh38
rs1050218290 Chr.5: 79640999-79640999 on Build GRCh38
rs772894889 Chr.5: 79640892-79640892 on Build GRCh38
rs1006630659 Chr.5: 79640725-79640727 on Build GRCh38
rs933617138 Chr.5: 79640950-79640950 on Build GRCh38
rs147020973 Chr.5: 79640746-79640746 on Build GRCh38
rs188344315 Chr.5: 79640919-79640919 on Build GRCh38
rs192695597 Chr.5: 79641037-79641037 on Build GRCh38
rs1034264397 Chr.5: 79641086-79641089 on Build GRCh38
rs771798862 Chr.5: 79640869-79640869 on Build GRCh38
rs759006283 Chr.5: 79641149-79641149 on Build GRCh38
rs994302213 Chr.5: 79641062-79641062 on Build GRCh38
rs773898358 Chr.5: 79640828-79640829 on Build GRCh38
rs151126262 Chr.5: 79641152-79641152 on Build GRCh38
rs983421006 Chr.5: 79641130-79641130 on Build GRCh38
rs200256692 Chr.5: 79641080-79641080 on Build GRCh38
rs202103836 Chr.5: 79640859-79640859 on Build GRCh38
rs1034377471 Chr.5: 79640809-79640809 on Build GRCh38
rs747930740 Chr.5: 79640853-79640853 on Build GRCh38
rs879632313 Chr.5: 79640705-79640706 on Build GRCh38
rs1038455203 Chr.5: 79640946-79640946 on Build GRCh38
rs745335099 Chr.5: 79640913-79640914 on Build GRCh38
rs1009204503 Chr.5: 79641023-79641023 on Build GRCh38
rs958859912 Chr.5: 79640770-79640770 on Build GRCh38
rs375373505 Chr.5: 79640814-79640814 on Build GRCh38

View Full Product Details