Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 97166789-97167301 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RIOK2
Entrez Gene ID
55781
Gene Name
RIO kinase 2
Gene Aliases
RIO2
UniGene
Hs.27021
Gene Chromosome Location
Chr.5: 97160867-97183301 on Build GRCh38
Transcripts
NM_001159749.1 NM_018343.2 XM_017009628.1

SNP Detail

SNP ID Chromosome Location
rs560656185 Chr.5: 97166886-97166886 on Build GRCh38
rs918163647 Chr.5: 97166922-97166922 on Build GRCh38
rs927353540 Chr.5: 97167026-97167026 on Build GRCh38
rs948281170 Chr.5: 97167188-97167188 on Build GRCh38
rs927384586 Chr.5: 97166872-97166872 on Build GRCh38
rs1053618843 Chr.5: 97167150-97167150 on Build GRCh38
rs532252361 Chr.5: 97166995-97166995 on Build GRCh38
rs1050147745 Chr.5: 97166947-97166947 on Build GRCh38
rs972268368 Chr.5: 97167077-97167077 on Build GRCh38
rs920118264 Chr.5: 97167197-97167197 on Build GRCh38
rs533961408 Chr.5: 97167204-97167204 on Build GRCh38
rs115410663 Chr.5: 97167200-97167200 on Build GRCh38
rs934981123 Chr.5: 97166911-97166912 on Build GRCh38
rs1000690652 Chr.5: 97166967-97166967 on Build GRCh38
rs1025061529 Chr.5: 97167203-97167203 on Build GRCh38
rs562467404 Chr.5: 97167054-97167054 on Build GRCh38
rs747972539 Chr.5: 97166845-97166845 on Build GRCh38
rs888849331 Chr.5: 97167118-97167118 on Build GRCh38
rs111844358 Chr.5: 97167265-97167265 on Build GRCh38
rs996024262 Chr.5: 97167024-97167024 on Build GRCh38
rs1036027510 Chr.5: 97166929-97166929 on Build GRCh38
rs1034338790 Chr.5: 97167215-97167215 on Build GRCh38
rs936456007 Chr.5: 97167126-97167126 on Build GRCh38
rs141340615 Chr.5: 97167166-97167166 on Build GRCh38
rs1032612630 Chr.5: 97166970-97166970 on Build GRCh38
rs139387663 Chr.5: 97166903-97166903 on Build GRCh38
rs184655799 Chr.5: 97166821-97166821 on Build GRCh38
rs1050115275 Chr.5: 97167105-97167105 on Build GRCh38
rs548451273 Chr.5: 97167148-97167148 on Build GRCh38
rs769415330 Chr.5: 97166937-97166937 on Build GRCh38
rs188329192 Chr.5: 97166925-97166925 on Build GRCh38
rs997184563 Chr.5: 97166939-97166939 on Build GRCh38
rs868522421 Chr.5: 97166999-97166999 on Build GRCh38
rs529687314 Chr.5: 97167070-97167070 on Build GRCh38
rs1024824184 Chr.5: 97167193-97167193 on Build GRCh38
rs540827612 Chr.5: 97167270-97167270 on Build GRCh38
rs1002817773 Chr.5: 97167214-97167214 on Build GRCh38
rs979617089 Chr.5: 97167052-97167052 on Build GRCh38
rs909455944 Chr.5: 97166915-97166915 on Build GRCh38
rs888880374 Chr.5: 97166952-97166952 on Build GRCh38
rs959073943 Chr.5: 97167034-97167034 on Build GRCh38
rs879481397 Chr.5: 97166913-97166914 on Build GRCh38
rs1008228545 Chr.5: 97166974-97166974 on Build GRCh38
rs1029217639 Chr.5: 97167033-97167033 on Build GRCh38
rs887128906 Chr.5: 97166951-97166951 on Build GRCh38
rs921708039 Chr.5: 97166851-97166851 on Build GRCh38
rs937487445 Chr.5: 97166882-97166882 on Build GRCh38
rs979137401 Chr.5: 97166900-97166900 on Build GRCh38
rs145313931 Chr.5: 97167049-97167049 on Build GRCh38
rs981427831 Chr.5: 97167016-97167016 on Build GRCh38
rs1045250985 Chr.5: 97167191-97167191 on Build GRCh38
rs971332780 Chr.5: 97167007-97167007 on Build GRCh38
rs545115588 Chr.5: 97166868-97166868 on Build GRCh38
rs974971812 Chr.5: 97166840-97166840 on Build GRCh38
rs892259795 Chr.5: 97167164-97167164 on Build GRCh38
rs932942841 Chr.5: 97167090-97167090 on Build GRCh38
rs774106938 Chr.5: 97167078-97167078 on Build GRCh38
rs772588887 Chr.5: 97166938-97166938 on Build GRCh38
rs150042014 Chr.5: 97166978-97166978 on Build GRCh38

View Full Product Details