Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 111729475-111729974 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NREP
Entrez Gene ID
9315
Gene Name
neuronal regeneration related protein
Gene Aliases
C5orf13, D4S114, P311, PRO1873, PTZ17, SEZ17
UniGene
Hs.36053
Gene Chromosome Location
Chr.5: 111729303-111976931 on Build GRCh38
Transcripts
NM_001142474.1 NM_001142475.1 NM_001142476.1 NM_001142477.1 NM_001142478.1 NM_001142479.1 NM_001142480.1 NM_001142481.1 NM_001142482.1 NM_001142483.1 NM_004772.2 XM_006714732.1 XM_006714733.2 NR_040093.1

Gene Symbol
STARD4-AS1
Entrez Gene ID
100505678
Gene Name
STARD4 antisense RNA 1
Gene Aliases
-
UniGene
Hs.745061
Gene Chromosome Location
Chr.5: 111512226-111739726 on Build GRCh38
Transcripts
NM_001142474.1 NM_001142475.1 NM_001142476.1 NM_001142477.1 NM_001142478.1 NM_001142479.1 NM_001142480.1 NM_001142481.1 NM_001142482.1 NM_001142483.1 NM_004772.2 XM_006714732.1 XM_006714733.2 NR_040093.1

SNP Detail

SNP ID Chromosome Location
rs965300581 Chr.5: 111729786-111729786 on Build GRCh38
rs529849888 Chr.5: 111729627-111729627 on Build GRCh38
rs762530556 Chr.5: 111729817-111729817 on Build GRCh38
rs906219552 Chr.5: 111729863-111729863 on Build GRCh38
rs573353577 Chr.5: 111729603-111729604 on Build GRCh38
rs71579156 Chr.5: 111729628-111729628 on Build GRCh38
rs1008955441 Chr.5: 111729944-111729944 on Build GRCh38
rs906768854 Chr.5: 111729638-111729638 on Build GRCh38
rs553375967 Chr.5: 111729805-111729806 on Build GRCh38
rs972562944 Chr.5: 111729536-111729536 on Build GRCh38
rs32253 Chr.5: 111729900-111729900 on Build GRCh38
rs1031148196 Chr.5: 111729654-111729654 on Build GRCh38
rs547710403 Chr.5: 111729609-111729609 on Build GRCh38
rs771401766 Chr.5: 111729804-111729804 on Build GRCh38
rs918433412 Chr.5: 111729540-111729540 on Build GRCh38
rs537171088 Chr.5: 111729662-111729662 on Build GRCh38
rs1029141449 Chr.5: 111729913-111729913 on Build GRCh38
rs889186046 Chr.5: 111729935-111729935 on Build GRCh38
rs539275151 Chr.5: 111729818-111729818 on Build GRCh38
rs10779 Chr.5: 111729649-111729649 on Build GRCh38
rs569752119 Chr.5: 111729650-111729650 on Build GRCh38
rs1029026900 Chr.5: 111729746-111729746 on Build GRCh38
rs541987908 Chr.5: 111729899-111729899 on Build GRCh38
rs916312986 Chr.5: 111729833-111729833 on Build GRCh38
rs984008379 Chr.5: 111729823-111729823 on Build GRCh38
rs781015600 Chr.5: 111729513-111729513 on Build GRCh38
rs1053126022 Chr.5: 111729882-111729882 on Build GRCh38
rs775636160 Chr.5: 111729596-111729597 on Build GRCh38
rs947697211 Chr.5: 111729616-111729616 on Build GRCh38
rs201168405 Chr.5: 111729616-111729616 on Build GRCh38
rs184848981 Chr.5: 111729752-111729752 on Build GRCh38
rs567619803 Chr.5: 111729615-111729615 on Build GRCh38
rs947811522 Chr.5: 111729835-111729836 on Build GRCh38
rs755219555 Chr.5: 111729819-111729819 on Build GRCh38
rs4618 Chr.5: 111729731-111729731 on Build GRCh38
rs1046070420 Chr.5: 111729851-111729851 on Build GRCh38
rs144163346 Chr.5: 111729520-111729520 on Build GRCh38
rs754905553 Chr.5: 111729509-111729509 on Build GRCh38
rs553580779 Chr.5: 111729781-111729781 on Build GRCh38
rs1048625637 Chr.5: 111729604-111729604 on Build GRCh38
rs534330002 Chr.5: 111729753-111729753 on Build GRCh38
rs769447249 Chr.5: 111729644-111729644 on Build GRCh38
rs192564761 Chr.5: 111729637-111729637 on Build GRCh38
rs538319377 Chr.5: 111729662-111729663 on Build GRCh38
rs780816127 Chr.5: 111729565-111729565 on Build GRCh38
rs931037824 Chr.5: 111729563-111729563 on Build GRCh38
rs554467199 Chr.5: 111729880-111729880 on Build GRCh38
rs748200880 Chr.5: 111729614-111729614 on Build GRCh38
rs973062658 Chr.5: 111729812-111729812 on Build GRCh38

View Full Product Details