Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 149994048-149994598 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TIGD6
Entrez Gene ID
81789
Gene Name
tigger transposable element derived 6
Gene Aliases
-
UniGene
Hs.169333
Gene Chromosome Location
Chr.5: 149993123-150001167 on Build GRCh38
Transcripts
NM_001243253.1 NM_030953.3

SNP Detail

SNP ID Chromosome Location
rs147857620 Chr.5: 149994370-149994370 on Build GRCh38
rs73798339 Chr.5: 149994177-149994177 on Build GRCh38
rs898215395 Chr.5: 149994250-149994250 on Build GRCh38
rs966865565 Chr.5: 149994172-149994172 on Build GRCh38
rs374047127 Chr.5: 149994456-149994456 on Build GRCh38
rs531565415 Chr.5: 149994406-149994406 on Build GRCh38
rs950946863 Chr.5: 149994337-149994337 on Build GRCh38
rs377095661 Chr.5: 149994089-149994089 on Build GRCh38
rs10036377 Chr.5: 149994453-149994453 on Build GRCh38
rs968501993 Chr.5: 149994436-149994436 on Build GRCh38
rs1028204865 Chr.5: 149994318-149994318 on Build GRCh38
rs750295950 Chr.5: 149994412-149994412 on Build GRCh38
rs1047691231 Chr.5: 149994081-149994081 on Build GRCh38
rs762651458 Chr.5: 149994194-149994194 on Build GRCh38
rs902675550 Chr.5: 149994477-149994477 on Build GRCh38
rs185905809 Chr.5: 149994168-149994168 on Build GRCh38
rs915589254 Chr.5: 149994212-149994212 on Build GRCh38
rs570246230 Chr.5: 149994158-149994158 on Build GRCh38
rs1011947954 Chr.5: 149994204-149994204 on Build GRCh38
rs868016821 Chr.5: 149994280-149994280 on Build GRCh38
rs553407742 Chr.5: 149994269-149994269 on Build GRCh38
rs191395649 Chr.5: 149994457-149994457 on Build GRCh38
rs1006659371 Chr.5: 149994153-149994153 on Build GRCh38
rs773374090 Chr.5: 149994244-149994244 on Build GRCh38
rs946836023 Chr.5: 149994140-149994140 on Build GRCh38
rs917810651 Chr.5: 149994486-149994486 on Build GRCh38
rs10041944 Chr.5: 149994456-149994456 on Build GRCh38
rs760939396 Chr.5: 149994266-149994266 on Build GRCh38
rs73798340 Chr.5: 149994459-149994459 on Build GRCh38
rs1019181016 Chr.5: 149994369-149994369 on Build GRCh38
rs1051808884 Chr.5: 149994468-149994468 on Build GRCh38
rs764425339 Chr.5: 149994563-149994563 on Build GRCh38
rs1005089994 Chr.5: 149994340-149994340 on Build GRCh38
rs928423965 Chr.5: 149994315-149994315 on Build GRCh38
rs766853277 Chr.5: 149994316-149994317 on Build GRCh38
rs778944463 Chr.5: 149994143-149994143 on Build GRCh38
rs907264048 Chr.5: 149994203-149994203 on Build GRCh38
rs770967190 Chr.5: 149994268-149994268 on Build GRCh38
rs577931817 Chr.5: 149994354-149994354 on Build GRCh38
rs929153529 Chr.5: 149994336-149994336 on Build GRCh38
rs541705144 Chr.5: 149994181-149994181 on Build GRCh38
rs1030417574 Chr.5: 149994171-149994171 on Build GRCh38
rs537231742 Chr.5: 149994163-149994163 on Build GRCh38
rs767228734 Chr.5: 149994321-149994321 on Build GRCh38
rs957059233 Chr.5: 149994196-149994196 on Build GRCh38
rs532723717 Chr.5: 149994285-149994285 on Build GRCh38
rs1056432295 Chr.5: 149994195-149994195 on Build GRCh38
rs181850661 Chr.5: 149994083-149994083 on Build GRCh38
rs947969282 Chr.5: 149994489-149994489 on Build GRCh38
rs1044713346 Chr.5: 149994523-149994523 on Build GRCh38

View Full Product Details