Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 177065632-177066140 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF346
Entrez Gene ID
23567
Gene Name
zinc finger protein 346
Gene Aliases
JAZ, Zfp346
UniGene
Hs.484259
Gene Chromosome Location
Chr.5: 177022680-177081189 on Build GRCh38
Transcripts
NM_001308213.1 NM_001308214.1 NM_001308215.1 NM_001308216.1 NM_001308218.1 NM_001308219.1 NM_001308221.1 NM_001308223.1 NM_012279.3 NR_131773.1 NR_131774.1 XM_005265860.2 XM_005265864.3 XM_011534494.1 XM_011534495.2 XM_011534496.2 XM_017009294.1 XM_017009295.1 XM_017009296.1 XM_017009297.1 XM_017009299.1 XM_017009300.1 XM_017009301.1 XM_017009302.1 XM_017009303.1

SNP Detail

SNP ID Chromosome Location
rs912870953 Chr.5: 177065877-177065882 on Build GRCh38
rs574636511 Chr.5: 177066015-177066015 on Build GRCh38
rs1004927477 Chr.5: 177066104-177066104 on Build GRCh38
rs891328778 Chr.5: 177065806-177065806 on Build GRCh38
rs537683314 Chr.5: 177065918-177065918 on Build GRCh38
rs981240944 Chr.5: 177066052-177066052 on Build GRCh38
rs936819863 Chr.5: 177066002-177066002 on Build GRCh38
rs939641084 Chr.5: 177065721-177065721 on Build GRCh38
rs997126015 Chr.5: 177065790-177065790 on Build GRCh38
rs903980087 Chr.5: 177065691-177065691 on Build GRCh38
rs541954303 Chr.5: 177066078-177066078 on Build GRCh38
rs1045493594 Chr.5: 177065686-177065686 on Build GRCh38
rs168489 Chr.5: 177065882-177065882 on Build GRCh38
rs1038046227 Chr.5: 177065750-177065750 on Build GRCh38
rs1008307093 Chr.5: 177065810-177065810 on Build GRCh38
rs180732483 Chr.5: 177065894-177065894 on Build GRCh38
rs1007995998 Chr.5: 177065890-177065890 on Build GRCh38
rs759610181 Chr.5: 177065760-177065760 on Build GRCh38
rs1055776296 Chr.5: 177065698-177065699 on Build GRCh38
rs397883962 Chr.5: 177065881-177065882 on Build GRCh38
rs146386684 Chr.5: 177065858-177065859 on Build GRCh38
rs949928979 Chr.5: 177066035-177066035 on Build GRCh38
rs991395928 Chr.5: 177066012-177066012 on Build GRCh38
rs547447936 Chr.5: 177065781-177065781 on Build GRCh38
rs987367737 Chr.5: 177065873-177065882 on Build GRCh38
rs527751937 Chr.5: 177066086-177066086 on Build GRCh38
rs979814580 Chr.5: 177065889-177065889 on Build GRCh38
rs59157655 Chr.5: 177066105-177066105 on Build GRCh38
rs145366452 Chr.5: 177065782-177065782 on Build GRCh38
rs142648749 Chr.5: 177065980-177065980 on Build GRCh38
rs1021011694 Chr.5: 177065858-177065858 on Build GRCh38
rs535559099 Chr.5: 177065776-177065776 on Build GRCh38
rs35976285 Chr.5: 177065883-177065884 on Build GRCh38
rs11377166 Chr.5: 177065859-177065860 on Build GRCh38
rs954674467 Chr.5: 177065813-177065813 on Build GRCh38
rs939798439 Chr.5: 177066089-177066089 on Build GRCh38
rs895427653 Chr.5: 177065702-177065702 on Build GRCh38
rs796486285 Chr.5: 177065881-177065882 on Build GRCh38
rs190330034 Chr.5: 177065655-177065655 on Build GRCh38
rs776605861 Chr.5: 177065692-177065692 on Build GRCh38
rs1028476625 Chr.5: 177065805-177065805 on Build GRCh38
rs539795813 Chr.5: 177065789-177065789 on Build GRCh38
rs568262277 Chr.5: 177065733-177065733 on Build GRCh38
rs570707572 Chr.5: 177065908-177065908 on Build GRCh38
rs1013984020 Chr.5: 177065743-177065743 on Build GRCh38

View Full Product Details