Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 3104113-3104515 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RIPK1
Entrez Gene ID
8737
Gene Name
receptor interacting serine/threonine kinase 1
Gene Aliases
RIP, RIP-1, RIP1
UniGene
Hs.519842
Gene Chromosome Location
Chr.6: 3063888-3115187 on Build GRCh38
Transcripts
NM_001317061.1 NM_003804.4 XM_005249457.3 XM_017011403.1 XM_017011404.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7106819 c.929A>C p.N310T Chr.6: 3104238-3104238 on Build GRCh38
7237052 c.926C>G p.S309* Chr.6: 3104235-3104235 on Build GRCh38
1443019 c.955C>T p.Q319* Chr.6: 3104264-3104264 on Build GRCh38
3830061 c.919G>C p.E307Q Chr.6: 3104228-3104228 on Build GRCh38
1443018 c.931G>A p.E311K Chr.6: 3104240-3104240 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768817778 Chr.6: 3104203-3104203 on Build GRCh38
rs184895368 Chr.6: 3104240-3104240 on Build GRCh38
rs759456633 Chr.6: 3104282-3104282 on Build GRCh38
rs1046179236 Chr.6: 3104491-3104491 on Build GRCh38
rs558097599 Chr.6: 3104365-3104365 on Build GRCh38
rs556069772 Chr.6: 3104239-3104239 on Build GRCh38
rs557022552 Chr.6: 3104426-3104426 on Build GRCh38
rs1002183207 Chr.6: 3104391-3104391 on Build GRCh38
rs767249748 Chr.6: 3104284-3104284 on Build GRCh38
rs143730914 Chr.6: 3104251-3104251 on Build GRCh38
rs10498658 Chr.6: 3104462-3104462 on Build GRCh38
rs761849349 Chr.6: 3104219-3104219 on Build GRCh38
rs773038989 Chr.6: 3104177-3104178 on Build GRCh38
rs905932844 Chr.6: 3104495-3104495 on Build GRCh38
rs779281120 Chr.6: 3104177-3104177 on Build GRCh38
rs370418004 Chr.6: 3104303-3104303 on Build GRCh38
rs368212631 Chr.6: 3104364-3104364 on Build GRCh38
rs755727869 Chr.6: 3104175-3104175 on Build GRCh38
rs897108393 Chr.6: 3104414-3104414 on Build GRCh38
rs755668565 Chr.6: 3104304-3104304 on Build GRCh38
rs751323055 Chr.6: 3104265-3104265 on Build GRCh38
rs776886079 Chr.6: 3104214-3104214 on Build GRCh38
rs148924234 Chr.6: 3104152-3104152 on Build GRCh38
rs960810045 Chr.6: 3104444-3104444 on Build GRCh38
rs976198343 Chr.6: 3104212-3104212 on Build GRCh38
rs577664248 Chr.6: 3104370-3104370 on Build GRCh38
rs1040193944 Chr.6: 3104166-3104166 on Build GRCh38
rs34934849 Chr.6: 3104420-3104421 on Build GRCh38
rs941564489 Chr.6: 3104310-3104310 on Build GRCh38
rs71552162 Chr.6: 3104359-3104359 on Build GRCh38
rs777276268 Chr.6: 3104357-3104357 on Build GRCh38
rs199575720 Chr.6: 3104179-3104179 on Build GRCh38
rs772466959 Chr.6: 3104186-3104186 on Build GRCh38
rs747405361 Chr.6: 3104199-3104199 on Build GRCh38
rs762882089 Chr.6: 3104245-3104245 on Build GRCh38
rs543110451 Chr.6: 3104402-3104402 on Build GRCh38
rs192366630 Chr.6: 3104189-3104189 on Build GRCh38

View Full Product Details