Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 25984307-25984851 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRIM38
Entrez Gene ID
10475
Gene Name
tripartite motif containing 38
Gene Aliases
RNF15, RORET
UniGene
Hs.584851
Gene Chromosome Location
Chr.6: 25962689-25988867 on Build GRCh38
Transcripts
NM_006355.4 XM_005248799.4 XM_005248800.4

SNP Detail

SNP ID Chromosome Location
rs920293312 Chr.6: 25984390-25984390 on Build GRCh38
rs1011290661 Chr.6: 25984394-25984394 on Build GRCh38
rs891406148 Chr.6: 25984709-25984709 on Build GRCh38
rs1019970779 Chr.6: 25984489-25984489 on Build GRCh38
rs940841475 Chr.6: 25984753-25984753 on Build GRCh38
rs12214908 Chr.6: 25984366-25984366 on Build GRCh38
rs375108408 Chr.6: 25984408-25984409 on Build GRCh38
rs930513656 Chr.6: 25984412-25984412 on Build GRCh38
rs964808882 Chr.6: 25984739-25984739 on Build GRCh38
rs138807424 Chr.6: 25984538-25984538 on Build GRCh38
rs189977640 Chr.6: 25984617-25984617 on Build GRCh38
rs952134929 Chr.6: 25984693-25984693 on Build GRCh38
rs928909573 Chr.6: 25984789-25984789 on Build GRCh38
rs983371089 Chr.6: 25984422-25984422 on Build GRCh38
rs937473427 Chr.6: 25984550-25984550 on Build GRCh38
rs967175016 Chr.6: 25984470-25984470 on Build GRCh38
rs114233662 Chr.6: 25984495-25984495 on Build GRCh38
rs984808496 Chr.6: 25984735-25984735 on Build GRCh38
rs778390321 Chr.6: 25984442-25984442 on Build GRCh38
rs1027286097 Chr.6: 25984810-25984810 on Build GRCh38
rs150237996 Chr.6: 25984354-25984354 on Build GRCh38
rs762023081 Chr.6: 25984736-25984736 on Build GRCh38
rs1055453430 Chr.6: 25984380-25984380 on Build GRCh38
rs528066929 Chr.6: 25984410-25984410 on Build GRCh38
rs141052293 Chr.6: 25984351-25984351 on Build GRCh38
rs892926031 Chr.6: 25984388-25984388 on Build GRCh38
rs1008793962 Chr.6: 25984720-25984720 on Build GRCh38
rs1045600297 Chr.6: 25984504-25984504 on Build GRCh38
rs776775338 Chr.6: 25984651-25984651 on Build GRCh38
rs1018483443 Chr.6: 25984728-25984728 on Build GRCh38
rs974528265 Chr.6: 25984794-25984794 on Build GRCh38
rs981615618 Chr.6: 25984781-25984781 on Build GRCh38
rs374903640 Chr.6: 25984616-25984616 on Build GRCh38
rs781378902 Chr.6: 25984579-25984579 on Build GRCh38
rs908066610 Chr.6: 25984743-25984743 on Build GRCh38
rs1008163482 Chr.6: 25984487-25984487 on Build GRCh38
rs1054828472 Chr.6: 25984597-25984597 on Build GRCh38
rs907686968 Chr.6: 25984424-25984424 on Build GRCh38

View Full Product Details