Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 154155459-154155952 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TPM3
Entrez Gene ID
7170
Gene Name
tropomyosin 3
Gene Aliases
CAPM1, CFTD, HEL-189, HEL-S-82p, NEM1, OK/SW-cl.5, TM-5, TM3, TM30, TM30nm, TM5, TPMsk3, TRK, hscp30
UniGene
Hs.535581
Gene Chromosome Location
Chr.1: 154155304-154192135 on Build GRCh38
Transcripts
NM_001043351.1 NM_001043352.1 NM_001043353.1 NM_001278188.1 NM_001278189.1 NM_001278190.1 NM_001278191.1 NM_153649.3 NR_103461.1

SNP Detail

SNP ID Chromosome Location
rs3208688 Chr.1: 154155619-154155619 on Build GRCh38
rs531973259 Chr.1: 154155620-154155620 on Build GRCh38
rs1003262811 Chr.1: 154155703-154155703 on Build GRCh38
rs927753566 Chr.1: 154155494-154155494 on Build GRCh38
rs749198898 Chr.1: 154155520-154155520 on Build GRCh38
rs369912190 Chr.1: 154155923-154155923 on Build GRCh38
rs771228014 Chr.1: 154155647-154155647 on Build GRCh38
rs887326835 Chr.1: 154155614-154155614 on Build GRCh38
rs4078456 Chr.1: 154155652-154155652 on Build GRCh38
rs986013428 Chr.1: 154155720-154155720 on Build GRCh38
rs138182436 Chr.1: 154155635-154155637 on Build GRCh38
rs990166048 Chr.1: 154155901-154155901 on Build GRCh38
rs937727568 Chr.1: 154155498-154155498 on Build GRCh38
rs911690017 Chr.1: 154155536-154155536 on Build GRCh38
rs924625088 Chr.1: 154155556-154155556 on Build GRCh38
rs138993462 Chr.1: 154155890-154155890 on Build GRCh38
rs564661817 Chr.1: 154155495-154155495 on Build GRCh38
rs3208687 Chr.1: 154155621-154155621 on Build GRCh38
rs563701240 Chr.1: 154155481-154155481 on Build GRCh38
rs551802898 Chr.1: 154155691-154155691 on Build GRCh38
rs1028361511 Chr.1: 154155863-154155863 on Build GRCh38
rs777996896 Chr.1: 154155543-154155543 on Build GRCh38
rs78397190 Chr.1: 154155864-154155864 on Build GRCh38
rs181731316 Chr.1: 154155707-154155707 on Build GRCh38
rs142354101 Chr.1: 154155894-154155894 on Build GRCh38
rs894724351 Chr.1: 154155608-154155608 on Build GRCh38
rs145405416 Chr.1: 154155853-154155856 on Build GRCh38
rs879157180 Chr.1: 154155625-154155626 on Build GRCh38
rs549452838 Chr.1: 154155545-154155545 on Build GRCh38
rs116842438 Chr.1: 154155540-154155540 on Build GRCh38
rs185606717 Chr.1: 154155606-154155606 on Build GRCh38
rs190467762 Chr.1: 154155706-154155706 on Build GRCh38
rs1041587439 Chr.1: 154155694-154155694 on Build GRCh38
rs1018803218 Chr.1: 154155768-154155768 on Build GRCh38
rs977505822 Chr.1: 154155554-154155554 on Build GRCh38
rs1004341859 Chr.1: 154155637-154155637 on Build GRCh38
rs997328874 Chr.1: 154155738-154155738 on Build GRCh38
rs776747237 Chr.1: 154155705-154155705 on Build GRCh38
rs965845361 Chr.1: 154155800-154155800 on Build GRCh38
rs751500305 Chr.1: 154155757-154155758 on Build GRCh38
rs920238001 Chr.1: 154155527-154155527 on Build GRCh38
rs768772404 Chr.1: 154155526-154155526 on Build GRCh38
rs558186581 Chr.1: 154155929-154155929 on Build GRCh38
rs936000641 Chr.1: 154155591-154155591 on Build GRCh38

View Full Product Details