Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 47477951-47478476 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD2AP
Entrez Gene ID
23607
Gene Name
CD2 associated protein
Gene Aliases
CMS
UniGene
Hs.485518
Gene Chromosome Location
Chr.6: 47477746-47627263 on Build GRCh38
Transcripts
NM_012120.2 XM_005248976.1 XM_017010641.1

SNP Detail

SNP ID Chromosome Location
rs532229799 Chr.6: 47477978-47477978 on Build GRCh38
rs560930115 Chr.6: 47478190-47478191 on Build GRCh38
rs772554140 Chr.6: 47478194-47478195 on Build GRCh38
rs962535167 Chr.6: 47478098-47478098 on Build GRCh38
rs777922110 Chr.6: 47478277-47478277 on Build GRCh38
rs1056065100 Chr.6: 47478026-47478026 on Build GRCh38
rs749459476 Chr.6: 47478282-47478282 on Build GRCh38
rs953502624 Chr.6: 47478117-47478117 on Build GRCh38
rs370143870 Chr.6: 47478264-47478264 on Build GRCh38
rs550810602 Chr.6: 47478236-47478236 on Build GRCh38
rs1056434 Chr.6: 47478053-47478053 on Build GRCh38
rs886061517 Chr.6: 47478241-47478242 on Build GRCh38
rs1029554908 Chr.6: 47478158-47478158 on Build GRCh38
rs1003986342 Chr.6: 47478079-47478079 on Build GRCh38
rs544549262 Chr.6: 47478294-47478294 on Build GRCh38
rs1028245197 Chr.6: 47478115-47478115 on Build GRCh38
rs529849638 Chr.6: 47478033-47478033 on Build GRCh38
rs751438191 Chr.6: 47478259-47478259 on Build GRCh38
rs558578035 Chr.6: 47478386-47478386 on Build GRCh38
rs988355427 Chr.6: 47478440-47478440 on Build GRCh38
rs544458907 Chr.6: 47477981-47477981 on Build GRCh38
rs12192946 Chr.6: 47478371-47478371 on Build GRCh38
rs544327170 Chr.6: 47478445-47478445 on Build GRCh38
rs762264070 Chr.6: 47478417-47478417 on Build GRCh38
rs768055057 Chr.6: 47478229-47478229 on Build GRCh38
rs916204452 Chr.6: 47478245-47478245 on Build GRCh38
rs893913619 Chr.6: 47478056-47478083 on Build GRCh38
rs1013923409 Chr.6: 47478042-47478042 on Build GRCh38
rs960880295 Chr.6: 47478100-47478100 on Build GRCh38
rs886839453 Chr.6: 47478360-47478360 on Build GRCh38
rs945520303 Chr.6: 47478169-47478180 on Build GRCh38
rs768196661 Chr.6: 47478424-47478424 on Build GRCh38
rs900163108 Chr.6: 47478236-47478237 on Build GRCh38
rs936228355 Chr.6: 47477986-47477986 on Build GRCh38
rs573455091 Chr.6: 47478318-47478318 on Build GRCh38
rs758123465 Chr.6: 47478237-47478237 on Build GRCh38
rs956758822 Chr.6: 47478395-47478395 on Build GRCh38
rs771046571 Chr.6: 47478103-47478103 on Build GRCh38
rs946137629 Chr.6: 47478176-47478176 on Build GRCh38
rs566721596 Chr.6: 47478078-47478078 on Build GRCh38
rs1023670980 Chr.6: 47478321-47478321 on Build GRCh38
rs969368611 Chr.6: 47478326-47478326 on Build GRCh38
rs538739578 Chr.6: 47478226-47478226 on Build GRCh38
rs781225870 Chr.6: 47478269-47478269 on Build GRCh38
rs761183938 Chr.6: 47478235-47478235 on Build GRCh38
rs886061513 Chr.6: 47478048-47478048 on Build GRCh38
rs1018996847 Chr.6: 47478020-47478020 on Build GRCh38
rs199562171 Chr.6: 47478215-47478215 on Build GRCh38
rs745454724 Chr.6: 47478007-47478007 on Build GRCh38
rs1056410566 Chr.6: 47478241-47478241 on Build GRCh38
rs751993865 Chr.6: 47478273-47478273 on Build GRCh38
rs1045399531 Chr.6: 47478281-47478281 on Build GRCh38
rs544576186 Chr.6: 47478160-47478167 on Build GRCh38
rs911968349 Chr.6: 47478173-47478173 on Build GRCh38
rs1052529182 Chr.6: 47478184-47478184 on Build GRCh38
rs533900030 Chr.6: 47478076-47478078 on Build GRCh38
rs567589455 Chr.6: 47478255-47478255 on Build GRCh38
rs925336925 Chr.6: 47478160-47478160 on Build GRCh38
rs886061514 Chr.6: 47478054-47478054 on Build GRCh38
rs944273817 Chr.6: 47478221-47478221 on Build GRCh38
rs554979605 Chr.6: 47478308-47478308 on Build GRCh38
rs979361943 Chr.6: 47478326-47478326 on Build GRCh38
rs748147507 Chr.6: 47478272-47478272 on Build GRCh38
rs1038097174 Chr.6: 47478443-47478443 on Build GRCh38
rs975706201 Chr.6: 47478109-47478109 on Build GRCh38
rs562538073 Chr.6: 47477991-47477991 on Build GRCh38
rs1008580714 Chr.6: 47478006-47478006 on Build GRCh38
rs1040062364 Chr.6: 47478222-47478222 on Build GRCh38
rs749530775 Chr.6: 47478203-47478203 on Build GRCh38
rs1024005555 Chr.6: 47478048-47478048 on Build GRCh38
rs760034318 Chr.6: 47478228-47478228 on Build GRCh38
rs886061516 Chr.6: 47478115-47478115 on Build GRCh38
rs948203089 Chr.6: 47478143-47478143 on Build GRCh38
rs894098539 Chr.6: 47478031-47478031 on Build GRCh38
rs987526270 Chr.6: 47478155-47478155 on Build GRCh38
rs9349406 Chr.6: 47478191-47478191 on Build GRCh38
rs1013918946 Chr.6: 47478303-47478303 on Build GRCh38
rs766268810 Chr.6: 47478238-47478238 on Build GRCh38
rs960682411 Chr.6: 47478298-47478298 on Build GRCh38
rs773265672 Chr.6: 47478196-47478196 on Build GRCh38
rs886061515 Chr.6: 47478078-47478080 on Build GRCh38
rs749334649 Chr.6: 47478091-47478091 on Build GRCh38
rs528486264 Chr.6: 47478081-47478081 on Build GRCh38
rs577058736 Chr.6: 47478391-47478391 on Build GRCh38
rs533804071 Chr.6: 47478363-47478363 on Build GRCh38

View Full Product Details