Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 83216360-83216907 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ME1
Entrez Gene ID
4199
Gene Name
malic enzyme 1
Gene Aliases
HUMNDME, MES
UniGene
Hs.21160
Gene Chromosome Location
Chr.6: 83210389-83431219 on Build GRCh38
Transcripts
NM_002395.5 XM_011535836.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7158757 c.1531C>G p.L511V Chr.6: 83216515-83216515 on Build GRCh38
1697583 c.1516A>T p.I506F Chr.6: 83216530-83216530 on Build GRCh38
75386 c.1462C>A p.Q488K Chr.6: 83216584-83216584 on Build GRCh38
4467718 c.1504C>T p.P502S Chr.6: 83216542-83216542 on Build GRCh38
4972906 c.1455A>G p.I485M Chr.6: 83216591-83216591 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754350171 Chr.6: 83216567-83216567 on Build GRCh38
rs779055872 Chr.6: 83216448-83216448 on Build GRCh38
rs755913240 Chr.6: 83216618-83216618 on Build GRCh38
rs761842278 Chr.6: 83216580-83216580 on Build GRCh38
rs908628383 Chr.6: 83216777-83216777 on Build GRCh38
rs374879325 Chr.6: 83216646-83216646 on Build GRCh38
rs561869394 Chr.6: 83216746-83216746 on Build GRCh38
rs775998450 Chr.6: 83216546-83216546 on Build GRCh38
rs374567163 Chr.6: 83216601-83216601 on Build GRCh38
rs982747166 Chr.6: 83216773-83216773 on Build GRCh38
rs753255330 Chr.6: 83216675-83216675 on Build GRCh38
rs370159373 Chr.6: 83216638-83216638 on Build GRCh38
rs745306723 Chr.6: 83216632-83216632 on Build GRCh38
rs370774453 Chr.6: 83216586-83216586 on Build GRCh38
rs113693452 Chr.6: 83216416-83216416 on Build GRCh38
rs370421410 Chr.6: 83216472-83216472 on Build GRCh38
rs774213170 Chr.6: 83216514-83216514 on Build GRCh38
rs145569223 Chr.6: 83216565-83216565 on Build GRCh38
rs778574695 Chr.6: 83216631-83216631 on Build GRCh38
rs541170033 Chr.6: 83216863-83216863 on Build GRCh38
rs770405129 Chr.6: 83216629-83216629 on Build GRCh38
rs1031084846 Chr.6: 83216445-83216445 on Build GRCh38
rs748796473 Chr.6: 83216497-83216497 on Build GRCh38
rs777402925 Chr.6: 83216707-83216707 on Build GRCh38
rs367861178 Chr.6: 83216832-83216832 on Build GRCh38
rs1033226542 Chr.6: 83216425-83216425 on Build GRCh38
rs895459348 Chr.6: 83216665-83216665 on Build GRCh38
rs796403771 Chr.6: 83216506-83216506 on Build GRCh38
rs541632705 Chr.6: 83216643-83216643 on Build GRCh38
rs972213524 Chr.6: 83216623-83216623 on Build GRCh38
rs759250240 Chr.6: 83216523-83216523 on Build GRCh38
rs770442498 Chr.6: 83216509-83216509 on Build GRCh38
rs766542346 Chr.6: 83216403-83216403 on Build GRCh38
rs377743345 Chr.6: 83216604-83216604 on Build GRCh38
rs367907680 Chr.6: 83216581-83216581 on Build GRCh38
rs752521310 Chr.6: 83216616-83216616 on Build GRCh38
rs1033631946 Chr.6: 83216703-83216703 on Build GRCh38
rs750706574 Chr.6: 83216591-83216591 on Build GRCh38
rs920445582 Chr.6: 83216870-83216870 on Build GRCh38
rs879135160 Chr.6: 83216584-83216584 on Build GRCh38
rs375254909 Chr.6: 83216541-83216541 on Build GRCh38
rs1029271116 Chr.6: 83216494-83216494 on Build GRCh38
rs754429126 Chr.6: 83216552-83216556 on Build GRCh38
rs1043545530 Chr.6: 83216690-83216690 on Build GRCh38
rs757919289 Chr.6: 83216693-83216693 on Build GRCh38
rs922635024 Chr.6: 83216486-83216486 on Build GRCh38
rs746062564 Chr.6: 83216449-83216449 on Build GRCh38
rs761057102 Chr.6: 83216554-83216554 on Build GRCh38
rs749887453 Chr.6: 83216657-83216657 on Build GRCh38
rs955339762 Chr.6: 83216763-83216763 on Build GRCh38
rs773163466 Chr.6: 83216485-83216485 on Build GRCh38
rs754838498 Chr.6: 83216451-83216451 on Build GRCh38
rs780951379 Chr.6: 83216457-83216457 on Build GRCh38
rs777609817 Chr.6: 83216621-83216621 on Build GRCh38
rs1020012510 Chr.6: 83216681-83216681 on Build GRCh38
rs113524520 Chr.6: 83216639-83216639 on Build GRCh38
rs112182338 Chr.6: 83216761-83216761 on Build GRCh38
rs901488100 Chr.6: 83216750-83216750 on Build GRCh38
rs903624099 Chr.6: 83216406-83216406 on Build GRCh38
rs368551004 Chr.6: 83216636-83216636 on Build GRCh38
rs759833549 Chr.6: 83216556-83216556 on Build GRCh38
rs564070506 Chr.6: 83216875-83216875 on Build GRCh38
rs780631259 Chr.6: 83216456-83216459 on Build GRCh38
rs112280660 Chr.6: 83216384-83216384 on Build GRCh38
rs997532947 Chr.6: 83216743-83216743 on Build GRCh38
rs780185399 Chr.6: 83216610-83216610 on Build GRCh38
rs763129607 Chr.6: 83216391-83216391 on Build GRCh38
rs947859326 Chr.6: 83216676-83216676 on Build GRCh38
rs749243530 Chr.6: 83216622-83216622 on Build GRCh38
rs751903740 Chr.6: 83216458-83216458 on Build GRCh38
rs963153594 Chr.6: 83216854-83216854 on Build GRCh38

View Full Product Details