Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 117707040-117707549 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NUS1
Entrez Gene ID
116150
Gene Name
NUS1 dehydrodolichyl diphosphate synthase subunit
Gene Aliases
C6orf68, MGC:7199, NgBR, TANGO14
UniGene
Hs.289008
Gene Chromosome Location
Chr.6: 117675454-117710727 on Build GRCh38
Transcripts
NM_138459.3

SNP Detail

SNP ID Chromosome Location
rs1052263 Chr.6: 117707247-117707247 on Build GRCh38
rs752181754 Chr.6: 117707189-117707189 on Build GRCh38
rs4028841 Chr.6: 117707218-117707218 on Build GRCh38
rs984587464 Chr.6: 117707201-117707201 on Build GRCh38
rs568808909 Chr.6: 117707105-117707105 on Build GRCh38
rs1026305251 Chr.6: 117707479-117707479 on Build GRCh38
rs1032412818 Chr.6: 117707380-117707380 on Build GRCh38
rs973047023 Chr.6: 117707460-117707460 on Build GRCh38
rs1052262 Chr.6: 117707219-117707219 on Build GRCh38
rs771922598 Chr.6: 117707064-117707064 on Build GRCh38
rs567506617 Chr.6: 117707213-117707213 on Build GRCh38
rs189173604 Chr.6: 117707141-117707141 on Build GRCh38
rs4028840 Chr.6: 117707113-117707113 on Build GRCh38
rs199562088 Chr.6: 117707171-117707172 on Build GRCh38
rs375623081 Chr.6: 117707182-117707183 on Build GRCh38
rs553483026 Chr.6: 117707506-117707506 on Build GRCh38
rs933255217 Chr.6: 117707311-117707311 on Build GRCh38
rs902545701 Chr.6: 117707355-117707355 on Build GRCh38
rs1001259886 Chr.6: 117707358-117707358 on Build GRCh38
rs908946351 Chr.6: 117707217-117707217 on Build GRCh38
rs973360172 Chr.6: 117707170-117707170 on Build GRCh38
rs1012255731 Chr.6: 117707408-117707408 on Build GRCh38
rs571153679 Chr.6: 117707419-117707419 on Build GRCh38
rs990929758 Chr.6: 117707101-117707101 on Build GRCh38
rs28414025 Chr.6: 117707138-117707138 on Build GRCh38
rs950439391 Chr.6: 117707485-117707485 on Build GRCh38
rs918931062 Chr.6: 117707184-117707184 on Build GRCh38
rs1042695384 Chr.6: 117707349-117707349 on Build GRCh38
rs1038898945 Chr.6: 117707251-117707251 on Build GRCh38
rs549936556 Chr.6: 117707327-117707327 on Build GRCh38
rs4028839 Chr.6: 117707062-117707062 on Build GRCh38
rs538322764 Chr.6: 117707502-117707502 on Build GRCh38
rs928958510 Chr.6: 117707193-117707193 on Build GRCh38
rs959559976 Chr.6: 117707078-117707078 on Build GRCh38
rs764928035 Chr.6: 117707165-117707168 on Build GRCh38
rs971014810 Chr.6: 117707170-117707171 on Build GRCh38
rs1052261 Chr.6: 117707178-117707178 on Build GRCh38
rs75648010 Chr.6: 117707093-117707093 on Build GRCh38
rs1052258 Chr.6: 117707083-117707083 on Build GRCh38
rs1025104183 Chr.6: 117707410-117707410 on Build GRCh38
rs895252474 Chr.6: 117707407-117707407 on Build GRCh38
rs181113936 Chr.6: 117707244-117707244 on Build GRCh38
rs901644079 Chr.6: 117707307-117707307 on Build GRCh38

View Full Product Details