Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 127266600-127267105 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNF146
Entrez Gene ID
81847
Gene Name
ring finger protein 146
Gene Aliases
-
UniGene
Hs.267120
Gene Chromosome Location
Chr.6: 127266340-127289893 on Build GRCh38
Transcripts
NM_001242844.1 NM_001242845.1 NM_001242846.1 NM_001242847.1 NM_001242848.1 NM_001242849.1 NM_001242850.1 NM_001242851.1 NM_001242852.1 NM_030963.3 XM_006715571.3 XM_011536161.2 XM_011536162.2 XM_011536163.2 XM_011536164.2 XM_017011336.1 XM_017011337.1 XM_017011338.1 XM_017011339.1 XM_017011340.1 XM_017011341.1 XM_017011342.1 XM_017011343.1

SNP Detail

SNP ID Chromosome Location
rs906119718 Chr.6: 127266969-127266969 on Build GRCh38
rs145142014 Chr.6: 127266823-127266823 on Build GRCh38
rs748667467 Chr.6: 127266765-127266765 on Build GRCh38
rs1000419295 Chr.6: 127266971-127266971 on Build GRCh38
rs1016772987 Chr.6: 127266980-127266980 on Build GRCh38
rs922273210 Chr.6: 127266860-127266860 on Build GRCh38
rs1007547584 Chr.6: 127266799-127266799 on Build GRCh38
rs913620759 Chr.6: 127266886-127266886 on Build GRCh38
rs569397323 Chr.6: 127266782-127266782 on Build GRCh38
rs955071914 Chr.6: 127267031-127267031 on Build GRCh38
rs984288994 Chr.6: 127266749-127266749 on Build GRCh38
rs186702072 Chr.6: 127266759-127266759 on Build GRCh38
rs906544659 Chr.6: 127266923-127266923 on Build GRCh38
rs943559691 Chr.6: 127266892-127266892 on Build GRCh38
rs933580326 Chr.6: 127266766-127266766 on Build GRCh38
rs1023252709 Chr.6: 127266847-127266848 on Build GRCh38
rs997676343 Chr.6: 127266709-127266709 on Build GRCh38
rs1030448385 Chr.6: 127266710-127266710 on Build GRCh38
rs778539602 Chr.6: 127266902-127266902 on Build GRCh38
rs985128299 Chr.6: 127267041-127267041 on Build GRCh38
rs933249830 Chr.6: 127266779-127266784 on Build GRCh38
rs1009664342 Chr.6: 127266907-127266907 on Build GRCh38
rs571274989 Chr.6: 127266707-127266707 on Build GRCh38
rs1026443776 Chr.6: 127266733-127266733 on Build GRCh38
rs1003479742 Chr.6: 127266786-127266786 on Build GRCh38
rs987883196 Chr.6: 127266885-127266885 on Build GRCh38
rs1003530771 Chr.6: 127266936-127266936 on Build GRCh38
rs993067568 Chr.6: 127266743-127266743 on Build GRCh38
rs889018055 Chr.6: 127266789-127266789 on Build GRCh38
rs937835026 Chr.6: 127266748-127266748 on Build GRCh38
rs567185839 Chr.6: 127266829-127266829 on Build GRCh38
rs544416416 Chr.6: 127266967-127266967 on Build GRCh38
rs1036651141 Chr.6: 127266762-127266762 on Build GRCh38
rs1016330345 Chr.6: 127266705-127266705 on Build GRCh38
rs988128937 Chr.6: 127266728-127266728 on Build GRCh38
rs939674222 Chr.6: 127266671-127266671 on Build GRCh38
rs889254498 Chr.6: 127266711-127266711 on Build GRCh38
rs961154489 Chr.6: 127266995-127266995 on Build GRCh38
rs933635997 Chr.6: 127266872-127266872 on Build GRCh38
rs1037663018 Chr.6: 127266813-127266813 on Build GRCh38
rs1051981293 Chr.6: 127266878-127266878 on Build GRCh38
rs189838722 Chr.6: 127266784-127266784 on Build GRCh38
rs1005830751 Chr.6: 127266950-127266950 on Build GRCh38
rs1020736014 Chr.6: 127267041-127267042 on Build GRCh38
rs980421504 Chr.6: 127266754-127266754 on Build GRCh38
rs889090155 Chr.6: 127266639-127266639 on Build GRCh38
rs538365821 Chr.6: 127266739-127266739 on Build GRCh38
rs1012747299 Chr.6: 127267021-127267021 on Build GRCh38
rs909661380 Chr.6: 127266837-127266837 on Build GRCh38
rs961102407 Chr.6: 127266838-127266838 on Build GRCh38
rs558681084 Chr.6: 127266883-127266883 on Build GRCh38
rs972421644 Chr.6: 127266847-127266847 on Build GRCh38
rs557100644 Chr.6: 127267003-127267003 on Build GRCh38
rs1032405574 Chr.6: 127266716-127266716 on Build GRCh38
rs1014459988 Chr.6: 127266962-127266962 on Build GRCh38
rs889302852 Chr.6: 127266768-127266768 on Build GRCh38
rs534154150 Chr.6: 127266879-127266879 on Build GRCh38
rs1033572331 Chr.6: 127266816-127266816 on Build GRCh38
rs1034037344 Chr.6: 127266942-127266942 on Build GRCh38
rs980475136 Chr.6: 127266850-127266850 on Build GRCh38
rs1014820168 Chr.6: 127266841-127266841 on Build GRCh38
rs877661 Chr.6: 127266770-127266770 on Build GRCh38
rs1032027011 Chr.6: 127266992-127266992 on Build GRCh38
rs910836814 Chr.6: 127267048-127267048 on Build GRCh38
rs972473504 Chr.6: 127267000-127267000 on Build GRCh38
rs748993549 Chr.6: 127266779-127266779 on Build GRCh38
rs747676907 Chr.6: 127266799-127266800 on Build GRCh38
rs958295845 Chr.6: 127266723-127266723 on Build GRCh38
rs1036601762 Chr.6: 127266675-127266675 on Build GRCh38
rs981976755 Chr.6: 127266742-127266742 on Build GRCh38
rs1020907144 Chr.6: 127266736-127266736 on Build GRCh38
rs959315417 Chr.6: 127266943-127266943 on Build GRCh38
rs1054310501 Chr.6: 127266775-127266775 on Build GRCh38
rs558041310 Chr.6: 127266744-127266744 on Build GRCh38
rs1009610858 Chr.6: 127266780-127266780 on Build GRCh38
rs1023138623 Chr.6: 127266968-127266968 on Build GRCh38
rs1026912287 Chr.6: 127266831-127266831 on Build GRCh38
rs899142806 Chr.6: 127266934-127266934 on Build GRCh38
rs910508724 Chr.6: 127266906-127266906 on Build GRCh38
rs959495822 Chr.6: 127266826-127266826 on Build GRCh38
rs781673535 Chr.6: 127266721-127266721 on Build GRCh38
rs1008061230 Chr.6: 127266715-127266715 on Build GRCh38
rs909619668 Chr.6: 127266756-127266756 on Build GRCh38
rs755309930 Chr.6: 127266689-127266689 on Build GRCh38
rs994111282 Chr.6: 127266729-127266729 on Build GRCh38
rs987268044 Chr.6: 127267042-127267043 on Build GRCh38
rs910351782 Chr.6: 127267045-127267045 on Build GRCh38
rs1051923569 Chr.6: 127266767-127266767 on Build GRCh38
rs577224911 Chr.6: 127266912-127266912 on Build GRCh38
rs1053956578 Chr.6: 127266896-127266896 on Build GRCh38

View Full Product Details