Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 136260312-136260819 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCLAF1
Entrez Gene ID
9774
Gene Name
BCL2 associated transcription factor 1
Gene Aliases
BTF, bK211L9.1
UniGene
Hs.486542
Gene Chromosome Location
Chr.6: 136256863-136290645 on Build GRCh38
Transcripts
NM_001077440.1 NM_001077441.1 NM_001301038.1 NM_014739.2 XM_005267237.3 XM_017011529.1 XM_017011530.1 XM_017011531.1 XM_017011532.1

SNP Detail

SNP ID Chromosome Location
rs1022172931 Chr.6: 136260754-136260754 on Build GRCh38
rs796112968 Chr.6: 136260644-136260644 on Build GRCh38
rs1035828810 Chr.6: 136260385-136260385 on Build GRCh38
rs906123281 Chr.6: 136260523-136260523 on Build GRCh38
rs547708959 Chr.6: 136260740-136260740 on Build GRCh38
rs1010423161 Chr.6: 136260750-136260750 on Build GRCh38
rs751916669 Chr.6: 136260395-136260396 on Build GRCh38
rs539167611 Chr.6: 136260349-136260349 on Build GRCh38
rs879748966 Chr.6: 136260537-136260537 on Build GRCh38
rs918466706 Chr.6: 136260451-136260451 on Build GRCh38
rs530859476 Chr.6: 136260370-136260370 on Build GRCh38
rs985399584 Chr.6: 136260425-136260425 on Build GRCh38
rs1003046030 Chr.6: 136260362-136260362 on Build GRCh38
rs984336209 Chr.6: 136260646-136260646 on Build GRCh38
rs1040932723 Chr.6: 136260444-136260444 on Build GRCh38
rs1012311029 Chr.6: 136260339-136260339 on Build GRCh38
rs538662536 Chr.6: 136260764-136260764 on Build GRCh38
rs554723798 Chr.6: 136260432-136260432 on Build GRCh38
rs745400431 Chr.6: 136260567-136260567 on Build GRCh38
rs985432572 Chr.6: 136260560-136260560 on Build GRCh38
rs914622734 Chr.6: 136260410-136260410 on Build GRCh38
rs970273121 Chr.6: 136260359-136260359 on Build GRCh38
rs148972267 Chr.6: 136260404-136260404 on Build GRCh38
rs1044714780 Chr.6: 136260513-136260513 on Build GRCh38
rs768098431 Chr.6: 136260704-136260707 on Build GRCh38
rs111296119 Chr.6: 136260744-136260744 on Build GRCh38
rs527542711 Chr.6: 136260676-136260676 on Build GRCh38
rs369456697 Chr.6: 136260501-136260502 on Build GRCh38
rs755957082 Chr.6: 136260411-136260411 on Build GRCh38
rs909634335 Chr.6: 136260672-136260672 on Build GRCh38
rs540515983 Chr.6: 136260504-136260504 on Build GRCh38
rs116261262 Chr.6: 136260615-136260615 on Build GRCh38
rs1044831666 Chr.6: 136260678-136260678 on Build GRCh38
rs538080705 Chr.6: 136260407-136260407 on Build GRCh38
rs929778464 Chr.6: 136260499-136260499 on Build GRCh38
rs558784509 Chr.6: 136260401-136260401 on Build GRCh38
rs918483709 Chr.6: 136260612-136260612 on Build GRCh38
rs766701834 Chr.6: 136260504-136260505 on Build GRCh38
rs776920842 Chr.6: 136260494-136260494 on Build GRCh38
rs777163723 Chr.6: 136260506-136260507 on Build GRCh38
rs542971362 Chr.6: 136260536-136260537 on Build GRCh38
rs906239541 Chr.6: 136260706-136260706 on Build GRCh38
rs976660722 Chr.6: 136260597-136260597 on Build GRCh38
rs762246770 Chr.6: 136260527-136260527 on Build GRCh38
rs186611139 Chr.6: 136260590-136260590 on Build GRCh38
rs1018263938 Chr.6: 136260585-136260585 on Build GRCh38
rs764445743 Chr.6: 136260464-136260464 on Build GRCh38
rs761093287 Chr.6: 136260435-136260435 on Build GRCh38
rs367788996 Chr.6: 136260660-136260660 on Build GRCh38
rs559764045 Chr.6: 136260505-136260505 on Build GRCh38
rs574831293 Chr.6: 136260450-136260450 on Build GRCh38
rs911157202 Chr.6: 136260427-136260427 on Build GRCh38
rs576767256 Chr.6: 136260506-136260506 on Build GRCh38
rs1006967937 Chr.6: 136260791-136260791 on Build GRCh38
rs561849512 Chr.6: 136260645-136260645 on Build GRCh38
rs566793633 Chr.6: 136260545-136260545 on Build GRCh38
rs969185896 Chr.6: 136260546-136260546 on Build GRCh38
rs75145909 Chr.6: 136260529-136260529 on Build GRCh38
rs1057400040 Chr.6: 136260743-136260743 on Build GRCh38
rs369430729 Chr.6: 136260343-136260343 on Build GRCh38
rs377678323 Chr.6: 136260734-136260734 on Build GRCh38
rs965346365 Chr.6: 136260595-136260595 on Build GRCh38

View Full Product Details