Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 20783294-20783787 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SP8
Entrez Gene ID
221833
Gene Name
Sp8 transcription factor
Gene Aliases
BTD
UniGene
Hs.195922
Gene Chromosome Location
Chr.7: 20782274-20786889 on Build GRCh38
Transcripts
NM_182700.5 NM_198956.3

SNP Detail

SNP ID Chromosome Location
rs547758415 Chr.7: 20783495-20783496 on Build GRCh38
rs545627561 Chr.7: 20783326-20783326 on Build GRCh38
rs68103805 Chr.7: 20783619-20783620 on Build GRCh38
rs949770660 Chr.7: 20783501-20783501 on Build GRCh38
rs910827837 Chr.7: 20783389-20783389 on Build GRCh38
rs1021012844 Chr.7: 20783720-20783720 on Build GRCh38
rs546737380 Chr.7: 20783502-20783503 on Build GRCh38
rs1018160457 Chr.7: 20783556-20783556 on Build GRCh38
rs59712697 Chr.7: 20783658-20783659 on Build GRCh38
rs937933781 Chr.7: 20783711-20783711 on Build GRCh38
rs1038994716 Chr.7: 20783597-20783597 on Build GRCh38
rs60840167 Chr.7: 20783425-20783426 on Build GRCh38
rs979061894 Chr.7: 20783487-20783487 on Build GRCh38
rs1020737517 Chr.7: 20783509-20783509 on Build GRCh38
rs190337145 Chr.7: 20783546-20783546 on Build GRCh38
rs964935170 Chr.7: 20783390-20783390 on Build GRCh38
rs976093739 Chr.7: 20783403-20783403 on Build GRCh38
rs891321156 Chr.7: 20783698-20783698 on Build GRCh38
rs976292082 Chr.7: 20783598-20783598 on Build GRCh38
rs941040872 Chr.7: 20783588-20783588 on Build GRCh38
rs537982679 Chr.7: 20783738-20783738 on Build GRCh38
rs761570479 Chr.7: 20783335-20783335 on Build GRCh38
rs1056347044 Chr.7: 20783725-20783725 on Build GRCh38
rs1038221675 Chr.7: 20783647-20783647 on Build GRCh38
rs994343941 Chr.7: 20783674-20783674 on Build GRCh38
rs1021543549 Chr.7: 20783337-20783337 on Build GRCh38
rs1047845349 Chr.7: 20783439-20783439 on Build GRCh38
rs964966401 Chr.7: 20783571-20783571 on Build GRCh38
rs59293237 Chr.7: 20783672-20783673 on Build GRCh38
rs896399806 Chr.7: 20783745-20783745 on Build GRCh38
rs202080088 Chr.7: 20783617-20783617 on Build GRCh38
rs1009576902 Chr.7: 20783703-20783703 on Build GRCh38
rs955451640 Chr.7: 20783382-20783382 on Build GRCh38
rs143163563 Chr.7: 20783639-20783639 on Build GRCh38
rs62453408 Chr.7: 20783357-20783357 on Build GRCh38
rs869292934 Chr.7: 20783673-20783674 on Build GRCh38
rs149116216 Chr.7: 20783488-20783488 on Build GRCh38
rs1001058278 Chr.7: 20783534-20783534 on Build GRCh38
rs187165584 Chr.7: 20783374-20783374 on Build GRCh38
rs942087881 Chr.7: 20783645-20783645 on Build GRCh38
rs1002049983 Chr.7: 20783477-20783477 on Build GRCh38
rs772035932 Chr.7: 20783422-20783442 on Build GRCh38
rs909320504 Chr.7: 20783643-20783643 on Build GRCh38
rs764862916 Chr.7: 20783362-20783362 on Build GRCh38
rs66552968 Chr.7: 20783426-20783427 on Build GRCh38
rs550775122 Chr.7: 20783448-20783448 on Build GRCh38
rs59073147 Chr.7: 20783618-20783619 on Build GRCh38
rs202003917 Chr.7: 20783426-20783426 on Build GRCh38
rs937136296 Chr.7: 20783493-20783493 on Build GRCh38
rs950727318 Chr.7: 20783628-20783628 on Build GRCh38
rs750009966 Chr.7: 20783395-20783395 on Build GRCh38
rs926426546 Chr.7: 20783680-20783680 on Build GRCh38
rs1042421883 Chr.7: 20783753-20783753 on Build GRCh38
rs941230648 Chr.7: 20783460-20783460 on Build GRCh38
rs35044068 Chr.7: 20783595-20783596 on Build GRCh38
rs905128865 Chr.7: 20783467-20783467 on Build GRCh38
rs1029765393 Chr.7: 20783695-20783695 on Build GRCh38
rs923512242 Chr.7: 20783613-20783613 on Build GRCh38
rs945073659 Chr.7: 20783748-20783748 on Build GRCh38
rs201200999 Chr.7: 20783619-20783619 on Build GRCh38
rs551332245 Chr.7: 20783502-20783502 on Build GRCh38
rs200447819 Chr.7: 20783427-20783427 on Build GRCh38
rs1038593928 Chr.7: 20783462-20783462 on Build GRCh38
rs968414136 Chr.7: 20783752-20783752 on Build GRCh38
rs533169664 Chr.7: 20783495-20783495 on Build GRCh38
rs375921320 Chr.7: 20783424-20783424 on Build GRCh38
rs370851373 Chr.7: 20783425-20783425 on Build GRCh38
rs923233956 Chr.7: 20783418-20783418 on Build GRCh38
rs73685720 Chr.7: 20783524-20783524 on Build GRCh38
rs555574956 Chr.7: 20783624-20783624 on Build GRCh38

View Full Product Details