Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 156530018-156530529 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IQGAP3
Entrez Gene ID
128239
Gene Name
IQ motif containing GTPase activating protein 3
Gene Aliases
-
UniGene
Hs.591495
Gene Chromosome Location
Chr.1: 156525405-156572604 on Build GRCh38
Transcripts
NM_178229.4 XM_011509198.2 XM_011509200.2 XM_011509201.2 XM_017000317.1 XM_017000318.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7189443 c.4391A>G p.D1464G Chr.1: 156530118-156530118 on Build GRCh38
424126 c.4271C>T p.S1424L Chr.1: 156530238-156530238 on Build GRCh38
424125 c.4332A>G p.L1444L Chr.1: 156530177-156530177 on Build GRCh38
424124 c.4398G>A p.L1466L Chr.1: 156530111-156530111 on Build GRCh38
3965880 c.4320C>G p.V1440V Chr.1: 156530189-156530189 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs539444768 Chr.1: 156530375-156530375 on Build GRCh38
rs201439773 Chr.1: 156530193-156530193 on Build GRCh38
rs1044754353 Chr.1: 156530280-156530280 on Build GRCh38
rs924100969 Chr.1: 156530383-156530383 on Build GRCh38
rs776760949 Chr.1: 156530290-156530290 on Build GRCh38
rs756088943 Chr.1: 156530345-156530345 on Build GRCh38
rs762101212 Chr.1: 156530238-156530238 on Build GRCh38
rs766006590 Chr.1: 156530331-156530331 on Build GRCh38
rs750097388 Chr.1: 156530191-156530191 on Build GRCh38
rs732361 Chr.1: 156530366-156530366 on Build GRCh38
rs201070239 Chr.1: 156530241-156530241 on Build GRCh38
rs766756159 Chr.1: 156530082-156530082 on Build GRCh38
rs751036559 Chr.1: 156530248-156530248 on Build GRCh38
rs751653682 Chr.1: 156530083-156530083 on Build GRCh38
rs756180940 Chr.1: 156530262-156530262 on Build GRCh38
rs777137475 Chr.1: 156530237-156530237 on Build GRCh38
rs1043877178 Chr.1: 156530065-156530065 on Build GRCh38
rs368700057 Chr.1: 156530214-156530214 on Build GRCh38
rs772234683 Chr.1: 156530226-156530226 on Build GRCh38
rs376708543 Chr.1: 156530342-156530342 on Build GRCh38
rs1171566 Chr.1: 156530177-156530177 on Build GRCh38
rs368191209 Chr.1: 156530146-156530146 on Build GRCh38
rs768499831 Chr.1: 156530073-156530073 on Build GRCh38
rs763620083 Chr.1: 156530097-156530099 on Build GRCh38
rs374144760 Chr.1: 156530197-156530197 on Build GRCh38
rs376519095 Chr.1: 156530312-156530312 on Build GRCh38
rs764813770 Chr.1: 156530190-156530190 on Build GRCh38
rs781305314 Chr.1: 156530196-156530196 on Build GRCh38
rs774732936 Chr.1: 156530172-156530172 on Build GRCh38
rs72996606 Chr.1: 156530060-156530060 on Build GRCh38
rs768730139 Chr.1: 156530289-156530289 on Build GRCh38
rs373374039 Chr.1: 156530269-156530269 on Build GRCh38
rs775613036 Chr.1: 156530227-156530227 on Build GRCh38
rs756872734 Chr.1: 156530124-156530124 on Build GRCh38
rs568433536 Chr.1: 156530192-156530192 on Build GRCh38
rs759473769 Chr.1: 156530173-156530173 on Build GRCh38
rs764537559 Chr.1: 156530236-156530236 on Build GRCh38
rs1026199165 Chr.1: 156530052-156530052 on Build GRCh38
rs763254546 Chr.1: 156530329-156530329 on Build GRCh38
rs555867777 Chr.1: 156530489-156530489 on Build GRCh38
rs377534676 Chr.1: 156530078-156530078 on Build GRCh38
rs549472046 Chr.1: 156530081-156530081 on Build GRCh38
rs771627762 Chr.1: 156530132-156530132 on Build GRCh38
rs543972331 Chr.1: 156530051-156530051 on Build GRCh38
rs756649345 Chr.1: 156530116-156530116 on Build GRCh38
rs1029723851 Chr.1: 156530318-156530318 on Build GRCh38
rs760701034 Chr.1: 156530235-156530235 on Build GRCh38
rs911488259 Chr.1: 156530053-156530053 on Build GRCh38
rs771234105 Chr.1: 156530155-156530155 on Build GRCh38
rs747056561 Chr.1: 156530286-156530286 on Build GRCh38
rs754553573 Chr.1: 156530250-156530250 on Build GRCh38
rs777605892 Chr.1: 156530266-156530266 on Build GRCh38
rs781214033 Chr.1: 156530251-156530251 on Build GRCh38
rs765579722 Chr.1: 156530240-156530240 on Build GRCh38
rs746505 Chr.1: 156530336-156530336 on Build GRCh38
rs776545556 Chr.1: 156530147-156530147 on Build GRCh38
rs576499704 Chr.1: 156530046-156530046 on Build GRCh38
rs1036193393 Chr.1: 156530295-156530295 on Build GRCh38
rs769722333 Chr.1: 156530215-156530215 on Build GRCh38
rs968270068 Chr.1: 156530369-156530369 on Build GRCh38
rs778568841 Chr.1: 156530126-156530126 on Build GRCh38
rs967900452 Chr.1: 156530417-156530417 on Build GRCh38
rs537935834 Chr.1: 156530485-156530485 on Build GRCh38
rs200332849 Chr.1: 156530113-156530113 on Build GRCh38
rs756914637 Chr.1: 156530054-156530055 on Build GRCh38
rs368737629 Chr.1: 156530053-156530054 on Build GRCh38
rs370750568 Chr.1: 156530100-156530100 on Build GRCh38
rs372435844 Chr.1: 156530176-156530176 on Build GRCh38
rs374068925 Chr.1: 156530117-156530117 on Build GRCh38
rs745334511 Chr.1: 156530130-156530130 on Build GRCh38
rs374814459 Chr.1: 156530415-156530415 on Build GRCh38
rs143959866 Chr.1: 156530247-156530247 on Build GRCh38
rs765779794 Chr.1: 156530306-156530306 on Build GRCh38
rs917633816 Chr.1: 156530472-156530472 on Build GRCh38
rs191632538 Chr.1: 156530270-156530270 on Build GRCh38
rs149623112 Chr.1: 156530175-156530175 on Build GRCh38
rs751917329 Chr.1: 156530210-156530233 on Build GRCh38
rs369608391 Chr.1: 156530261-156530261 on Build GRCh38
rs767860195 Chr.1: 156530102-156530102 on Build GRCh38
rs746092752 Chr.1: 156530224-156530224 on Build GRCh38
rs139798063 Chr.1: 156530195-156530195 on Build GRCh38
rs386635735 Chr.1: 156530175-156530177 on Build GRCh38
rs369838719 Chr.1: 156530333-156530333 on Build GRCh38
rs758045685 Chr.1: 156530242-156530242 on Build GRCh38
rs766894424 Chr.1: 156530130-156530130 on Build GRCh38
rs979259231 Chr.1: 156530433-156530433 on Build GRCh38
rs1006175376 Chr.1: 156530293-156530293 on Build GRCh38
rs750722564 Chr.1: 156530465-156530465 on Build GRCh38
rs772302314 Chr.1: 156530277-156530277 on Build GRCh38
rs373353178 Chr.1: 156530260-156530260 on Build GRCh38
rs199748084 Chr.1: 156530220-156530220 on Build GRCh38
rs951499993 Chr.1: 156530052-156530053 on Build GRCh38
rs115742679 Chr.1: 156530304-156530304 on Build GRCh38
rs374687381 Chr.1: 156530145-156530145 on Build GRCh38
rs759048607 Chr.1: 156530334-156530334 on Build GRCh38
rs527875561 Chr.1: 156530153-156530153 on Build GRCh38
rs775597435 Chr.1: 156530281-156530281 on Build GRCh38
rs777603569 Chr.1: 156530349-156530349 on Build GRCh38
rs879038664 Chr.1: 156530410-156530410 on Build GRCh38
rs199533607 Chr.1: 156530184-156530184 on Build GRCh38
rs992753023 Chr.1: 156530437-156530437 on Build GRCh38
rs755337798 Chr.1: 156530235-156530238 on Build GRCh38
rs947802592 Chr.1: 156530482-156530482 on Build GRCh38
rs757641500 Chr.1: 156530185-156530185 on Build GRCh38

View Full Product Details