Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 55497524-55498016 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VOPP1
Entrez Gene ID
81552
Gene Name
vesicular, overexpressed in cancer, prosurvival protein 1
Gene Aliases
ECOP, GASP
UniGene
Hs.488307
Gene Chromosome Location
Chr.7: 55434406-55572520 on Build GRCh38
Transcripts
NM_001284282.1 NM_001284283.1 NM_001284284.1 NM_001321242.1 NM_001321243.1 NM_001321244.1 NM_001321246.1 NM_001321247.1 NM_001321248.1 NM_001321249.1 NM_001321250.1 NM_001321251.1 NM_030796.4 XM_011515539.1 XM_011515540.2 XM_011515541.2 XM_011515546.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3639800 c.54C>T p.L18L Chr.7: 55497637-55497637 on Build GRCh38
4862794 c.148C>T p.R50W Chr.7: 55497647-55497647 on Build GRCh38
601887 c.44C>T p.A15V Chr.7: 55497647-55497647 on Build GRCh38
3923839 c.37C>T p.L13L Chr.7: 55497654-55497654 on Build GRCh38
4892141 c.158C>T p.S53F Chr.7: 55497637-55497637 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766492998 Chr.7: 55497754-55497754 on Build GRCh38
rs765114951 Chr.7: 55497554-55497556 on Build GRCh38
rs891862078 Chr.7: 55497735-55497735 on Build GRCh38
rs749022382 Chr.7: 55497579-55497579 on Build GRCh38
rs112572178 Chr.7: 55497623-55497623 on Build GRCh38
rs758847862 Chr.7: 55497554-55497557 on Build GRCh38
rs71547935 Chr.7: 55497615-55497615 on Build GRCh38
rs762208927 Chr.7: 55497694-55497725 on Build GRCh38
rs58951420 Chr.7: 55497568-55497570 on Build GRCh38
rs750529839 Chr.7: 55497633-55497633 on Build GRCh38
rs575468925 Chr.7: 55497559-55497559 on Build GRCh38
rs769414502 Chr.7: 55497567-55497568 on Build GRCh38
rs747399146 Chr.7: 55497570-55497570 on Build GRCh38
rs774541984 Chr.7: 55497596-55497597 on Build GRCh38
rs148360132 Chr.7: 55497739-55497739 on Build GRCh38
rs149888342 Chr.7: 55497553-55497554 on Build GRCh38
rs764019062 Chr.7: 55497596-55497596 on Build GRCh38
rs560158176 Chr.7: 55497780-55497780 on Build GRCh38
rs772662024 Chr.7: 55497563-55497563 on Build GRCh38
rs183481368 Chr.7: 55497635-55497635 on Build GRCh38
rs879175033 Chr.7: 55497753-55497753 on Build GRCh38
rs765520861 Chr.7: 55497716-55497716 on Build GRCh38
rs1031447605 Chr.7: 55497738-55497738 on Build GRCh38
rs550516559 Chr.7: 55497884-55497884 on Build GRCh38
rs761883234 Chr.7: 55497713-55497713 on Build GRCh38
rs755315913 Chr.7: 55497646-55497646 on Build GRCh38
rs568803224 Chr.7: 55497555-55497556 on Build GRCh38
rs17673849 Chr.7: 55497919-55497919 on Build GRCh38
rs753927048 Chr.7: 55497566-55497566 on Build GRCh38
rs815954 Chr.7: 55497787-55497787 on Build GRCh38
rs561311509 Chr.7: 55497569-55497570 on Build GRCh38
rs202190740 Chr.7: 55497989-55497990 on Build GRCh38
rs1033052434 Chr.7: 55497562-55497562 on Build GRCh38
rs374585522 Chr.7: 55497588-55497588 on Build GRCh38
rs1024063186 Chr.7: 55497741-55497741 on Build GRCh38
rs796749202 Chr.7: 55497556-55497559 on Build GRCh38
rs907123090 Chr.7: 55497743-55497743 on Build GRCh38
rs776596470 Chr.7: 55497565-55497566 on Build GRCh38
rs548425478 Chr.7: 55497745-55497745 on Build GRCh38
rs770730198 Chr.7: 55497563-55497564 on Build GRCh38
rs762992096 Chr.7: 55497719-55497719 on Build GRCh38
rs1049372676 Chr.7: 55497605-55497605 on Build GRCh38
rs867943736 Chr.7: 55497841-55497841 on Build GRCh38
rs773126361 Chr.7: 55497718-55497718 on Build GRCh38
rs1021537971 Chr.7: 55497749-55497749 on Build GRCh38
rs907225002 Chr.7: 55497900-55497900 on Build GRCh38
rs117645790 Chr.7: 55497820-55497820 on Build GRCh38
rs1054361929 Chr.7: 55497943-55497943 on Build GRCh38
rs762152091 Chr.7: 55497609-55497609 on Build GRCh38
rs878870912 Chr.7: 55497554-55497554 on Build GRCh38
rs536517408 Chr.7: 55497576-55497576 on Build GRCh38
rs1019292100 Chr.7: 55497782-55497782 on Build GRCh38
rs778844285 Chr.7: 55497967-55497967 on Build GRCh38
rs751542194 Chr.7: 55497785-55497785 on Build GRCh38
rs762626984 Chr.7: 55497570-55497571 on Build GRCh38
rs538309074 Chr.7: 55497845-55497845 on Build GRCh38
rs372325187 Chr.7: 55497594-55497596 on Build GRCh38
rs370104426 Chr.7: 55497720-55497720 on Build GRCh38
rs779097426 Chr.7: 55497567-55497567 on Build GRCh38
rs951891039 Chr.7: 55497617-55497617 on Build GRCh38
rs58370285 Chr.7: 55497556-55497556 on Build GRCh38
rs754091209 Chr.7: 55497599-55497599 on Build GRCh38
rs202033592 Chr.7: 55497734-55497734 on Build GRCh38
rs780340196 Chr.7: 55497569-55497569 on Build GRCh38
rs774138253 Chr.7: 55497564-55497564 on Build GRCh38
rs536555377 Chr.7: 55497968-55497968 on Build GRCh38
rs554616126 Chr.7: 55497580-55497580 on Build GRCh38
rs984959011 Chr.7: 55497840-55497840 on Build GRCh38
rs756934629 Chr.7: 55497678-55497678 on Build GRCh38
rs955723446 Chr.7: 55497844-55497844 on Build GRCh38
rs748884467 Chr.7: 55497674-55497674 on Build GRCh38
rs938754806 Chr.7: 55497935-55497935 on Build GRCh38
rs577403967 Chr.7: 55497701-55497701 on Build GRCh38
rs985497579 Chr.7: 55497760-55497760 on Build GRCh38
rs564823906 Chr.7: 55497647-55497647 on Build GRCh38
rs745402895 Chr.7: 55497699-55497699 on Build GRCh38
rs1011326992 Chr.7: 55497980-55497980 on Build GRCh38
rs796685751 Chr.7: 55497555-55497557 on Build GRCh38
rs760576880 Chr.7: 55497589-55497589 on Build GRCh38
rs775348578 Chr.7: 55497568-55497569 on Build GRCh38
rs547189739 Chr.7: 55497793-55497793 on Build GRCh38
rs773975227 Chr.7: 55497581-55497581 on Build GRCh38
rs778225998 Chr.7: 55497686-55497686 on Build GRCh38
rs879221852 Chr.7: 55497568-55497570 on Build GRCh38
rs796896664 Chr.7: 55497555-55497558 on Build GRCh38
rs1000243186 Chr.7: 55497561-55497561 on Build GRCh38
rs776491396 Chr.7: 55497711-55497711 on Build GRCh38
rs747551674 Chr.7: 55497752-55497752 on Build GRCh38
rs373731224 Chr.7: 55497639-55497639 on Build GRCh38
rs1053165697 Chr.7: 55497947-55497947 on Build GRCh38
rs376201866 Chr.7: 55497613-55497613 on Build GRCh38
rs934011566 Chr.7: 55497558-55497558 on Build GRCh38
rs889873797 Chr.7: 55497560-55497560 on Build GRCh38
rs911450011 Chr.7: 55497872-55497872 on Build GRCh38
rs377339047 Chr.7: 55497740-55497740 on Build GRCh38
rs59713326 Chr.7: 55497565-55497565 on Build GRCh38
rs191334730 Chr.7: 55497582-55497582 on Build GRCh38
rs775534773 Chr.7: 55497703-55497703 on Build GRCh38
rs766903922 Chr.7: 55497638-55497638 on Build GRCh38
rs199539089 Chr.7: 55497704-55497704 on Build GRCh38
rs373905741 Chr.7: 55497565-55497567 on Build GRCh38
rs768557629 Chr.7: 55497709-55497709 on Build GRCh38
rs777503574 Chr.7: 55497790-55497790 on Build GRCh38

View Full Product Details