Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 158076022-158076518 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIRREL
Entrez Gene ID
55243
Gene Name
kin of IRRE like (Drosophila)
Gene Aliases
NEPH1
UniGene
Hs.272234
Gene Chromosome Location
Chr.1: 157993273-158100262 on Build GRCh38
Transcripts
NM_001286349.1 NM_018240.6 XM_005245305.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6192012 c.122C>A p.P41H Chr.1: 158076182-158076182 on Build GRCh38
1985600 c.126T>C p.C42C Chr.1: 158076186-158076186 on Build GRCh38
4696286 c.168C>T p.D56D Chr.1: 158076228-158076228 on Build GRCh38
4796119 c.5G>A p.G2D Chr.1: 158076248-158076248 on Build GRCh38
1255906 c.65G>A p.R22H Chr.1: 158076125-158076125 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769466372 Chr.1: 158076152-158076152 on Build GRCh38
rs62642493 Chr.1: 158076122-158076122 on Build GRCh38
rs748515520 Chr.1: 158076258-158076258 on Build GRCh38
rs368832771 Chr.1: 158076314-158076314 on Build GRCh38
rs781460289 Chr.1: 158076254-158076254 on Build GRCh38
rs748825421 Chr.1: 158076280-158076280 on Build GRCh38
rs757378555 Chr.1: 158076086-158076086 on Build GRCh38
rs762552084 Chr.1: 158076161-158076161 on Build GRCh38
rs777557454 Chr.1: 158076274-158076274 on Build GRCh38
rs563432262 Chr.1: 158076397-158076397 on Build GRCh38
rs201035657 Chr.1: 158076310-158076310 on Build GRCh38
rs774448722 Chr.1: 158076284-158076284 on Build GRCh38
rs891179069 Chr.1: 158076132-158076132 on Build GRCh38
rs771052077 Chr.1: 158076169-158076169 on Build GRCh38
rs772892194 Chr.1: 158076153-158076153 on Build GRCh38
rs758969672 Chr.1: 158076114-158076114 on Build GRCh38
rs138764677 Chr.1: 158076244-158076244 on Build GRCh38
rs567633103 Chr.1: 158076365-158076365 on Build GRCh38
rs944962156 Chr.1: 158076449-158076449 on Build GRCh38
rs772012319 Chr.1: 158076286-158076286 on Build GRCh38
rs75560227 Chr.1: 158076448-158076448 on Build GRCh38
rs759436908 Chr.1: 158076285-158076285 on Build GRCh38
rs768835287 Chr.1: 158076121-158076121 on Build GRCh38
rs770502338 Chr.1: 158076283-158076283 on Build GRCh38
rs943888963 Chr.1: 158076482-158076482 on Build GRCh38
rs200949520 Chr.1: 158076306-158076306 on Build GRCh38
rs75187201 Chr.1: 158076469-158076469 on Build GRCh38
rs557900518 Chr.1: 158076174-158076174 on Build GRCh38
rs780542461 Chr.1: 158076228-158076228 on Build GRCh38
rs745914249 Chr.1: 158076096-158076096 on Build GRCh38
rs979901967 Chr.1: 158076323-158076323 on Build GRCh38
rs761979056 Chr.1: 158076183-158076183 on Build GRCh38
rs981154126 Chr.1: 158076303-158076303 on Build GRCh38
rs1042485717 Chr.1: 158076483-158076483 on Build GRCh38
rs780525861 Chr.1: 158076116-158076116 on Build GRCh38
rs752022419 Chr.1: 158076229-158076229 on Build GRCh38
rs371294912 Chr.1: 158076170-158076170 on Build GRCh38
rs1053217438 Chr.1: 158076418-158076418 on Build GRCh38
rs769403326 Chr.1: 158076262-158076262 on Build GRCh38
rs539625524 Chr.1: 158076124-158076124 on Build GRCh38
rs912858240 Chr.1: 158076422-158076422 on Build GRCh38
rs764393617 Chr.1: 158076064-158076064 on Build GRCh38
rs536706183 Chr.1: 158076186-158076186 on Build GRCh38
rs780523041 Chr.1: 158076349-158076349 on Build GRCh38
rs77874069 Chr.1: 158076442-158076442 on Build GRCh38
rs191487619 Chr.1: 158076175-158076175 on Build GRCh38
rs1052701730 Chr.1: 158076090-158076090 on Build GRCh38
rs919826369 Chr.1: 158076342-158076342 on Build GRCh38
rs181320133 Chr.1: 158076089-158076089 on Build GRCh38
rs149775287 Chr.1: 158076307-158076307 on Build GRCh38
rs1050039797 Chr.1: 158076125-158076125 on Build GRCh38
rs545337814 Chr.1: 158076366-158076366 on Build GRCh38
rs925696617 Chr.1: 158076060-158076060 on Build GRCh38
rs201637798 Chr.1: 158076292-158076292 on Build GRCh38
rs753963750 Chr.1: 158076082-158076082 on Build GRCh38
rs747385256 Chr.1: 158076117-158076117 on Build GRCh38
rs758414667 Chr.1: 158076196-158076196 on Build GRCh38

View Full Product Details