Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 158086523-158087070 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIRREL
Entrez Gene ID
55243
Gene Name
kin of IRRE like (Drosophila)
Gene Aliases
NEPH1
UniGene
Hs.272234
Gene Chromosome Location
Chr.1: 157993273-158100262 on Build GRCh38
Transcripts
NM_001286349.1 NM_018240.6 XM_005245305.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6527830 c.644T>C p.I215T Chr.1: 158086729-158086729 on Build GRCh38
6527831 c.644T>C p.I215T Chr.1: 158086729-158086729 on Build GRCh38
7264578 c.657G>A p.V219V Chr.1: 158086742-158086742 on Build GRCh38
3711178 c.658C>A p.H220N Chr.1: 158086743-158086743 on Build GRCh38
1583573 c.645C>T p.I215I Chr.1: 158086730-158086730 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs11264909 Chr.1: 158086989-158086989 on Build GRCh38
rs964930898 Chr.1: 158086554-158086554 on Build GRCh38
rs139544241 Chr.1: 158087021-158087021 on Build GRCh38
rs1047631371 Chr.1: 158086738-158086738 on Build GRCh38
rs756271701 Chr.1: 158086801-158086801 on Build GRCh38
rs373621646 Chr.1: 158086869-158086869 on Build GRCh38
rs762459853 Chr.1: 158086793-158086793 on Build GRCh38
rs954703549 Chr.1: 158086579-158086579 on Build GRCh38
rs749578512 Chr.1: 158086771-158086771 on Build GRCh38
rs1043491627 Chr.1: 158087016-158087016 on Build GRCh38
rs780382516 Chr.1: 158086847-158086847 on Build GRCh38
rs1030382651 Chr.1: 158086851-158086851 on Build GRCh38
rs979018258 Chr.1: 158086655-158086655 on Build GRCh38
rs749029312 Chr.1: 158086672-158086672 on Build GRCh38
rs201608779 Chr.1: 158086758-158086759 on Build GRCh38
rs942759526 Chr.1: 158086903-158086903 on Build GRCh38
rs751993059 Chr.1: 158086589-158086589 on Build GRCh38
rs60115669 Chr.1: 158087005-158087005 on Build GRCh38
rs934171234 Chr.1: 158086570-158086570 on Build GRCh38
rs1012346907 Chr.1: 158086839-158086839 on Build GRCh38
rs978966253 Chr.1: 158086927-158086927 on Build GRCh38
rs750545390 Chr.1: 158086782-158086782 on Build GRCh38
rs762400981 Chr.1: 158086944-158086944 on Build GRCh38
rs900310046 Chr.1: 158086849-158086849 on Build GRCh38
rs956149763 Chr.1: 158086809-158086828 on Build GRCh38
rs780474346 Chr.1: 158086746-158086746 on Build GRCh38
rs927472352 Chr.1: 158086675-158086675 on Build GRCh38
rs974051464 Chr.1: 158086906-158086906 on Build GRCh38
rs543970318 Chr.1: 158086954-158086954 on Build GRCh38
rs577374783 Chr.1: 158086952-158086952 on Build GRCh38
rs41273463 Chr.1: 158086759-158086759 on Build GRCh38
rs1038309946 Chr.1: 158086654-158086654 on Build GRCh38
rs755888560 Chr.1: 158086647-158086647 on Build GRCh38
rs769854392 Chr.1: 158086726-158086726 on Build GRCh38
rs374520719 Chr.1: 158086577-158086577 on Build GRCh38
rs896230979 Chr.1: 158086731-158086731 on Build GRCh38
rs554856265 Chr.1: 158086565-158086565 on Build GRCh38
rs988897219 Chr.1: 158086866-158086866 on Build GRCh38
rs1011970822 Chr.1: 158087006-158087006 on Build GRCh38
rs540418640 Chr.1: 158086810-158086810 on Build GRCh38
rs146029367 Chr.1: 158086569-158086569 on Build GRCh38
rs188683712 Chr.1: 158086697-158086697 on Build GRCh38
rs927556593 Chr.1: 158086951-158086951 on Build GRCh38
rs939196749 Chr.1: 158086987-158086987 on Build GRCh38
rs986957988 Chr.1: 158086882-158086882 on Build GRCh38
rs7364493 Chr.1: 158086861-158086861 on Build GRCh38
rs996425330 Chr.1: 158086850-158086850 on Build GRCh38
rs777659418 Chr.1: 158086666-158086666 on Build GRCh38
rs947637833 Chr.1: 158086610-158086610 on Build GRCh38
rs978377165 Chr.1: 158086858-158086858 on Build GRCh38
rs1030849991 Chr.1: 158086571-158086571 on Build GRCh38
rs1016095047 Chr.1: 158086551-158086551 on Build GRCh38
rs911314091 Chr.1: 158086884-158086884 on Build GRCh38
rs538710206 Chr.1: 158086616-158086616 on Build GRCh38
rs139318668 Chr.1: 158086837-158086838 on Build GRCh38
rs772538040 Chr.1: 158087041-158087041 on Build GRCh38
rs913448453 Chr.1: 158086596-158086596 on Build GRCh38
rs747088272 Chr.1: 158086742-158086742 on Build GRCh38
rs555277710 Chr.1: 158086816-158086816 on Build GRCh38
rs911295213 Chr.1: 158086578-158086578 on Build GRCh38
rs941694049 Chr.1: 158087029-158087029 on Build GRCh38
rs1047539324 Chr.1: 158087013-158087013 on Build GRCh38
rs1016211826 Chr.1: 158086845-158086845 on Build GRCh38
rs192057980 Chr.1: 158086923-158086923 on Build GRCh38
rs762334234 Chr.1: 158086730-158086730 on Build GRCh38
rs1047424625 Chr.1: 158086783-158086783 on Build GRCh38
rs886138627 Chr.1: 158086785-158086785 on Build GRCh38

View Full Product Details