Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 102456219-102456746 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALKBH4
Entrez Gene ID
54784
Gene Name
alkB homolog 4, lysine demethylase
Gene Aliases
ABH4
UniGene
Hs.658598
Gene Chromosome Location
Chr.7: 102456220-102464874 on Build GRCh38
Transcripts
NM_017621.3 XM_005250464.2 XM_005250465.2 NM_001126340.2 NM_001271818.1 NM_001271819.1 NM_032831.3

Gene Symbol
ORAI2
Entrez Gene ID
80228
Gene Name
ORAI calcium release-activated calcium modulator 2
Gene Aliases
C7orf19, CBCIP2, MEM142B, TMEM142B
UniGene
Hs.363308
Gene Chromosome Location
Chr.7: 102433530-102456821 on Build GRCh38
Transcripts
NM_017621.3 XM_005250464.2 XM_005250465.2 NM_001126340.2 NM_001271818.1 NM_001271819.1 NM_032831.3

SNP Detail

SNP ID Chromosome Location
rs987656077 Chr.7: 102456701-102456701 on Build GRCh38
rs894455702 Chr.7: 102456631-102456631 on Build GRCh38
rs1009488457 Chr.7: 102456394-102456394 on Build GRCh38
rs77522041 Chr.7: 102456386-102456386 on Build GRCh38
rs749263912 Chr.7: 102456649-102456649 on Build GRCh38
rs898326976 Chr.7: 102456531-102456531 on Build GRCh38
rs947639178 Chr.7: 102456280-102456280 on Build GRCh38
rs958805697 Chr.7: 102456439-102456439 on Build GRCh38
rs1039667005 Chr.7: 102456668-102456668 on Build GRCh38
rs763697062 Chr.7: 102456410-102456410 on Build GRCh38
rs1046923430 Chr.7: 102456559-102456559 on Build GRCh38
rs546513349 Chr.7: 102456329-102456329 on Build GRCh38
rs934961450 Chr.7: 102456330-102456330 on Build GRCh38
rs924897540 Chr.7: 102456480-102456480 on Build GRCh38
rs981078642 Chr.7: 102456532-102456532 on Build GRCh38
rs1022228607 Chr.7: 102456455-102456455 on Build GRCh38
rs951032546 Chr.7: 102456518-102456518 on Build GRCh38
rs986451266 Chr.7: 102456499-102456499 on Build GRCh38
rs1002910109 Chr.7: 102456411-102456411 on Build GRCh38
rs1031780027 Chr.7: 102456666-102456666 on Build GRCh38
rs887064232 Chr.7: 102456572-102456572 on Build GRCh38
rs905012306 Chr.7: 102456296-102456296 on Build GRCh38
rs933570711 Chr.7: 102456492-102456492 on Build GRCh38
rs890751701 Chr.7: 102456413-102456413 on Build GRCh38
rs868409358 Chr.7: 102456438-102456438 on Build GRCh38
rs978082374 Chr.7: 102456474-102456474 on Build GRCh38
rs754865704 Chr.7: 102456665-102456665 on Build GRCh38
rs1053774635 Chr.7: 102456381-102456381 on Build GRCh38
rs890437662 Chr.7: 102456658-102456658 on Build GRCh38
rs1008216171 Chr.7: 102456415-102456415 on Build GRCh38
rs576056534 Chr.7: 102456302-102456302 on Build GRCh38
rs557780683 Chr.7: 102456720-102456720 on Build GRCh38
rs994968905 Chr.7: 102456689-102456689 on Build GRCh38
rs1026703416 Chr.7: 102456498-102456498 on Build GRCh38
rs890913990 Chr.7: 102456356-102456356 on Build GRCh38
rs923609609 Chr.7: 102456308-102456308 on Build GRCh38
rs963709700 Chr.7: 102456723-102456723 on Build GRCh38
rs561787500 Chr.7: 102456529-102456529 on Build GRCh38
rs568757143 Chr.7: 102456700-102456700 on Build GRCh38
rs1053806298 Chr.7: 102456544-102456544 on Build GRCh38
rs563832365 Chr.7: 102456254-102456254 on Build GRCh38
rs539389003 Chr.7: 102456702-102456702 on Build GRCh38
rs778806995 Chr.7: 102456669-102456669 on Build GRCh38
rs539071712 Chr.7: 102456677-102456677 on Build GRCh38
rs942449186 Chr.7: 102456357-102456357 on Build GRCh38
rs931127783 Chr.7: 102456539-102456539 on Build GRCh38
rs1025520963 Chr.7: 102456396-102456396 on Build GRCh38
rs1007831378 Chr.7: 102456664-102456664 on Build GRCh38
rs977656310 Chr.7: 102456652-102456652 on Build GRCh38
rs550503159 Chr.7: 102456645-102456645 on Build GRCh38
rs912218746 Chr.7: 102456508-102456508 on Build GRCh38
rs553470803 Chr.7: 102456593-102456593 on Build GRCh38
rs757358914 Chr.7: 102456420-102456420 on Build GRCh38
rs968874612 Chr.7: 102456456-102456456 on Build GRCh38
rs1039973130 Chr.7: 102456375-102456375 on Build GRCh38
rs926495408 Chr.7: 102456533-102456533 on Build GRCh38
rs1050709409 Chr.7: 102456346-102456346 on Build GRCh38
rs913948901 Chr.7: 102456581-102456581 on Build GRCh38
rs528889294 Chr.7: 102456638-102456638 on Build GRCh38
rs781184603 Chr.7: 102456524-102456524 on Build GRCh38
rs898302658 Chr.7: 102456376-102456376 on Build GRCh38
rs1035706457 Chr.7: 102456585-102456585 on Build GRCh38
rs565302730 Chr.7: 102456639-102456639 on Build GRCh38
rs1002773779 Chr.7: 102456580-102456580 on Build GRCh38
rs142611777 Chr.7: 102456304-102456304 on Build GRCh38
rs116995449 Chr.7: 102456409-102456409 on Build GRCh38
rs77123592 Chr.7: 102456435-102456435 on Build GRCh38
rs991559303 Chr.7: 102456444-102456444 on Build GRCh38

View Full Product Details