Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 154946420-154946916 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PAXIP1
Entrez Gene ID
22976
Gene Name
PAX interacting protein 1
Gene Aliases
CAGF28, CAGF29, PACIP1, PAXIP1L, PTIP, TNRC2
UniGene
Hs.443881
Gene Chromosome Location
Chr.7: 154943690-155003393 on Build GRCh38
Transcripts
NM_007349.3 XM_005249539.1 XM_011515982.2 XM_011515983.1 XM_017011881.1 NR_024476.1 NR_024477.1

Gene Symbol
PAXIP1-AS2
Entrez Gene ID
100132707
Gene Name
PAXIP1 antisense RNA 2
Gene Aliases
PAXIP1OS
UniGene
Hs.586358
Gene Chromosome Location
Chr.7: 154928517-154949490 on Build GRCh38
Transcripts
NM_007349.3 XM_005249539.1 XM_011515982.2 XM_011515983.1 XM_017011881.1 NR_024476.1 NR_024477.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
150431 c.2935A>G p.M979V Chr.7: 154946699-154946699 on Build GRCh38
369105 c.2938G>T p.E980* Chr.7: 154946696-154946696 on Build GRCh38
1496552 c.3007C>T p.R1003* Chr.7: 154946536-154946536 on Build GRCh38
4762968 c.2921C>G p.S974C Chr.7: 154946713-154946713 on Build GRCh38
6109471 c.2866A>G p.T956A Chr.7: 154946768-154946768 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774684247 Chr.7: 154946577-154946577 on Build GRCh38
rs756344863 Chr.7: 154946754-154946754 on Build GRCh38
rs61752002 Chr.7: 154946694-154946694 on Build GRCh38
rs200574109 Chr.7: 154946491-154946491 on Build GRCh38
rs760228731 Chr.7: 154946610-154946610 on Build GRCh38
rs369240498 Chr.7: 154946500-154946500 on Build GRCh38
rs1041597927 Chr.7: 154946874-154946874 on Build GRCh38
rs754964854 Chr.7: 154946765-154946765 on Build GRCh38
rs758829567 Chr.7: 154946799-154946799 on Build GRCh38
rs373635173 Chr.7: 154946660-154946660 on Build GRCh38
rs762107361 Chr.7: 154946579-154946579 on Build GRCh38
rs747800895 Chr.7: 154946528-154946528 on Build GRCh38
rs776112228 Chr.7: 154946860-154946860 on Build GRCh38
rs749818360 Chr.7: 154946659-154946659 on Build GRCh38
rs748534474 Chr.7: 154946675-154946675 on Build GRCh38
rs368699452 Chr.7: 154946474-154946474 on Build GRCh38
rs3501 Chr.7: 154946699-154946699 on Build GRCh38
rs773216114 Chr.7: 154946652-154946653 on Build GRCh38
rs765031748 Chr.7: 154946620-154946620 on Build GRCh38
rs749770706 Chr.7: 154946569-154946569 on Build GRCh38
rs373308839 Chr.7: 154946619-154946619 on Build GRCh38
rs765151848 Chr.7: 154946522-154946522 on Build GRCh38
rs376214339 Chr.7: 154946517-154946517 on Build GRCh38
rs781585176 Chr.7: 154946642-154946642 on Build GRCh38
rs770407366 Chr.7: 154946696-154946696 on Build GRCh38
rs923098480 Chr.7: 154946885-154946885 on Build GRCh38
rs765292789 Chr.7: 154946700-154946700 on Build GRCh38
rs770626267 Chr.7: 154946835-154946835 on Build GRCh38
rs369541196 Chr.7: 154946580-154946580 on Build GRCh38
rs756186230 Chr.7: 154946654-154946654 on Build GRCh38
rs763947571 Chr.7: 154946708-154946708 on Build GRCh38
rs758222542 Chr.7: 154946635-154946635 on Build GRCh38
rs945934801 Chr.7: 154946851-154946851 on Build GRCh38
rs745881397 Chr.7: 154946444-154946444 on Build GRCh38
rs750740629 Chr.7: 154946753-154946753 on Build GRCh38
rs376467437 Chr.7: 154946649-154946649 on Build GRCh38
rs772405233 Chr.7: 154946680-154946680 on Build GRCh38
rs542075029 Chr.7: 154946476-154946476 on Build GRCh38
rs773913644 Chr.7: 154946478-154946478 on Build GRCh38
rs1036807007 Chr.7: 154946876-154946876 on Build GRCh38
rs753956808 Chr.7: 154946756-154946756 on Build GRCh38
rs762887463 Chr.7: 154946707-154946707 on Build GRCh38
rs867868549 Chr.7: 154946650-154946650 on Build GRCh38
rs745971788 Chr.7: 154946535-154946535 on Build GRCh38
rs753793164 Chr.7: 154946492-154946492 on Build GRCh38
rs200215885 Chr.7: 154946655-154946655 on Build GRCh38
rs183889833 Chr.7: 154946832-154946832 on Build GRCh38
rs914295101 Chr.7: 154946664-154946664 on Build GRCh38
rs778100830 Chr.7: 154946812-154946812 on Build GRCh38
rs760374133 Chr.7: 154946685-154946685 on Build GRCh38
rs779120651 Chr.7: 154946443-154946444 on Build GRCh38
rs77319557 Chr.7: 154946488-154946488 on Build GRCh38
rs1015181241 Chr.7: 154946644-154946644 on Build GRCh38
rs770202739 Chr.7: 154946449-154946449 on Build GRCh38
rs776022267 Chr.7: 154946473-154946473 on Build GRCh38
rs748301397 Chr.7: 154946484-154946487 on Build GRCh38
rs746099872 Chr.7: 154946623-154946623 on Build GRCh38
rs192118427 Chr.7: 154946499-154946499 on Build GRCh38
rs748626037 Chr.7: 154946783-154946783 on Build GRCh38
rs958617978 Chr.7: 154946766-154946766 on Build GRCh38
rs779445797 Chr.7: 154946774-154946774 on Build GRCh38
rs553823627 Chr.7: 154946648-154946648 on Build GRCh38
rs945882454 Chr.7: 154946513-154946513 on Build GRCh38
rs765883313 Chr.7: 154946616-154946616 on Build GRCh38
rs775515966 Chr.7: 154946702-154946702 on Build GRCh38
rs972025587 Chr.7: 154946646-154946646 on Build GRCh38
rs772893135 Chr.7: 154946683-154946683 on Build GRCh38
rs914506466 Chr.7: 154946833-154946833 on Build GRCh38
rs752541952 Chr.7: 154946629-154946629 on Build GRCh38
rs772803152 Chr.7: 154946604-154946604 on Build GRCh38
rs772222813 Chr.7: 154946546-154946546 on Build GRCh38
rs117213645 Chr.7: 154946482-154946482 on Build GRCh38
rs769860072 Chr.7: 154946546-154946546 on Build GRCh38
rs779354009 Chr.7: 154946661-154946661 on Build GRCh38
rs768966226 Chr.7: 154946570-154946570 on Build GRCh38
rs780471773 Chr.7: 154946555-154946555 on Build GRCh38
rs967034739 Chr.7: 154946665-154946665 on Build GRCh38
rs867060120 Chr.7: 154946544-154946544 on Build GRCh38
rs61752001 Chr.7: 154946508-154946508 on Build GRCh38
rs1041545529 Chr.7: 154946597-154946597 on Build GRCh38

View Full Product Details