Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 166848857-166849365 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POGK
Entrez Gene ID
57645
Gene Name
pogo transposable element with KRAB domain
Gene Aliases
BASS2, KRBOX2, LST003
UniGene
Hs.432752
Gene Chromosome Location
Chr.1: 166839404-166856344 on Build GRCh38
Transcripts
NM_001314014.1 NM_017542.4 XM_005245370.3 XM_011509814.2 XM_011509815.1 XM_017001914.1 XM_017001915.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6483451 c.492G>C p.W164C Chr.1: 166849071-166849071 on Build GRCh38
898972 c.703C>T p.R235C Chr.1: 166849282-166849282 on Build GRCh38
1499416 c.604G>A p.A202T Chr.1: 166849183-166849183 on Build GRCh38
4880456 c.750A>G p.A250A Chr.1: 166849329-166849329 on Build GRCh38
6751761 c.566A>C p.K189T Chr.1: 166849145-166849145 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770055510 Chr.1: 166849315-166849315 on Build GRCh38
rs34864748 Chr.1: 166849140-166849141 on Build GRCh38
rs139118899 Chr.1: 166849215-166849215 on Build GRCh38
rs140373616 Chr.1: 166849066-166849066 on Build GRCh38
rs371202482 Chr.1: 166849334-166849334 on Build GRCh38
rs781090181 Chr.1: 166849108-166849108 on Build GRCh38
rs770571769 Chr.1: 166849135-166849135 on Build GRCh38
rs755901637 Chr.1: 166849125-166849125 on Build GRCh38
rs770518724 Chr.1: 166849051-166849051 on Build GRCh38
rs1043069783 Chr.1: 166848924-166848924 on Build GRCh38
rs751500100 Chr.1: 166849091-166849091 on Build GRCh38
rs573690726 Chr.1: 166849079-166849079 on Build GRCh38
rs762404380 Chr.1: 166849046-166849046 on Build GRCh38
rs148807160 Chr.1: 166849308-166849308 on Build GRCh38
rs764497288 Chr.1: 166848952-166848952 on Build GRCh38
rs112544772 Chr.1: 166849176-166849176 on Build GRCh38
rs777165406 Chr.1: 166849172-166849172 on Build GRCh38
rs745409732 Chr.1: 166849304-166849304 on Build GRCh38
rs544727342 Chr.1: 166848970-166848970 on Build GRCh38
rs916136060 Chr.1: 166848880-166848880 on Build GRCh38
rs769334733 Chr.1: 166849160-166849160 on Build GRCh38
rs565818418 Chr.1: 166849246-166849246 on Build GRCh38
rs773691171 Chr.1: 166849322-166849322 on Build GRCh38
rs749033738 Chr.1: 166849274-166849274 on Build GRCh38
rs201306097 Chr.1: 166849182-166849182 on Build GRCh38
rs553788517 Chr.1: 166849085-166849085 on Build GRCh38
rs774072094 Chr.1: 166849142-166849142 on Build GRCh38
rs143520580 Chr.1: 166849073-166849073 on Build GRCh38
rs757029029 Chr.1: 166849283-166849283 on Build GRCh38
rs529097030 Chr.1: 166848963-166848963 on Build GRCh38
rs772892848 Chr.1: 166849045-166849045 on Build GRCh38
rs754972305 Chr.1: 166849099-166849099 on Build GRCh38
rs111752271 Chr.1: 166849113-166849113 on Build GRCh38
rs766578918 Chr.1: 166849189-166849189 on Build GRCh38
rs777023778 Chr.1: 166849309-166849309 on Build GRCh38
rs747937758 Chr.1: 166849121-166849121 on Build GRCh38
rs61744366 Chr.1: 166849126-166849126 on Build GRCh38
rs761310983 Chr.1: 166848889-166848889 on Build GRCh38
rs992049377 Chr.1: 166849088-166849088 on Build GRCh38
rs375997972 Chr.1: 166849106-166849106 on Build GRCh38
rs202168363 Chr.1: 166849036-166849036 on Build GRCh38
rs748914183 Chr.1: 166849131-166849131 on Build GRCh38
rs769610784 Chr.1: 166849314-166849314 on Build GRCh38
rs200681234 Chr.1: 166848990-166848990 on Build GRCh38
rs745358664 Chr.1: 166849149-166849149 on Build GRCh38
rs538402061 Chr.1: 166849179-166849179 on Build GRCh38
rs147324383 Chr.1: 166849258-166849258 on Build GRCh38
rs767478116 Chr.1: 166849208-166849208 on Build GRCh38
rs1022499443 Chr.1: 166849022-166849022 on Build GRCh38
rs772660669 Chr.1: 166848920-166848920 on Build GRCh38
rs77068976 Chr.1: 166848955-166848955 on Build GRCh38
rs763286049 Chr.1: 166849326-166849326 on Build GRCh38
rs763093136 Chr.1: 166849186-166849186 on Build GRCh38
rs776030495 Chr.1: 166848916-166848916 on Build GRCh38
rs776286043 Chr.1: 166848887-166848887 on Build GRCh38
rs764695456 Chr.1: 166849065-166849065 on Build GRCh38
rs755023488 Chr.1: 166849201-166849201 on Build GRCh38
rs774659230 Chr.1: 166849037-166849037 on Build GRCh38
rs761145631 Chr.1: 166848934-166848934 on Build GRCh38
rs144587246 Chr.1: 166849052-166849052 on Build GRCh38
rs907483954 Chr.1: 166848882-166848882 on Build GRCh38
rs368935998 Chr.1: 166848911-166848911 on Build GRCh38
rs781298242 Chr.1: 166848919-166848919 on Build GRCh38
rs768449678 Chr.1: 166849013-166849013 on Build GRCh38
rs113329811 Chr.1: 166849181-166849181 on Build GRCh38
rs2075946 Chr.1: 166849329-166849329 on Build GRCh38
rs994749216 Chr.1: 166849012-166849012 on Build GRCh38
rs765417149 Chr.1: 166849069-166849069 on Build GRCh38
rs751692773 Chr.1: 166849197-166849197 on Build GRCh38
rs774520557 Chr.1: 166849333-166849333 on Build GRCh38
rs746897176 Chr.1: 166849010-166849010 on Build GRCh38
rs777559044 Chr.1: 166849272-166849272 on Build GRCh38
rs753218448 Chr.1: 166848925-166848925 on Build GRCh38
rs780894099 Chr.1: 166849032-166849032 on Build GRCh38
rs147708165 Chr.1: 166848954-166848954 on Build GRCh38
rs758471368 Chr.1: 166848982-166848982 on Build GRCh38
rs767952551 Chr.1: 166848923-166848923 on Build GRCh38
rs962868946 Chr.1: 166849161-166849161 on Build GRCh38
rs779182131 Chr.1: 166848967-166848967 on Build GRCh38
rs202109752 Chr.1: 166849290-166849290 on Build GRCh38
rs762507449 Chr.1: 166848921-166848921 on Build GRCh38
rs77682365 Chr.1: 166849185-166849185 on Build GRCh38

View Full Product Details