Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 73793569-73794084 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UBE2W
Entrez Gene ID
55284
Gene Name
ubiquitin conjugating enzyme E2 W (putative)
Gene Aliases
UBC-16, UBC16
UniGene
Hs.128841
Gene Chromosome Location
Chr.8: 73780097-73878910 on Build GRCh38
Transcripts
NM_001001481.2 NM_001271015.1 NM_018299.4 NR_073119.1 NR_073120.1 NR_073121.1

SNP Detail

SNP ID Chromosome Location
rs529523225 Chr.8: 73793653-73793653 on Build GRCh38
rs372744397 Chr.8: 73793771-73793771 on Build GRCh38
rs894816714 Chr.8: 73793676-73793676 on Build GRCh38
rs375753648 Chr.8: 73794014-73794014 on Build GRCh38
rs981605652 Chr.8: 73794002-73794002 on Build GRCh38
rs879190021 Chr.8: 73793705-73793705 on Build GRCh38
rs990193332 Chr.8: 73793950-73793950 on Build GRCh38
rs373157611 Chr.8: 73793907-73793907 on Build GRCh38
rs1018198633 Chr.8: 73793892-73793892 on Build GRCh38
rs879571410 Chr.8: 73793711-73793711 on Build GRCh38
rs768472897 Chr.8: 73793863-73793863 on Build GRCh38
rs896961239 Chr.8: 73793759-73793759 on Build GRCh38
rs3739349 Chr.8: 73793651-73793651 on Build GRCh38
rs957403307 Chr.8: 73793945-73793945 on Build GRCh38
rs560346343 Chr.8: 73793997-73793997 on Build GRCh38
rs928933351 Chr.8: 73793616-73793616 on Build GRCh38
rs377306564 Chr.8: 73794035-73794035 on Build GRCh38
rs758414758 Chr.8: 73793627-73793627 on Build GRCh38
rs1020393908 Chr.8: 73793959-73793959 on Build GRCh38
rs544501485 Chr.8: 73793745-73793745 on Build GRCh38
rs1054772918 Chr.8: 73793660-73793660 on Build GRCh38
rs746820306 Chr.8: 73793740-73793740 on Build GRCh38
rs183444055 Chr.8: 73794008-73794008 on Build GRCh38
rs1029921990 Chr.8: 73793848-73793848 on Build GRCh38
rs941080508 Chr.8: 73793679-73793679 on Build GRCh38
rs114676188 Chr.8: 73793984-73793984 on Build GRCh38
rs868231543 Chr.8: 73793812-73793812 on Build GRCh38
rs577381225 Chr.8: 73793663-73793664 on Build GRCh38
rs139128343 Chr.8: 73793768-73793769 on Build GRCh38
rs368841087 Chr.8: 73793731-73793731 on Build GRCh38
rs777927055 Chr.8: 73793872-73793872 on Build GRCh38
rs73322646 Chr.8: 73793960-73793960 on Build GRCh38
rs192629589 Chr.8: 73793770-73793770 on Build GRCh38

View Full Product Details