Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 130079810-130080320 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASAP1
Entrez Gene ID
50807
Gene Name
ArfGAP with SH3 domain, ankyrin repeat and PH domain 1
Gene Aliases
AMAP1, CENTB4, DDEF1, PAG2, PAP, ZG14P
UniGene
Hs.655552
Gene Chromosome Location
Chr.8: 130052104-130443848 on Build GRCh38
Transcripts
NM_001247996.1 NM_018482.3 XM_005250925.1 XM_006716563.3 XM_006716564.1 XM_006716565.3 XM_006716566.1 XM_011517052.2 XM_011517053.1 XM_017013467.1 XM_017013468.1 XM_017013469.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6605420 c.2577C>T p.N859N Chr.8: 130079967-130079967 on Build GRCh38
4663589 c.2628A>G p.L876L Chr.8: 130079916-130079916 on Build GRCh38
6179991 c.2578G>T p.D860Y Chr.8: 130079966-130079966 on Build GRCh38
278994 c.2625A>G p.G875G Chr.8: 130079919-130079919 on Build GRCh38
3896753 c.2639C>T p.S880L Chr.8: 130079905-130079905 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781750725 Chr.8: 130080009-130080009 on Build GRCh38
rs549439026 Chr.8: 130080229-130080229 on Build GRCh38
rs946165687 Chr.8: 130080073-130080073 on Build GRCh38
rs563931565 Chr.8: 130080181-130080181 on Build GRCh38
rs138900968 Chr.8: 130079958-130079958 on Build GRCh38
rs755770115 Chr.8: 130079888-130079888 on Build GRCh38
rs985077633 Chr.8: 130079836-130079836 on Build GRCh38
rs759703780 Chr.8: 130079964-130079964 on Build GRCh38
rs74531144 Chr.8: 130080106-130080106 on Build GRCh38
rs778832409 Chr.8: 130079916-130079916 on Build GRCh38
rs751906732 Chr.8: 130079985-130079985 on Build GRCh38
rs536814056 Chr.8: 130080055-130080055 on Build GRCh38
rs555562548 Chr.8: 130080057-130080057 on Build GRCh38
rs990767450 Chr.8: 130080045-130080045 on Build GRCh38
rs1015002729 Chr.8: 130080147-130080147 on Build GRCh38
rs200185585 Chr.8: 130079874-130079874 on Build GRCh38
rs933399237 Chr.8: 130079864-130079864 on Build GRCh38
rs951055590 Chr.8: 130079834-130079834 on Build GRCh38
rs964928263 Chr.8: 130080065-130080065 on Build GRCh38
rs746386440 Chr.8: 130080074-130080074 on Build GRCh38
rs938365766 Chr.8: 130080082-130080082 on Build GRCh38
rs370789875 Chr.8: 130079967-130079967 on Build GRCh38
rs771698608 Chr.8: 130080138-130080138 on Build GRCh38
rs895023337 Chr.8: 130079897-130079897 on Build GRCh38
rs764639162 Chr.8: 130079984-130079984 on Build GRCh38
rs748822682 Chr.8: 130079908-130079908 on Build GRCh38
rs35210585 Chr.8: 130079972-130079972 on Build GRCh38
rs576293757 Chr.8: 130080030-130080030 on Build GRCh38
rs762900734 Chr.8: 130079998-130079999 on Build GRCh38
rs995458003 Chr.8: 130080104-130080104 on Build GRCh38
rs1025789688 Chr.8: 130080046-130080046 on Build GRCh38
rs779646389 Chr.8: 130079904-130079904 on Build GRCh38
rs370155967 Chr.8: 130079861-130079861 on Build GRCh38
rs933430573 Chr.8: 130080077-130080077 on Build GRCh38
rs115908456 Chr.8: 130079862-130079862 on Build GRCh38
rs1055474693 Chr.8: 130080094-130080094 on Build GRCh38
rs770383193 Chr.8: 130080100-130080100 on Build GRCh38
rs35813100 Chr.8: 130080015-130080016 on Build GRCh38
rs538692514 Chr.8: 130079910-130079910 on Build GRCh38
rs990427604 Chr.8: 130080231-130080231 on Build GRCh38
rs551464063 Chr.8: 130079886-130079886 on Build GRCh38
rs772933905 Chr.8: 130079956-130079956 on Build GRCh38
rs1053993411 Chr.8: 130080127-130080127 on Build GRCh38
rs765339752 Chr.8: 130079966-130079966 on Build GRCh38
rs545024575 Chr.8: 130080178-130080178 on Build GRCh38
rs6984045 Chr.8: 130080167-130080167 on Build GRCh38
rs966091400 Chr.8: 130080154-130080154 on Build GRCh38
rs778454635 Chr.8: 130080021-130080021 on Build GRCh38
rs997521215 Chr.8: 130080163-130080163 on Build GRCh38
rs376417360 Chr.8: 130079887-130079887 on Build GRCh38
rs886972103 Chr.8: 130080131-130080131 on Build GRCh38
rs772036962 Chr.8: 130079950-130079950 on Build GRCh38
rs557581643 Chr.8: 130080004-130080004 on Build GRCh38
rs1003984741 Chr.8: 130080143-130080143 on Build GRCh38
rs541114620 Chr.8: 130080075-130080075 on Build GRCh38
rs377386876 Chr.8: 130079856-130079856 on Build GRCh38
rs923994000 Chr.8: 130080098-130080098 on Build GRCh38
rs763932894 Chr.8: 130080018-130080018 on Build GRCh38
rs545784511 Chr.8: 130080265-130080265 on Build GRCh38
rs776946460 Chr.8: 130079962-130079962 on Build GRCh38
rs748103594 Chr.8: 130079928-130079928 on Build GRCh38
rs531256713 Chr.8: 130080209-130080209 on Build GRCh38
rs753198304 Chr.8: 130079875-130079875 on Build GRCh38
rs978220882 Chr.8: 130080086-130080086 on Build GRCh38
rs760488638 Chr.8: 130079957-130079957 on Build GRCh38
rs150967413 Chr.8: 130080064-130080064 on Build GRCh38
rs561515439 Chr.8: 130080289-130080289 on Build GRCh38
rs775685211 Chr.8: 130080101-130080101 on Build GRCh38
rs549091693 Chr.8: 130080273-130080273 on Build GRCh38
rs368803236 Chr.8: 130079986-130079986 on Build GRCh38
rs763427510 Chr.8: 130079975-130079975 on Build GRCh38

View Full Product Details