Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 114902024-114902555 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TNFSF8
Entrez Gene ID
944
Gene Name
tumor necrosis factor superfamily member 8
Gene Aliases
CD153, CD30L, CD30LG, TNLG3A
UniGene
Hs.494901
Gene Chromosome Location
Chr.9: 114893343-114930595 on Build GRCh38
Transcripts
NM_001244.3 NM_001252290.1

SNP Detail

SNP ID Chromosome Location
rs966007984 Chr.9: 114902156-114902156 on Build GRCh38
rs762071843 Chr.9: 114902503-114902503 on Build GRCh38
rs1005935034 Chr.9: 114902444-114902444 on Build GRCh38
rs1040613037 Chr.9: 114902338-114902338 on Build GRCh38
rs957985543 Chr.9: 114902487-114902487 on Build GRCh38
rs760122613 Chr.9: 114902169-114902169 on Build GRCh38
rs966179856 Chr.9: 114902112-114902112 on Build GRCh38
rs982345563 Chr.9: 114902291-114902291 on Build GRCh38
rs567674674 Chr.9: 114902412-114902412 on Build GRCh38
rs901365017 Chr.9: 114902466-114902466 on Build GRCh38
rs552478553 Chr.9: 114902354-114902354 on Build GRCh38
rs3181375 Chr.9: 114902470-114902470 on Build GRCh38
rs371222825 Chr.9: 114902209-114902209 on Build GRCh38
rs868458216 Chr.9: 114902119-114902119 on Build GRCh38
rs916669963 Chr.9: 114902274-114902274 on Build GRCh38
rs77691228 Chr.9: 114902100-114902100 on Build GRCh38
rs570192275 Chr.9: 114902505-114902505 on Build GRCh38
rs1030991168 Chr.9: 114902178-114902178 on Build GRCh38
rs892266887 Chr.9: 114902106-114902106 on Build GRCh38
rs1026521905 Chr.9: 114902408-114902408 on Build GRCh38
rs934482715 Chr.9: 114902262-114902262 on Build GRCh38
rs989530787 Chr.9: 114902229-114902229 on Build GRCh38
rs947888537 Chr.9: 114902051-114902051 on Build GRCh38
rs113570103 Chr.9: 114902377-114902377 on Build GRCh38
rs1008682042 Chr.9: 114902439-114902439 on Build GRCh38
rs540696234 Chr.9: 114902192-114902192 on Build GRCh38
rs763199969 Chr.9: 114902370-114902370 on Build GRCh38
rs928560558 Chr.9: 114902249-114902249 on Build GRCh38
rs955996457 Chr.9: 114902499-114902499 on Build GRCh38
rs930322400 Chr.9: 114902343-114902343 on Build GRCh38
rs1036253407 Chr.9: 114902325-114902325 on Build GRCh38
rs569572380 Chr.9: 114902355-114902355 on Build GRCh38
rs913986257 Chr.9: 114902282-114902282 on Build GRCh38
rs940583213 Chr.9: 114902312-114902312 on Build GRCh38
rs988693010 Chr.9: 114902509-114902509 on Build GRCh38
rs948200051 Chr.9: 114902286-114902286 on Build GRCh38
rs375365206 Chr.9: 114902128-114902128 on Build GRCh38
rs561264795 Chr.9: 114902131-114902131 on Build GRCh38
rs1004834877 Chr.9: 114902091-114902091 on Build GRCh38
rs117673429 Chr.9: 114902293-114902293 on Build GRCh38
rs3181376 Chr.9: 114902461-114902461 on Build GRCh38
rs898857281 Chr.9: 114902095-114902095 on Build GRCh38
rs1031265440 Chr.9: 114902482-114902482 on Build GRCh38
rs888862079 Chr.9: 114902387-114902387 on Build GRCh38
rs1007474254 Chr.9: 114902129-114902129 on Build GRCh38
rs140962100 Chr.9: 114902336-114902336 on Build GRCh38
rs900600944 Chr.9: 114902329-114902329 on Build GRCh38
rs1028714550 Chr.9: 114902101-114902101 on Build GRCh38
rs576242534 Chr.9: 114902092-114902092 on Build GRCh38
rs185002643 Chr.9: 114902071-114902071 on Build GRCh38
rs993655089 Chr.9: 114902396-114902396 on Build GRCh38
rs548167795 Chr.9: 114902401-114902401 on Build GRCh38
rs759239412 Chr.9: 114902210-114902210 on Build GRCh38
rs1044208582 Chr.9: 114902292-114902292 on Build GRCh38
rs914373625 Chr.9: 114902529-114902529 on Build GRCh38
rs892580337 Chr.9: 114902049-114902049 on Build GRCh38
rs981353284 Chr.9: 114902213-114902213 on Build GRCh38
rs2295801 Chr.9: 114902164-114902164 on Build GRCh38
rs774704147 Chr.9: 114902491-114902491 on Build GRCh38
rs906439265 Chr.9: 114902081-114902081 on Build GRCh38
rs62578679 Chr.9: 114902289-114902289 on Build GRCh38

View Full Product Details