Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 30243355-30243870 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAGEB1
Entrez Gene ID
4112
Gene Name
MAGE family member B1
Gene Aliases
CT3.1, DAM10, MAGE-Xp, MAGEL1
UniGene
Hs.73021
Gene Chromosome Location
Chr.X: 30243731-30252038 on Build GRCh38
Transcripts
NM_002363.4 NM_177415.2 NM_002367.3

Gene Symbol
MAGEB4
Entrez Gene ID
4115
Gene Name
MAGE family member B4
Gene Aliases
CT3.6
UniGene
Hs.113291
Gene Chromosome Location
Chr.X: 30241940-30244193 on Build GRCh38
Transcripts
NM_002363.4 NM_177415.2 NM_002367.3

SNP Detail

SNP ID Chromosome Location
rs913011055 Chr.X: 30243486-30243486 on Build GRCh38
rs925791010 Chr.X: 30243417-30243417 on Build GRCh38
rs1041271714 Chr.X: 30243542-30243542 on Build GRCh38
rs879157814 Chr.X: 30243832-30243832 on Build GRCh38
rs571587034 Chr.X: 30243451-30243451 on Build GRCh38
rs949809542 Chr.X: 30243564-30243564 on Build GRCh38
rs898743185 Chr.X: 30243694-30243694 on Build GRCh38
rs1002199287 Chr.X: 30243615-30243615 on Build GRCh38
rs186948407 Chr.X: 30243580-30243580 on Build GRCh38
rs1015391683 Chr.X: 30243760-30243760 on Build GRCh38
rs932937776 Chr.X: 30243613-30243613 on Build GRCh38
rs1003233952 Chr.X: 30243820-30243820 on Build GRCh38
rs934698038 Chr.X: 30243501-30243501 on Build GRCh38
rs905496831 Chr.X: 30243589-30243589 on Build GRCh38
rs1008284741 Chr.X: 30243747-30243747 on Build GRCh38
rs935875480 Chr.X: 30243420-30243420 on Build GRCh38
rs184313512 Chr.X: 30243682-30243682 on Build GRCh38
rs896812164 Chr.X: 30243802-30243802 on Build GRCh38
rs764335827 Chr.X: 30243471-30243471 on Build GRCh38
rs924609317 Chr.X: 30243425-30243425 on Build GRCh38
rs766925306 Chr.X: 30243702-30243702 on Build GRCh38
rs751675977 Chr.X: 30243566-30243569 on Build GRCh38
rs994367668 Chr.X: 30243708-30243708 on Build GRCh38
rs762175884 Chr.X: 30243574-30243574 on Build GRCh38
rs191787003 Chr.X: 30243659-30243659 on Build GRCh38
rs955661774 Chr.X: 30243741-30243741 on Build GRCh38
rs73205834 Chr.X: 30243709-30243709 on Build GRCh38
rs751853154 Chr.X: 30243627-30243627 on Build GRCh38
rs755475254 Chr.X: 30243785-30243787 on Build GRCh38

View Full Product Details