Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 119079350-119079862 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA1210
Entrez Gene ID
57481
Gene Name
KIAA1210
Gene Aliases
-
UniGene
Hs.97594
Gene Chromosome Location
Chr.X: 119078635-119151082 on Build GRCh38
Transcripts
NM_020721.1 XM_006724774.3 XM_017029688.1 XM_017029689.1

SNP Detail

SNP ID Chromosome Location
rs894982398 Chr.X: 119079604-119079604 on Build GRCh38
rs1020763703 Chr.X: 119079549-119079549 on Build GRCh38
rs932301755 Chr.X: 119079463-119079463 on Build GRCh38
rs903269198 Chr.X: 119079556-119079556 on Build GRCh38
rs768141228 Chr.X: 119079828-119079828 on Build GRCh38
rs192017735 Chr.X: 119079423-119079423 on Build GRCh38
rs1033052093 Chr.X: 119079581-119079581 on Build GRCh38
rs985046034 Chr.X: 119079533-119079533 on Build GRCh38
rs998919097 Chr.X: 119079580-119079580 on Build GRCh38
rs145306011 Chr.X: 119079664-119079664 on Build GRCh38
rs1042623712 Chr.X: 119079827-119079827 on Build GRCh38
rs762368154 Chr.X: 119079767-119079767 on Build GRCh38
rs112748559 Chr.X: 119079743-119079743 on Build GRCh38
rs181297838 Chr.X: 119079808-119079808 on Build GRCh38
rs778467393 Chr.X: 119079547-119079547 on Build GRCh38
rs908675341 Chr.X: 119079670-119079670 on Build GRCh38
rs776500902 Chr.X: 119079764-119079764 on Build GRCh38
rs985016356 Chr.X: 119079792-119079792 on Build GRCh38
rs189427984 Chr.X: 119079392-119079392 on Build GRCh38
rs148892485 Chr.X: 119079622-119079622 on Build GRCh38
rs891037036 Chr.X: 119079518-119079518 on Build GRCh38
rs189839571 Chr.X: 119079551-119079551 on Build GRCh38
rs898197233 Chr.X: 119079459-119079459 on Build GRCh38
rs891055980 Chr.X: 119079772-119079772 on Build GRCh38
rs939744754 Chr.X: 119079424-119079424 on Build GRCh38
rs1038050442 Chr.X: 119079425-119079425 on Build GRCh38
rs901590496 Chr.X: 119079730-119079730 on Build GRCh38
rs765270813 Chr.X: 119079793-119079793 on Build GRCh38
rs976268343 Chr.X: 119079480-119079480 on Build GRCh38
rs919657213 Chr.X: 119079753-119079753 on Build GRCh38
rs759152203 Chr.X: 119079731-119079731 on Build GRCh38
rs776343016 Chr.X: 119079694-119079694 on Build GRCh38
rs957409642 Chr.X: 119079612-119079612 on Build GRCh38
rs909181837 Chr.X: 119079542-119079542 on Build GRCh38
rs938806385 Chr.X: 119079574-119079574 on Build GRCh38
rs999134785 Chr.X: 119079678-119079678 on Build GRCh38
rs761927296 Chr.X: 119079774-119079774 on Build GRCh38
rs748841950 Chr.X: 119079394-119079394 on Build GRCh38
rs903804286 Chr.X: 119079670-119079670 on Build GRCh38
rs771817453 Chr.X: 119079602-119079602 on Build GRCh38
rs971647409 Chr.X: 119079641-119079641 on Build GRCh38
rs999209063 Chr.X: 119079413-119079413 on Build GRCh38
rs184484182 Chr.X: 119079505-119079505 on Build GRCh38
rs1049696771 Chr.X: 119079495-119079495 on Build GRCh38

View Full Product Details