Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 123860063-123860559 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
XIAP
Entrez Gene ID
331
Gene Name
X-linked inhibitor of apoptosis
Gene Aliases
API3, BIRC4, IAP-3, ILP1, MIHA, XLP2, hIAP-3, hIAP3
UniGene
Hs.356076
Gene Chromosome Location
Chr.X: 123859812-123913979 on Build GRCh38
Transcripts
NM_001167.3 NM_001204401.1 NR_037916.1 XM_006724754.2 XM_011531329.2

SNP Detail

SNP ID Chromosome Location
rs752335325 Chr.X: 123860137-123860137 on Build GRCh38
rs904220374 Chr.X: 123860511-123860511 on Build GRCh38
rs899119343 Chr.X: 123860106-123860106 on Build GRCh38
rs890339054 Chr.X: 123860474-123860474 on Build GRCh38
rs1008712712 Chr.X: 123860233-123860233 on Build GRCh38
rs940451441 Chr.X: 123860448-123860448 on Build GRCh38
rs1037798613 Chr.X: 123860454-123860454 on Build GRCh38
rs996091304 Chr.X: 123860177-123860177 on Build GRCh38
rs1011124253 Chr.X: 123860429-123860429 on Build GRCh38
rs1019713935 Chr.X: 123860258-123860258 on Build GRCh38
rs777487668 Chr.X: 123860380-123860380 on Build GRCh38
rs920262092 Chr.X: 123860460-123860460 on Build GRCh38
rs777523821 Chr.X: 123860364-123860366 on Build GRCh38
rs751160417 Chr.X: 123860212-123860212 on Build GRCh38
rs999597553 Chr.X: 123860534-123860534 on Build GRCh38
rs931613580 Chr.X: 123860465-123860465 on Build GRCh38
rs1053353200 Chr.X: 123860249-123860249 on Build GRCh38
rs768133907 Chr.X: 123860259-123860259 on Build GRCh38
rs902635325 Chr.X: 123860520-123860520 on Build GRCh38
rs982203371 Chr.X: 123860440-123860440 on Build GRCh38
rs182150836 Chr.X: 123860388-123860388 on Build GRCh38
rs970886041 Chr.X: 123860430-123860430 on Build GRCh38
rs755667889 Chr.X: 123860232-123860232 on Build GRCh38
rs991018044 Chr.X: 123860401-123860401 on Build GRCh38
rs1037553738 Chr.X: 123860157-123860157 on Build GRCh38
rs757670783 Chr.X: 123860231-123860231 on Build GRCh38
rs1032341619 Chr.X: 123860330-123860330 on Build GRCh38
rs1041565183 Chr.X: 123860508-123860508 on Build GRCh38
rs750736414 Chr.X: 123860328-123860328 on Build GRCh38
rs200382204 Chr.X: 123860337-123860337 on Build GRCh38
rs1050280946 Chr.X: 123860180-123860180 on Build GRCh38
rs749263812 Chr.X: 123860434-123860435 on Build GRCh38
rs944530495 Chr.X: 123860507-123860507 on Build GRCh38
rs916800576 Chr.X: 123860406-123860406 on Build GRCh38
rs1050352735 Chr.X: 123860470-123860470 on Build GRCh38
rs907666065 Chr.X: 123860445-123860445 on Build GRCh38

View Full Product Details