Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 135051547-135052043 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM127B
Entrez Gene ID
26071
Gene Name
family with sequence similarity 127 member B
Gene Aliases
CXX1b, MAR8A
UniGene
Hs.460924
Gene Chromosome Location
Chr.X: 135050932-135052191 on Build GRCh38
Transcripts
NM_001078172.1 NM_001134321.1

SNP Detail

SNP ID Chromosome Location
rs766787503 Chr.X: 135051775-135051775 on Build GRCh38
rs760249665 Chr.X: 135051997-135051997 on Build GRCh38
rs778040826 Chr.X: 135051818-135051818 on Build GRCh38
rs778135155 Chr.X: 135051928-135051928 on Build GRCh38
rs202158024 Chr.X: 135051695-135051695 on Build GRCh38
rs771782175 Chr.X: 135051987-135051987 on Build GRCh38
rs758581758 Chr.X: 135051812-135051812 on Build GRCh38
rs774020193 Chr.X: 135051866-135051866 on Build GRCh38
rs1047506130 Chr.X: 135051673-135051673 on Build GRCh38
rs755393015 Chr.X: 135051896-135051896 on Build GRCh38
rs368118052 Chr.X: 135051834-135051834 on Build GRCh38
rs753081824 Chr.X: 135051908-135051908 on Build GRCh38
rs201721834 Chr.X: 135051811-135051811 on Build GRCh38
rs867161086 Chr.X: 135051994-135051994 on Build GRCh38
rs768283470 Chr.X: 135051865-135051865 on Build GRCh38
rs772786999 Chr.X: 135051992-135051992 on Build GRCh38
rs376649703 Chr.X: 135051973-135051973 on Build GRCh38
rs886099824 Chr.X: 135051713-135051713 on Build GRCh38
rs777226889 Chr.X: 135052005-135052005 on Build GRCh38
rs755234360 Chr.X: 135051787-135051787 on Build GRCh38
rs763396568 Chr.X: 135051971-135051971 on Build GRCh38
rs749121395 Chr.X: 135051844-135051844 on Build GRCh38
rs754300787 Chr.X: 135051785-135051785 on Build GRCh38
rs766918867 Chr.X: 135051886-135051886 on Build GRCh38
rs766971377 Chr.X: 135051767-135051767 on Build GRCh38
rs779210625 Chr.X: 135051800-135051800 on Build GRCh38
rs1038862127 Chr.X: 135051626-135051626 on Build GRCh38
rs754476110 Chr.X: 135051822-135051822 on Build GRCh38
rs913879082 Chr.X: 135051567-135051567 on Build GRCh38
rs765819241 Chr.X: 135052019-135052019 on Build GRCh38
rs770301029 Chr.X: 135052004-135052004 on Build GRCh38
rs762651220 Chr.X: 135051737-135051737 on Build GRCh38
rs943278908 Chr.X: 135051625-135051625 on Build GRCh38
rs774177879 Chr.X: 135051979-135051979 on Build GRCh38
rs765478668 Chr.X: 135051900-135051900 on Build GRCh38
rs960936476 Chr.X: 135051905-135051905 on Build GRCh38
rs2498776 Chr.X: 135051967-135051967 on Build GRCh38
rs761491585 Chr.X: 135051871-135051871 on Build GRCh38
rs775410910 Chr.X: 135051569-135051569 on Build GRCh38
rs775224705 Chr.X: 135051839-135051839 on Build GRCh38
rs979756010 Chr.X: 135051573-135051573 on Build GRCh38
rs754244413 Chr.X: 135051888-135051888 on Build GRCh38
rs773815748 Chr.X: 135051763-135051763 on Build GRCh38
rs754913310 Chr.X: 135051771-135051771 on Build GRCh38
rs767132415 Chr.X: 135051875-135051875 on Build GRCh38
rs933051905 Chr.X: 135051644-135051644 on Build GRCh38
rs772795477 Chr.X: 135051930-135051930 on Build GRCh38
rs760101362 Chr.X: 135051884-135051884 on Build GRCh38
rs925153773 Chr.X: 135051620-135051620 on Build GRCh38
rs372367356 Chr.X: 135051969-135051969 on Build GRCh38
rs760003240 Chr.X: 135052007-135052007 on Build GRCh38
rs775412817 Chr.X: 135051733-135051733 on Build GRCh38
rs901699957 Chr.X: 135051641-135051641 on Build GRCh38
rs375655465 Chr.X: 135051823-135051823 on Build GRCh38
rs763805000 Chr.X: 135051753-135051753 on Build GRCh38
rs992644711 Chr.X: 135051907-135051907 on Build GRCh38
rs898723335 Chr.X: 135051822-135051822 on Build GRCh38
rs772770497 Chr.X: 135051880-135051880 on Build GRCh38
rs771859549 Chr.X: 135051924-135051924 on Build GRCh38
rs747922476 Chr.X: 135051986-135051986 on Build GRCh38
rs778881173 Chr.X: 135051801-135051801 on Build GRCh38
rs756658939 Chr.X: 135051772-135051772 on Build GRCh38

View Full Product Details