Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 70052463-70052952 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
H2AFY2
Entrez Gene ID
55506
Gene Name
H2A histone family member Y2
Gene Aliases
macroH2A2
UniGene
Hs.499953
Gene Chromosome Location
Chr.10: 70052601-70112284 on Build GRCh38
Transcripts
NM_018649.2

SNP Detail

SNP ID Chromosome Location
rs1030680077 Chr.10: 70052512-70052512 on Build GRCh38
rs965747730 Chr.10: 70052825-70052825 on Build GRCh38
rs575407613 Chr.10: 70052575-70052575 on Build GRCh38
rs926788902 Chr.10: 70052605-70052605 on Build GRCh38
rs74139255 Chr.10: 70052556-70052556 on Build GRCh38
rs1007394245 Chr.10: 70052792-70052792 on Build GRCh38
rs978485094 Chr.10: 70052882-70052882 on Build GRCh38
rs927418471 Chr.10: 70052634-70052634 on Build GRCh38
rs554076664 Chr.10: 70052855-70052855 on Build GRCh38
rs1042357240 Chr.10: 70052664-70052664 on Build GRCh38
rs753247266 Chr.10: 70052849-70052850 on Build GRCh38
rs537937830 Chr.10: 70052593-70052593 on Build GRCh38
rs985983923 Chr.10: 70052492-70052492 on Build GRCh38
rs763873719 Chr.10: 70052713-70052713 on Build GRCh38
rs1049792512 Chr.10: 70052576-70052576 on Build GRCh38
rs1034104055 Chr.10: 70052897-70052897 on Build GRCh38
rs78134463 Chr.10: 70052684-70052684 on Build GRCh38
rs1019984690 Chr.10: 70052847-70052847 on Build GRCh38
rs958867600 Chr.10: 70052924-70052924 on Build GRCh38
rs968442447 Chr.10: 70052860-70052860 on Build GRCh38
rs930879026 Chr.10: 70052573-70052573 on Build GRCh38
rs1018831092 Chr.10: 70052814-70052814 on Build GRCh38
rs965820967 Chr.10: 70052543-70052543 on Build GRCh38
rs900308400 Chr.10: 70052827-70052827 on Build GRCh38
rs533114583 Chr.10: 70052498-70052498 on Build GRCh38
rs978905598 Chr.10: 70052629-70052629 on Build GRCh38
rs1030212375 Chr.10: 70052829-70052829 on Build GRCh38
rs890401744 Chr.10: 70052833-70052833 on Build GRCh38
rs747505955 Chr.10: 70052900-70052900 on Build GRCh38
rs1039775311 Chr.10: 70052817-70052817 on Build GRCh38
rs941479132 Chr.10: 70052761-70052761 on Build GRCh38
rs757957028 Chr.10: 70052612-70052612 on Build GRCh38
rs917498175 Chr.10: 70052660-70052660 on Build GRCh38
rs371719882 Chr.10: 70052783-70052784 on Build GRCh38
rs953348629 Chr.10: 70052721-70052721 on Build GRCh38
rs937524737 Chr.10: 70052641-70052641 on Build GRCh38
rs918922076 Chr.10: 70052558-70052558 on Build GRCh38
rs35395876 Chr.10: 70052563-70052563 on Build GRCh38
rs953351209 Chr.10: 70052487-70052487 on Build GRCh38
rs777945849 Chr.10: 70052882-70052883 on Build GRCh38
rs888513870 Chr.10: 70052725-70052725 on Build GRCh38
rs929987886 Chr.10: 70052661-70052661 on Build GRCh38
rs1000092612 Chr.10: 70052674-70052674 on Build GRCh38
rs572356889 Chr.10: 70052927-70052927 on Build GRCh38
rs549720741 Chr.10: 70052595-70052595 on Build GRCh38
rs926856122 Chr.10: 70052843-70052843 on Build GRCh38
rs1009902336 Chr.10: 70052839-70052839 on Build GRCh38
rs911629473 Chr.10: 70052542-70052542 on Build GRCh38
rs996412450 Chr.10: 70052828-70052828 on Build GRCh38
rs114931839 Chr.10: 70052650-70052650 on Build GRCh38
rs903865322 Chr.10: 70052667-70052667 on Build GRCh38
rs988891138 Chr.10: 70052565-70052565 on Build GRCh38
rs918091856 Chr.10: 70052920-70052920 on Build GRCh38
rs984497380 Chr.10: 70052837-70052837 on Build GRCh38

View Full Product Details