Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 102834680-102835174 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP17A1
Entrez Gene ID
1586
Gene Name
cytochrome P450 family 17 subfamily A member 1
Gene Aliases
CPT7, CYP17, P450C17, S17AH
UniGene
Hs.438016
Gene Chromosome Location
Chr.10: 102830531-102837533 on Build GRCh38
Transcripts
NM_000102.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6128135 c.601T>C p.Y201H Chr.10: 102834850-102834850 on Build GRCh38
5580751 c.656C>T p.P219L Chr.10: 102834795-102834795 on Build GRCh38
127756 c.462T>C p.C154C Chr.10: 102834989-102834989 on Build GRCh38
6064773 c.468G>T p.M156I Chr.10: 102834983-102834983 on Build GRCh38
1718557 c.496G>A p.D166N Chr.10: 102834955-102834955 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1004467 Chr.10: 102834750-102834750 on Build GRCh38
rs995317924 Chr.10: 102835088-102835088 on Build GRCh38
rs71471253 Chr.10: 102835066-102835066 on Build GRCh38
rs950945162 Chr.10: 102835096-102835096 on Build GRCh38
rs765148467 Chr.10: 102834840-102834840 on Build GRCh38
rs769089279 Chr.10: 102834877-102834877 on Build GRCh38
rs142435666 Chr.10: 102834823-102834823 on Build GRCh38
rs926270668 Chr.10: 102835140-102835140 on Build GRCh38
rs58822002 Chr.10: 102834996-102834996 on Build GRCh38
rs752540777 Chr.10: 102834930-102834930 on Build GRCh38
rs743575 Chr.10: 102835149-102835149 on Build GRCh38
rs770597902 Chr.10: 102835005-102835005 on Build GRCh38
rs767247558 Chr.10: 102834803-102834803 on Build GRCh38
rs764087679 Chr.10: 102835065-102835065 on Build GRCh38
rs776597678 Chr.10: 102834782-102834782 on Build GRCh38
rs745927932 Chr.10: 102834964-102834964 on Build GRCh38
rs769287572 Chr.10: 102834976-102834976 on Build GRCh38
rs754941252 Chr.10: 102834834-102834834 on Build GRCh38
rs974261500 Chr.10: 102835037-102835037 on Build GRCh38
rs747064195 Chr.10: 102834763-102834763 on Build GRCh38
rs375350911 Chr.10: 102834797-102834797 on Build GRCh38
rs758354727 Chr.10: 102834934-102834934 on Build GRCh38
rs879802265 Chr.10: 102834791-102834791 on Build GRCh38
rs533771699 Chr.10: 102834722-102834722 on Build GRCh38
rs760333930 Chr.10: 102834887-102834887 on Build GRCh38
rs556134667 Chr.10: 102834828-102834828 on Build GRCh38
rs776190569 Chr.10: 102835026-102835026 on Build GRCh38
rs980336663 Chr.10: 102835127-102835127 on Build GRCh38
rs752593169 Chr.10: 102835040-102835045 on Build GRCh38
rs369081864 Chr.10: 102834929-102834929 on Build GRCh38
rs758206006 Chr.10: 102835060-102835072 on Build GRCh38
rs775909420 Chr.10: 102834795-102834795 on Build GRCh38
rs751361358 Chr.10: 102834945-102834945 on Build GRCh38
rs778700597 Chr.10: 102834734-102834734 on Build GRCh38
rs575801994 Chr.10: 102834844-102834844 on Build GRCh38
rs1026422207 Chr.10: 102835091-102835091 on Build GRCh38
rs752518511 Chr.10: 102834842-102834842 on Build GRCh38
rs148082634 Chr.10: 102834923-102834923 on Build GRCh38
rs749751359 Chr.10: 102834974-102834974 on Build GRCh38
rs7922856 Chr.10: 102835108-102835108 on Build GRCh38
rs775901707 Chr.10: 102834874-102834874 on Build GRCh38
rs577311150 Chr.10: 102835114-102835114 on Build GRCh38
rs777854065 Chr.10: 102835061-102835062 on Build GRCh38
rs774849280 Chr.10: 102834984-102834984 on Build GRCh38
rs200063521 Chr.10: 102834805-102834805 on Build GRCh38
rs970242783 Chr.10: 102835112-102835112 on Build GRCh38
rs76228098 Chr.10: 102834794-102834794 on Build GRCh38
rs565377365 Chr.10: 102835101-102835101 on Build GRCh38
rs375364403 Chr.10: 102834875-102834875 on Build GRCh38
rs762422304 Chr.10: 102834770-102834770 on Build GRCh38
rs957752302 Chr.10: 102835148-102835148 on Build GRCh38
rs745991883 Chr.10: 102834863-102834863 on Build GRCh38
rs780472449 Chr.10: 102835125-102835125 on Build GRCh38
rs104894150 Chr.10: 102834850-102834850 on Build GRCh38
rs369805604 Chr.10: 102835075-102835075 on Build GRCh38
rs373613340 Chr.10: 102835056-102835056 on Build GRCh38
rs755443 Chr.10: 102835097-102835097 on Build GRCh38
rs766199096 Chr.10: 102834894-102834894 on Build GRCh38
rs558207891 Chr.10: 102834731-102834731 on Build GRCh38
rs61754273 Chr.10: 102834861-102834861 on Build GRCh38
rs45583734 Chr.10: 102835071-102835071 on Build GRCh38
rs760032797 Chr.10: 102834783-102834783 on Build GRCh38
rs770270001 Chr.10: 102834785-102834785 on Build GRCh38
rs942302876 Chr.10: 102835084-102835086 on Build GRCh38
rs776193931 Chr.10: 102834901-102834901 on Build GRCh38
rs777576356 Chr.10: 102834845-102834845 on Build GRCh38
rs764602894 Chr.10: 102835046-102835046 on Build GRCh38
rs945233088 Chr.10: 102834919-102834919 on Build GRCh38
rs750052005 Chr.10: 102834813-102834813 on Build GRCh38
rs774995068 Chr.10: 102835062-102835062 on Build GRCh38
rs770311570 Chr.10: 102834868-102834868 on Build GRCh38
rs757454955 Chr.10: 102834958-102834958 on Build GRCh38
rs746879000 Chr.10: 102835067-102835067 on Build GRCh38
rs772270950 Chr.10: 102834737-102834737 on Build GRCh38
rs903593909 Chr.10: 102835090-102835090 on Build GRCh38
rs781316969 Chr.10: 102834961-102834961 on Build GRCh38
rs759268771 Chr.10: 102835041-102835041 on Build GRCh38
rs767962487 Chr.10: 102834968-102834968 on Build GRCh38
rs200231398 Chr.10: 102834881-102834881 on Build GRCh38
rs779250805 Chr.10: 102835036-102835037 on Build GRCh38
rs762813674 Chr.10: 102835064-102835064 on Build GRCh38
rs372323467 Chr.10: 102834862-102834862 on Build GRCh38
rs141821705 Chr.10: 102834967-102834967 on Build GRCh38
rs748584208 Chr.10: 102834994-102834994 on Build GRCh38
rs760421916 Chr.10: 102834816-102834816 on Build GRCh38

View Full Product Details