Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 123688988-123689488 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR26
Entrez Gene ID
2849
Gene Name
G protein-coupled receptor 26
Gene Aliases
-
UniGene
Hs.12751
Gene Chromosome Location
Chr.10: 123666355-123697399 on Build GRCh38
Transcripts
NM_153442.3

SNP Detail

SNP ID Chromosome Location
rs1045211182 Chr.10: 123689272-123689272 on Build GRCh38
rs986226217 Chr.10: 123689198-123689198 on Build GRCh38
rs906807241 Chr.10: 123689277-123689277 on Build GRCh38
rs551901673 Chr.10: 123689113-123689113 on Build GRCh38
rs79489240 Chr.10: 123689267-123689267 on Build GRCh38
rs567865256 Chr.10: 123689269-123689269 on Build GRCh38
rs905335277 Chr.10: 123689283-123689283 on Build GRCh38
rs58170593 Chr.10: 123689266-123689266 on Build GRCh38
rs986008629 Chr.10: 123689338-123689338 on Build GRCh38
rs533180126 Chr.10: 123689020-123689020 on Build GRCh38
rs972023898 Chr.10: 123689139-123689139 on Build GRCh38
rs901189756 Chr.10: 123689212-123689212 on Build GRCh38
rs12414023 Chr.10: 123689229-123689229 on Build GRCh38
rs976883447 Chr.10: 123689419-123689419 on Build GRCh38
rs935308570 Chr.10: 123689228-123689228 on Build GRCh38
rs528651386 Chr.10: 123689294-123689294 on Build GRCh38
rs192389959 Chr.10: 123689325-123689325 on Build GRCh38
rs933241735 Chr.10: 123689187-123689188 on Build GRCh38
rs528199231 Chr.10: 123689268-123689268 on Build GRCh38
rs539481321 Chr.10: 123689339-123689339 on Build GRCh38
rs969107346 Chr.10: 123689032-123689032 on Build GRCh38
rs187724815 Chr.10: 123689024-123689024 on Build GRCh38
rs1002649377 Chr.10: 123689070-123689070 on Build GRCh38
rs954678881 Chr.10: 123689458-123689458 on Build GRCh38
rs766981850 Chr.10: 123689254-123689254 on Build GRCh38
rs878944588 Chr.10: 123689254-123689255 on Build GRCh38
rs981786524 Chr.10: 123689057-123689057 on Build GRCh38
rs1033672106 Chr.10: 123689080-123689080 on Build GRCh38
rs140176525 Chr.10: 123689064-123689064 on Build GRCh38
rs945587333 Chr.10: 123689236-123689236 on Build GRCh38
rs942765689 Chr.10: 123689183-123689183 on Build GRCh38
rs1219821 Chr.10: 123689329-123689329 on Build GRCh38
rs965842148 Chr.10: 123689417-123689417 on Build GRCh38
rs371532141 Chr.10: 123689252-123689253 on Build GRCh38
rs398054812 Chr.10: 123689265-123689266 on Build GRCh38
rs796237624 Chr.10: 123689253-123689253 on Build GRCh38
rs759604109 Chr.10: 123689163-123689163 on Build GRCh38
rs866696842 Chr.10: 123689328-123689328 on Build GRCh38
rs1035159697 Chr.10: 123689314-123689314 on Build GRCh38
rs1015653550 Chr.10: 123689114-123689114 on Build GRCh38
rs993557725 Chr.10: 123689327-123689327 on Build GRCh38
rs58984643 Chr.10: 123689253-123689254 on Build GRCh38
rs922663215 Chr.10: 123689455-123689455 on Build GRCh38
rs745809556 Chr.10: 123689266-123689267 on Build GRCh38
rs376538154 Chr.10: 123689401-123689401 on Build GRCh38
rs952930411 Chr.10: 123689082-123689082 on Build GRCh38
rs532350281 Chr.10: 123689442-123689442 on Build GRCh38
rs371207434 Chr.10: 123689205-123689205 on Build GRCh38
rs1055756434 Chr.10: 123689303-123689303 on Build GRCh38
rs920489795 Chr.10: 123689145-123689145 on Build GRCh38
rs910396918 Chr.10: 123689364-123689364 on Build GRCh38
rs765236269 Chr.10: 123689169-123689169 on Build GRCh38
rs78410128 Chr.10: 123689266-123689266 on Build GRCh38
rs930492367 Chr.10: 123689149-123689149 on Build GRCh38

View Full Product Details