Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 809998-810490 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPLP2
Entrez Gene ID
6181
Gene Name
ribosomal protein lateral stalk subunit P2
Gene Aliases
D11S2243E, LP2, P2, RPP2
UniGene
Hs.437594
Gene Chromosome Location
Chr.11: 809936-812876 on Build GRCh38
Transcripts
NM_001004.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6769288 c.91G>T p.G31C Chr.11: 810325-810325 on Build GRCh38
3720491 c.100G>T p.A34S Chr.11: 810334-810334 on Build GRCh38
6421271 c.21C>A p.Y7* Chr.11: 810255-810255 on Build GRCh38
1746604 c.97G>A p.E33K Chr.11: 810331-810331 on Build GRCh38
6413347 c.2T>C p.M1T Chr.11: 810236-810236 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758999487 Chr.11: 810374-810374 on Build GRCh38
rs755880156 Chr.11: 810388-810388 on Build GRCh38
rs1045650514 Chr.11: 810161-810161 on Build GRCh38
rs758070555 Chr.11: 810209-810209 on Build GRCh38
rs780975281 Chr.11: 810339-810339 on Build GRCh38
rs1013714002 Chr.11: 810240-810240 on Build GRCh38
rs560855971 Chr.11: 810212-810212 on Build GRCh38
rs780607024 Chr.11: 810218-810218 on Build GRCh38
rs145622642 Chr.11: 810251-810251 on Build GRCh38
rs866990660 Chr.11: 810060-810060 on Build GRCh38
rs367573432 Chr.11: 810367-810367 on Build GRCh38
rs780328939 Chr.11: 810268-810268 on Build GRCh38
rs774923485 Chr.11: 810293-810293 on Build GRCh38
rs546666558 Chr.11: 810249-810249 on Build GRCh38
rs552951679 Chr.11: 810460-810460 on Build GRCh38
rs752237180 Chr.11: 810259-810259 on Build GRCh38
rs200336941 Chr.11: 810267-810267 on Build GRCh38
rs749273033 Chr.11: 810353-810353 on Build GRCh38
rs549711977 Chr.11: 810105-810105 on Build GRCh38
rs1803013 Chr.11: 810315-810315 on Build GRCh38
rs769624915 Chr.11: 810292-810292 on Build GRCh38
rs749797199 Chr.11: 810219-810219 on Build GRCh38
rs745663573 Chr.11: 810342-810342 on Build GRCh38
rs200920555 Chr.11: 810256-810256 on Build GRCh38
rs146929030 Chr.11: 810113-810113 on Build GRCh38
rs920355012 Chr.11: 810128-810128 on Build GRCh38
rs1001051325 Chr.11: 810435-810435 on Build GRCh38
rs751189347 Chr.11: 810300-810300 on Build GRCh38
rs111850318 Chr.11: 810305-810305 on Build GRCh38
rs17155725 Chr.11: 810019-810027 on Build GRCh38
rs771021879 Chr.11: 810371-810371 on Build GRCh38
rs878963307 Chr.11: 810232-810233 on Build GRCh38
rs201228279 Chr.11: 810370-810370 on Build GRCh38
rs879358846 Chr.11: 810158-810158 on Build GRCh38
rs1013373191 Chr.11: 810020-810020 on Build GRCh38
rs756707581 Chr.11: 810330-810330 on Build GRCh38
rs775375372 Chr.11: 810352-810352 on Build GRCh38
rs138243875 Chr.11: 810318-810318 on Build GRCh38
rs1020907453 Chr.11: 810054-810054 on Build GRCh38
rs11540443 Chr.11: 810016-810016 on Build GRCh38
rs878978501 Chr.11: 810325-810325 on Build GRCh38
rs370622305 Chr.11: 810243-810243 on Build GRCh38
rs552937310 Chr.11: 810025-810025 on Build GRCh38
rs538751053 Chr.11: 810168-810168 on Build GRCh38
rs142278470 Chr.11: 810319-810319 on Build GRCh38
rs930422716 Chr.11: 810134-810134 on Build GRCh38
rs761392767 Chr.11: 810303-810303 on Build GRCh38
rs976230677 Chr.11: 810154-810154 on Build GRCh38
rs763750323 Chr.11: 810263-810263 on Build GRCh38
rs774314279 Chr.11: 810229-810229 on Build GRCh38
rs752327022 Chr.11: 810202-810202 on Build GRCh38
rs1053542138 Chr.11: 810452-810452 on Build GRCh38
rs781498380 Chr.11: 810211-810211 on Build GRCh38
rs200161310 Chr.11: 810345-810345 on Build GRCh38
rs544855806 Chr.11: 810091-810091 on Build GRCh38
rs889654519 Chr.11: 810461-810461 on Build GRCh38
rs544374482 Chr.11: 810147-810147 on Build GRCh38
rs759443353 Chr.11: 810255-810255 on Build GRCh38
rs757444355 Chr.11: 810377-810377 on Build GRCh38
rs929496679 Chr.11: 810162-810162 on Build GRCh38
rs763633895 Chr.11: 810298-810298 on Build GRCh38
rs955639423 Chr.11: 810056-810056 on Build GRCh38
rs768757273 Chr.11: 810221-810221 on Build GRCh38
rs768582692 Chr.11: 810405-810405 on Build GRCh38
rs190451004 Chr.11: 810100-810100 on Build GRCh38
rs140116545 Chr.11: 810244-810244 on Build GRCh38
rs768104043 Chr.11: 810354-810354 on Build GRCh38
rs756527968 Chr.11: 810216-810216 on Build GRCh38
rs910581196 Chr.11: 810138-810138 on Build GRCh38
rs770740675 Chr.11: 810288-810288 on Build GRCh38
rs772331823 Chr.11: 810282-810282 on Build GRCh38
rs772424395 Chr.11: 810239-810239 on Build GRCh38
rs199757717 Chr.11: 810286-810286 on Build GRCh38
rs888013713 Chr.11: 810170-810170 on Build GRCh38
rs879158068 Chr.11: 810361-810361 on Build GRCh38
rs776315521 Chr.11: 810291-810291 on Build GRCh38
rs527404049 Chr.11: 810089-810090 on Build GRCh38
rs1042507562 Chr.11: 810426-810426 on Build GRCh38
rs532925478 Chr.11: 810199-810200 on Build GRCh38
rs753238870 Chr.11: 810373-810373 on Build GRCh38
rs1001497843 Chr.11: 810207-810207 on Build GRCh38
rs537273414 Chr.11: 810385-810385 on Build GRCh38
rs754156104 Chr.11: 810312-810312 on Build GRCh38
rs986936477 Chr.11: 810302-810302 on Build GRCh38
rs974381460 Chr.11: 810116-810116 on Build GRCh38
rs896226891 Chr.11: 810019-810019 on Build GRCh38
rs878889845 Chr.11: 810358-810359 on Build GRCh38
rs773850872 Chr.11: 810407-810407 on Build GRCh38
rs771091638 Chr.11: 810335-810335 on Build GRCh38
rs755467714 Chr.11: 810273-810273 on Build GRCh38
rs1032234131 Chr.11: 810101-810101 on Build GRCh38
rs374810591 Chr.11: 810438-810438 on Build GRCh38
rs902241828 Chr.11: 810430-810430 on Build GRCh38
rs367869473 Chr.11: 810223-810223 on Build GRCh38
rs779844757 Chr.11: 810396-810396 on Build GRCh38
rs572844964 Chr.11: 810034-810034 on Build GRCh38
rs764233512 Chr.11: 810376-810376 on Build GRCh38
rs536340668 Chr.11: 810441-810441 on Build GRCh38
rs779066946 Chr.11: 810277-810277 on Build GRCh38
rs202225033 Chr.11: 810397-810397 on Build GRCh38
rs373702898 Chr.11: 810362-810362 on Build GRCh38
rs748338928 Chr.11: 810234-810234 on Build GRCh38
rs572766158 Chr.11: 810369-810369 on Build GRCh38
rs529786599 Chr.11: 810227-810227 on Build GRCh38
rs764696182 Chr.11: 810184-810184 on Build GRCh38
rs944778328 Chr.11: 810149-810149 on Build GRCh38
rs147750535 Chr.11: 810279-810279 on Build GRCh38
rs986076468 Chr.11: 810123-810123 on Build GRCh38
rs758525359 Chr.11: 810329-810329 on Build GRCh38
rs907630539 Chr.11: 810398-810398 on Build GRCh38
rs905786333 Chr.11: 810205-810205 on Build GRCh38
rs762444786 Chr.11: 810297-810297 on Build GRCh38
rs747052106 Chr.11: 810279-810281 on Build GRCh38
rs996355892 Chr.11: 810199-810199 on Build GRCh38
rs778140990 Chr.11: 810285-810285 on Build GRCh38

View Full Product Details